Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87952023-87952296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTEN
Entrez Gene ID
5728
Gene Name
phosphatase and tensin homolog
Gene Aliases
10q23del, BZS, CWS1, DEC, GLM2, MHAM, MMAC1, PTEN1, TEP1
UniGene
Hs.500466
Gene Chromosome Location
Chr.10: 87863438-87971930 on Build GRCh38
Transcripts
NM_000314.6 NM_001304717.2 NM_001304718.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7339121 c.528_528delT p.Y177fs*6 Chr.10: 87952153-87952153 on Build GRCh38
6975907 c.583T>C p.F195L Chr.10: 87952208-87952208 on Build GRCh38
6959267 c.520T>C p.Y174H Chr.10: 87952145-87952145 on Build GRCh38
4920 c.567_571delACCAG p.R189fs*11 Chr.10: 87952192-87952196 on Build GRCh38
4165560 c.611C>A p.P204Q Chr.10: 87952236-87952236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951327461 Chr.10: 87952044-87952044 on Build GRCh38
rs192026853 Chr.10: 87952050-87952050 on Build GRCh38
rs1043121029 Chr.10: 87952066-87952066 on Build GRCh38
rs786204943 Chr.10: 87952185-87952185 on Build GRCh38
rs121909221 Chr.10: 87952135-87952135 on Build GRCh38
rs904398124 Chr.10: 87952069-87952069 on Build GRCh38
rs750907391 Chr.10: 87952076-87952077 on Build GRCh38
rs372870950 Chr.10: 87952055-87952055 on Build GRCh38
rs876659456 Chr.10: 87952210-87952211 on Build GRCh38
rs727504114 Chr.10: 87952261-87952261 on Build GRCh38
rs992603618 Chr.10: 87952047-87952047 on Build GRCh38
rs786201867 Chr.10: 87952147-87952147 on Build GRCh38
rs786204944 Chr.10: 87952229-87952229 on Build GRCh38
rs606231170 Chr.10: 87952189-87952189 on Build GRCh38
rs780943695 Chr.10: 87952121-87952121 on Build GRCh38
rs786204890 Chr.10: 87952172-87952174 on Build GRCh38
rs786204864 Chr.10: 87952136-87952136 on Build GRCh38
rs794729664 Chr.10: 87952170-87952170 on Build GRCh38
rs878853012 Chr.10: 87952156-87952156 on Build GRCh38
rs764273954 Chr.10: 87952072-87952072 on Build GRCh38
rs397514559 Chr.10: 87952125-87952125 on Build GRCh38
rs568851024 Chr.10: 87952204-87952204 on Build GRCh38
rs786204867 Chr.10: 87952224-87952224 on Build GRCh38
rs587781895 Chr.10: 87952176-87952176 on Build GRCh38
rs765083317 Chr.10: 87952076-87952076 on Build GRCh38
rs587776673 Chr.10: 87952132-87952132 on Build GRCh38
rs757498880 Chr.10: 87952152-87952152 on Build GRCh38
rs761470435 Chr.10: 87952074-87952074 on Build GRCh38
rs786204865 Chr.10: 87952137-87952137 on Build GRCh38
rs765433422 Chr.10: 87952250-87952250 on Build GRCh38
rs746280047 Chr.10: 87952163-87952163 on Build GRCh38
rs138336847 Chr.10: 87952264-87952264 on Build GRCh38
rs539074063 Chr.10: 87952237-87952237 on Build GRCh38
rs776857453 Chr.10: 87952073-87952073 on Build GRCh38
rs587782603 Chr.10: 87952118-87952118 on Build GRCh38
rs876660507 Chr.10: 87952134-87952134 on Build GRCh38
rs786204862 Chr.10: 87952117-87952117 on Build GRCh38
rs776763121 Chr.10: 87952238-87952238 on Build GRCh38
rs786204901 Chr.10: 87952210-87952210 on Build GRCh38
rs374478092 Chr.10: 87952198-87952198 on Build GRCh38
rs587781538 Chr.10: 87952221-87952221 on Build GRCh38
rs786204863 Chr.10: 87952119-87952119 on Build GRCh38
rs878853941 Chr.10: 87952170-87952171 on Build GRCh38
rs587780711 Chr.10: 87952151-87952153 on Build GRCh38
rs370162160 Chr.10: 87952249-87952249 on Build GRCh38
rs779349848 Chr.10: 87952162-87952162 on Build GRCh38
rs587776670 Chr.10: 87952211-87952211 on Build GRCh38
rs766292424 Chr.10: 87952085-87952085 on Build GRCh38
rs747072478 Chr.10: 87952219-87952219 on Build GRCh38
rs786204866 Chr.10: 87952158-87952158 on Build GRCh38
rs750267449 Chr.10: 87952078-87952078 on Build GRCh38
rs397515374 Chr.10: 87952159-87952160 on Build GRCh38
rs864622341 Chr.10: 87952146-87952146 on Build GRCh38
rs587782473 Chr.10: 87952223-87952223 on Build GRCh38
rs121913293 Chr.10: 87952142-87952142 on Build GRCh38
rs945711198 Chr.10: 87952065-87952065 on Build GRCh38
rs756623620 Chr.10: 87952098-87952099 on Build GRCh38
rs757799618 Chr.10: 87952180-87952180 on Build GRCh38
rs104894184 Chr.10: 87952159-87952159 on Build GRCh38
rs121913294 Chr.10: 87952143-87952143 on Build GRCh38
rs758341794 Chr.10: 87952079-87952079 on Build GRCh38
rs786202733 Chr.10: 87952165-87952165 on Build GRCh38
rs876659227 Chr.10: 87952174-87952174 on Build GRCh38
rs772631069 Chr.10: 87952202-87952202 on Build GRCh38
rs587781784 Chr.10: 87952116-87952116 on Build GRCh38
rs121909232 Chr.10: 87952258-87952258 on Build GRCh38
rs786202773 Chr.10: 87952243-87952243 on Build GRCh38
rs550267891 Chr.10: 87952087-87952087 on Build GRCh38
rs587782316 Chr.10: 87952145-87952145 on Build GRCh38
rs786204868 Chr.10: 87952235-87952235 on Build GRCh38

View Full Product Details