Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42130364-42130623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB5C
Entrez Gene ID
5878
Gene Name
RAB5C, member RAS oncogene family
Gene Aliases
L1880, RAB5CL, RAB5L, RABL
UniGene
Hs.650382
Gene Chromosome Location
Chr.17: 42124976-42155044 on Build GRCh38
Transcripts
NM_001252039.1 NM_004583.3 NM_201434.2 XM_011525090.1 XM_011525091.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6761598 c.118C>T p.R40C Chr.17: 42130385-42130385 on Build GRCh38
4066518 c.112G>A p.V38I Chr.17: 42130391-42130391 on Build GRCh38
3770608 c.92C>T p.A31V Chr.17: 42130411-42130411 on Build GRCh38
6523536 c.3G>T p.M1I Chr.17: 42130500-42130500 on Build GRCh38
3517624 c.89C>T p.S30F Chr.17: 42130414-42130414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757930639 Chr.17: 42130593-42130593 on Build GRCh38
rs201855065 Chr.17: 42130492-42130492 on Build GRCh38
rs749662869 Chr.17: 42130438-42130438 on Build GRCh38
rs759380944 Chr.17: 42130539-42130539 on Build GRCh38
rs781707295 Chr.17: 42130410-42130410 on Build GRCh38
rs759435852 Chr.17: 42130463-42130463 on Build GRCh38
rs755324556 Chr.17: 42130497-42130497 on Build GRCh38
rs187972504 Chr.17: 42130537-42130537 on Build GRCh38
rs560209770 Chr.17: 42130536-42130536 on Build GRCh38
rs774625517 Chr.17: 42130546-42130546 on Build GRCh38
rs370734734 Chr.17: 42130533-42130533 on Build GRCh38
rs771533462 Chr.17: 42130448-42130448 on Build GRCh38
rs750913164 Chr.17: 42130479-42130479 on Build GRCh38
rs968864221 Chr.17: 42130494-42130494 on Build GRCh38
rs756552315 Chr.17: 42130484-42130484 on Build GRCh38
rs563186025 Chr.17: 42130477-42130477 on Build GRCh38
rs751777474 Chr.17: 42130471-42130471 on Build GRCh38
rs1021354841 Chr.17: 42130488-42130488 on Build GRCh38
rs754165479 Chr.17: 42130495-42130495 on Build GRCh38
rs745516376 Chr.17: 42130502-42130502 on Build GRCh38
rs73311653 Chr.17: 42130455-42130455 on Build GRCh38
rs924960833 Chr.17: 42130411-42130411 on Build GRCh38
rs746412388 Chr.17: 42130413-42130413 on Build GRCh38
rs760778533 Chr.17: 42130575-42130575 on Build GRCh38
rs777485845 Chr.17: 42130529-42130529 on Build GRCh38
rs768234973 Chr.17: 42130440-42130440 on Build GRCh38
rs761919578 Chr.17: 42130474-42130474 on Build GRCh38
rs199902477 Chr.17: 42130508-42130508 on Build GRCh38
rs771607685 Chr.17: 42130517-42130517 on Build GRCh38
rs144263947 Chr.17: 42130468-42130468 on Build GRCh38
rs761997455 Chr.17: 42130569-42130569 on Build GRCh38
rs780450538 Chr.17: 42130433-42130433 on Build GRCh38
rs779213765 Chr.17: 42130580-42130580 on Build GRCh38
rs751617564 Chr.17: 42130385-42130385 on Build GRCh38
rs11542397 Chr.17: 42130541-42130541 on Build GRCh38
rs941942730 Chr.17: 42130515-42130528 on Build GRCh38
rs376067518 Chr.17: 42130507-42130507 on Build GRCh38
rs767789812 Chr.17: 42130573-42130573 on Build GRCh38
rs750431886 Chr.17: 42130574-42130574 on Build GRCh38
rs773171823 Chr.17: 42130452-42130452 on Build GRCh38
rs1014328871 Chr.17: 42130585-42130585 on Build GRCh38
rs754394450 Chr.17: 42130577-42130577 on Build GRCh38
rs778465129 Chr.17: 42130446-42130446 on Build GRCh38
rs202098551 Chr.17: 42130392-42130392 on Build GRCh38
rs776510757 Chr.17: 42130458-42130458 on Build GRCh38
rs752383662 Chr.17: 42130583-42130583 on Build GRCh38
rs955928227 Chr.17: 42130532-42130532 on Build GRCh38
rs756462674 Chr.17: 42130425-42130425 on Build GRCh38
rs567303604 Chr.17: 42130578-42130578 on Build GRCh38

View Full Product Details