Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43415667-43415892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TP53BP1
Entrez Gene ID
7158
Gene Name
tumor protein p53 binding protein 1
Gene Aliases
53BP1, TDRD30, TP53, p202, p53BP1
UniGene
Hs.440968
Gene Chromosome Location
Chr.15: 43403064-43510509 on Build GRCh38
Transcripts
NM_001141979.1 NM_001141980.1 NM_005657.2 XM_011521985.1 XM_011521986.2 XM_017022533.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6569675 c.4966C>T p.R1656* Chr.15: 43415717-43415717 on Build GRCh38
6806558 c.4911C>T p.N1637N Chr.15: 43415772-43415772 on Build GRCh38
961971 c.4921G>A p.A1641T Chr.15: 43415747-43415747 on Build GRCh38
6538278 c.4997T>C p.M1666T Chr.15: 43415686-43415686 on Build GRCh38
961970 c.4970C>T p.P1657L Chr.15: 43415698-43415698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758859555 Chr.15: 43415777-43415777 on Build GRCh38
rs776632909 Chr.15: 43415821-43415821 on Build GRCh38
rs759267622 Chr.15: 43415842-43415842 on Build GRCh38
rs376828160 Chr.15: 43415864-43415864 on Build GRCh38
rs140309161 Chr.15: 43415771-43415771 on Build GRCh38
rs377703429 Chr.15: 43415752-43415752 on Build GRCh38
rs753325618 Chr.15: 43415858-43415858 on Build GRCh38
rs767106305 Chr.15: 43415696-43415696 on Build GRCh38
rs374006962 Chr.15: 43415813-43415813 on Build GRCh38
rs774183732 Chr.15: 43415691-43415691 on Build GRCh38
rs775203698 Chr.15: 43415847-43415847 on Build GRCh38
rs1002151907 Chr.15: 43415702-43415702 on Build GRCh38
rs201180017 Chr.15: 43415756-43415756 on Build GRCh38
rs747204356 Chr.15: 43415814-43415814 on Build GRCh38
rs760299459 Chr.15: 43415854-43415854 on Build GRCh38
rs118147882 Chr.15: 43415857-43415857 on Build GRCh38
rs1034600964 Chr.15: 43415743-43415743 on Build GRCh38
rs767220200 Chr.15: 43415845-43415845 on Build GRCh38
rs770897067 Chr.15: 43415824-43415824 on Build GRCh38
rs369627426 Chr.15: 43415757-43415757 on Build GRCh38
rs768781465 Chr.15: 43415817-43415817 on Build GRCh38
rs373841537 Chr.15: 43415695-43415695 on Build GRCh38
rs752303589 Chr.15: 43415750-43415750 on Build GRCh38
rs774292410 Chr.15: 43415837-43415837 on Build GRCh38
rs749201688 Chr.15: 43415823-43415823 on Build GRCh38
rs746192779 Chr.15: 43415775-43415775 on Build GRCh38
rs878966132 Chr.15: 43415748-43415748 on Build GRCh38
rs1020489968 Chr.15: 43415872-43415872 on Build GRCh38

View Full Product Details