Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 158891218-158891443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR60
Entrez Gene ID
55112
Gene Name
WD repeat domain 60
Gene Aliases
FAP163, SRPS6, SRTD8
UniGene
Hs.389945
Gene Chromosome Location
Chr.7: 158839250-158958695 on Build GRCh38
Transcripts
NM_018051.4 XM_005249549.1 XM_006716041.2 XM_011516367.1 XM_011516368.2 XM_011516369.2 XM_017012381.1 XM_017012382.1 XM_017012383.1 XM_017012384.1 XM_017012385.1 XM_017012386.1 XM_017012387.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4741015 c.1048G>A p.E350K Chr.7: 158891322-158891322 on Build GRCh38
6109392 c.998A>G p.H333R Chr.7: 158891272-158891272 on Build GRCh38
6869457 c.1057G>A p.V353M Chr.7: 158891331-158891331 on Build GRCh38
1449721 c.1000G>A p.E334K Chr.7: 158891274-158891274 on Build GRCh38
7280334 c.1040C>T p.P347L Chr.7: 158891314-158891314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747741412 Chr.7: 158891383-158891383 on Build GRCh38
rs201852398 Chr.7: 158891321-158891321 on Build GRCh38
rs749260080 Chr.7: 158891370-158891370 on Build GRCh38
rs202014035 Chr.7: 158891315-158891315 on Build GRCh38
rs528993084 Chr.7: 158891330-158891330 on Build GRCh38
rs979284175 Chr.7: 158891388-158891389 on Build GRCh38
rs1006424601 Chr.7: 158891291-158891291 on Build GRCh38
rs767982901 Chr.7: 158891372-158891372 on Build GRCh38
rs964449939 Chr.7: 158891405-158891405 on Build GRCh38
rs112908949 Chr.7: 158891347-158891347 on Build GRCh38
rs1017678902 Chr.7: 158891406-158891406 on Build GRCh38
rs769768860 Chr.7: 158891301-158891301 on Build GRCh38
rs200061970 Chr.7: 158891322-158891322 on Build GRCh38
rs745805834 Chr.7: 158891295-158891295 on Build GRCh38
rs780973838 Chr.7: 158891292-158891292 on Build GRCh38
rs919078942 Chr.7: 158891248-158891248 on Build GRCh38
rs28584840 Chr.7: 158891368-158891368 on Build GRCh38
rs374394657 Chr.7: 158891257-158891257 on Build GRCh38
rs183547176 Chr.7: 158891317-158891317 on Build GRCh38
rs748753193 Chr.7: 158891313-158891313 on Build GRCh38
rs976919044 Chr.7: 158891408-158891408 on Build GRCh38
rs758617099 Chr.7: 158891269-158891269 on Build GRCh38
rs779914446 Chr.7: 158891369-158891369 on Build GRCh38
rs201126878 Chr.7: 158891274-158891274 on Build GRCh38
rs764250579 Chr.7: 158891337-158891337 on Build GRCh38
rs780955388 Chr.7: 158891350-158891350 on Build GRCh38
rs902794954 Chr.7: 158891329-158891329 on Build GRCh38
rs752873400 Chr.7: 158891266-158891266 on Build GRCh38
rs781003265 Chr.7: 158891346-158891346 on Build GRCh38
rs778551596 Chr.7: 158891377-158891377 on Build GRCh38
rs73529717 Chr.7: 158891314-158891314 on Build GRCh38
rs886100816 Chr.7: 158891277-158891277 on Build GRCh38
rs367589644 Chr.7: 158891356-158891356 on Build GRCh38
rs976029909 Chr.7: 158891245-158891245 on Build GRCh38
rs193215755 Chr.7: 158891392-158891392 on Build GRCh38
rs201238212 Chr.7: 158891331-158891331 on Build GRCh38
rs576071107 Chr.7: 158891238-158891238 on Build GRCh38
rs756947785 Chr.7: 158891283-158891283 on Build GRCh38
rs777820131 Chr.7: 158891273-158891273 on Build GRCh38
rs140137568 Chr.7: 158891306-158891306 on Build GRCh38
rs1032478409 Chr.7: 158891360-158891360 on Build GRCh38
rs968728887 Chr.7: 158891387-158891387 on Build GRCh38

View Full Product Details