Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149367319-149367560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCYOX1L
Entrez Gene ID
78991
Gene Name
prenylcysteine oxidase 1 like
Gene Aliases
-
UniGene
Hs.644397
Gene Chromosome Location
Chr.5: 149358007-149375112 on Build GRCh38
Transcripts
NM_001301054.1 NM_001301057.1 NM_024028.3 XM_011537680.1 XM_011537681.1 XM_011537682.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1634059 c.750T>A p.C250* Chr.5: 149367427-149367427 on Build GRCh38
6007171 c.749G>A p.C250Y Chr.5: 149367426-149367426 on Build GRCh38
3365916 c.810C>A p.T270T Chr.5: 149367487-149367487 on Build GRCh38
6377462 c.821C>A p.T274K Chr.5: 149367498-149367498 on Build GRCh38
5474788 c.823+10C>T p.? Chr.5: 149367510-149367510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757380710 Chr.5: 149367512-149367512 on Build GRCh38
rs764424388 Chr.5: 149367371-149367381 on Build GRCh38
rs761317238 Chr.5: 149367485-149367485 on Build GRCh38
rs759973337 Chr.5: 149367338-149367338 on Build GRCh38
rs747914525 Chr.5: 149367538-149367538 on Build GRCh38
rs754260788 Chr.5: 149367507-149367507 on Build GRCh38
rs558974556 Chr.5: 149367456-149367456 on Build GRCh38
rs775751017 Chr.5: 149367347-149367347 on Build GRCh38
rs753935868 Chr.5: 149367354-149367354 on Build GRCh38
rs938242434 Chr.5: 149367482-149367482 on Build GRCh38
rs775986270 Chr.5: 149367479-149367479 on Build GRCh38
rs201278534 Chr.5: 149367448-149367448 on Build GRCh38
rs751855957 Chr.5: 149367428-149367429 on Build GRCh38
rs754781532 Chr.5: 149367418-149367418 on Build GRCh38
rs372542183 Chr.5: 149367349-149367349 on Build GRCh38
rs200328826 Chr.5: 149367534-149367534 on Build GRCh38
rs377373868 Chr.5: 149367431-149367431 on Build GRCh38
rs768065702 Chr.5: 149367468-149367468 on Build GRCh38
rs748148118 Chr.5: 149367429-149367429 on Build GRCh38
rs746028890 Chr.5: 149367416-149367416 on Build GRCh38
rs187849573 Chr.5: 149367375-149367375 on Build GRCh38
rs764329879 Chr.5: 149367350-149367350 on Build GRCh38
rs375616927 Chr.5: 149367515-149367515 on Build GRCh38
rs369997493 Chr.5: 149367516-149367516 on Build GRCh38
rs533718703 Chr.5: 149367430-149367430 on Build GRCh38
rs759201042 Chr.5: 149367463-149367463 on Build GRCh38
rs748797626 Chr.5: 149367454-149367454 on Build GRCh38
rs767941098 Chr.5: 149367342-149367342 on Build GRCh38
rs779229030 Chr.5: 149367405-149367405 on Build GRCh38
rs763551144 Chr.5: 149367445-149367445 on Build GRCh38
rs142537559 Chr.5: 149367423-149367423 on Build GRCh38
rs764632833 Chr.5: 149367500-149367500 on Build GRCh38
rs781020744 Chr.5: 149367537-149367537 on Build GRCh38
rs369557809 Chr.5: 149367474-149367474 on Build GRCh38
rs757462073 Chr.5: 149367377-149367377 on Build GRCh38

View Full Product Details