Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80221512-80221783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC26A11
Entrez Gene ID
284129
Gene Name
solute carrier family 26 member 11
Gene Aliases
-
UniGene
Hs.4866
Gene Chromosome Location
Chr.17: 80220287-80253509 on Build GRCh38
Transcripts
NM_001166347.1 NM_001166348.1 NM_001166349.1 NM_173626.3 XM_006721833.1 XM_011524652.1 XM_011524653.1 XM_011524654.1 XM_017024505.1 XM_017024506.1 XM_017024507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
985571 c.218C>A p.A73D Chr.17: 80221778-80221778 on Build GRCh38
5913941 c.131C>T p.S44F Chr.17: 80221691-80221691 on Build GRCh38
2803368 c.13G>A p.V5M Chr.17: 80221573-80221573 on Build GRCh38
4894923 c.57C>T p.A19A Chr.17: 80221617-80221617 on Build GRCh38
6780922 c.198G>T p.A66A Chr.17: 80221758-80221758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200412027 Chr.17: 80221629-80221629 on Build GRCh38
rs375394500 Chr.17: 80221757-80221757 on Build GRCh38
rs754796731 Chr.17: 80221762-80221762 on Build GRCh38
rs749266673 Chr.17: 80221568-80221568 on Build GRCh38
rs780138047 Chr.17: 80221745-80221745 on Build GRCh38
rs768102743 Chr.17: 80221676-80221676 on Build GRCh38
rs373471904 Chr.17: 80221668-80221668 on Build GRCh38
rs528825807 Chr.17: 80221620-80221620 on Build GRCh38
rs746824309 Chr.17: 80221586-80221586 on Build GRCh38
rs777226815 Chr.17: 80221698-80221698 on Build GRCh38
rs774697312 Chr.17: 80221604-80221604 on Build GRCh38
rs866220843 Chr.17: 80221758-80221758 on Build GRCh38
rs368234615 Chr.17: 80221733-80221733 on Build GRCh38
rs761226870 Chr.17: 80221599-80221599 on Build GRCh38
rs561848151 Chr.17: 80221580-80221580 on Build GRCh38
rs764858387 Chr.17: 80221601-80221601 on Build GRCh38
rs752883092 Chr.17: 80221559-80221559 on Build GRCh38
rs551429137 Chr.17: 80221673-80221673 on Build GRCh38
rs768042728 Chr.17: 80221612-80221612 on Build GRCh38
rs986227818 Chr.17: 80221624-80221624 on Build GRCh38
rs372017696 Chr.17: 80221720-80221720 on Build GRCh38
rs750461173 Chr.17: 80221724-80221724 on Build GRCh38
rs774948597 Chr.17: 80221565-80221567 on Build GRCh38
rs142733492 Chr.17: 80221747-80221747 on Build GRCh38
rs760446324 Chr.17: 80221541-80221541 on Build GRCh38
rs758365577 Chr.17: 80221644-80221644 on Build GRCh38
rs771963387 Chr.17: 80221540-80221540 on Build GRCh38
rs888376925 Chr.17: 80221691-80221691 on Build GRCh38
rs926725408 Chr.17: 80221671-80221671 on Build GRCh38
rs941955741 Chr.17: 80221717-80221717 on Build GRCh38
rs146795725 Chr.17: 80221622-80221622 on Build GRCh38
rs779092910 Chr.17: 80221641-80221641 on Build GRCh38
rs1030532951 Chr.17: 80221572-80221572 on Build GRCh38
rs753351051 Chr.17: 80221545-80221545 on Build GRCh38
rs1046170087 Chr.17: 80221640-80221640 on Build GRCh38
rs765404032 Chr.17: 80221722-80221722 on Build GRCh38
rs766730336 Chr.17: 80221551-80221551 on Build GRCh38
rs370502982 Chr.17: 80221664-80221664 on Build GRCh38
rs754153623 Chr.17: 80221623-80221623 on Build GRCh38
rs760128984 Chr.17: 80221608-80221608 on Build GRCh38
rs1030841823 Chr.17: 80221763-80221763 on Build GRCh38
rs752456721 Chr.17: 80221539-80221539 on Build GRCh38
rs745969268 Chr.17: 80221643-80221643 on Build GRCh38
rs200345707 Chr.17: 80221555-80221555 on Build GRCh38
rs761402151 Chr.17: 80221547-80221547 on Build GRCh38
rs531057270 Chr.17: 80221558-80221558 on Build GRCh38
rs199781652 Chr.17: 80221693-80221693 on Build GRCh38
rs768366152 Chr.17: 80221594-80221594 on Build GRCh38
rs760084288 Chr.17: 80221668-80221685 on Build GRCh38
rs763873938 Chr.17: 80221542-80221542 on Build GRCh38
rs1031981022 Chr.17: 80221626-80221626 on Build GRCh38
rs897619139 Chr.17: 80221540-80221540 on Build GRCh38
rs76464091 Chr.17: 80221648-80221648 on Build GRCh38
rs757264826 Chr.17: 80221635-80221635 on Build GRCh38
rs561639116 Chr.17: 80221598-80221598 on Build GRCh38
rs775389353 Chr.17: 80221583-80221583 on Build GRCh38
rs776410925 Chr.17: 80221663-80221663 on Build GRCh38
rs753007585 Chr.17: 80221619-80221619 on Build GRCh38
rs148454524 Chr.17: 80221694-80221694 on Build GRCh38
rs949620294 Chr.17: 80221665-80221665 on Build GRCh38
rs756288564 Chr.17: 80221565-80221565 on Build GRCh38
rs759183117 Chr.17: 80221534-80221534 on Build GRCh38
rs144562896 Chr.17: 80221686-80221686 on Build GRCh38
rs549799845 Chr.17: 80221581-80221581 on Build GRCh38
rs771043425 Chr.17: 80221574-80221574 on Build GRCh38
rs768641516 Chr.17: 80221659-80221659 on Build GRCh38

View Full Product Details