Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38081660-38081893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIPA1L3
Entrez Gene ID
23094
Gene Name
signal induced proliferation associated 1 like 3
Gene Aliases
CTRCT45, SPAL3, SPAR3
UniGene
Hs.655502
Gene Chromosome Location
Chr.19: 37906823-38208372 on Build GRCh38
Transcripts
NM_015073.2 XM_005258671.4 XM_011526657.2 XM_017026517.1 XM_017026518.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1287927 c.320G>T p.G107V Chr.19: 38081885-38081885 on Build GRCh38
6778752 c.223C>T p.P75S Chr.19: 38081788-38081788 on Build GRCh38
5547778 c.89_100del12 p.T31_Y34delTGDY Chr.19: 38081654-38081665 on Build GRCh38
4726158 c.285G>A p.E95E Chr.19: 38081850-38081850 on Build GRCh38
4726157 c.157A>C p.S53R Chr.19: 38081722-38081722 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779147500 Chr.19: 38081834-38081834 on Build GRCh38
rs530042626 Chr.19: 38081742-38081742 on Build GRCh38
rs772021253 Chr.19: 38081761-38081761 on Build GRCh38
rs71354964 Chr.19: 38081735-38081735 on Build GRCh38
rs746621772 Chr.19: 38081761-38081762 on Build GRCh38
rs569252662 Chr.19: 38081739-38081740 on Build GRCh38
rs371571630 Chr.19: 38081727-38081727 on Build GRCh38
rs761656220 Chr.19: 38081684-38081684 on Build GRCh38
rs746943953 Chr.19: 38081867-38081867 on Build GRCh38
rs754135148 Chr.19: 38081817-38081817 on Build GRCh38
rs564479011 Chr.19: 38081728-38081739 on Build GRCh38
rs750466221 Chr.19: 38081727-38081728 on Build GRCh38
rs760130502 Chr.19: 38081711-38081711 on Build GRCh38
rs748167678 Chr.19: 38081757-38081757 on Build GRCh38
rs372329721 Chr.19: 38081803-38081803 on Build GRCh38
rs780440383 Chr.19: 38081745-38081745 on Build GRCh38
rs548172526 Chr.19: 38081786-38081786 on Build GRCh38
rs747927445 Chr.19: 38081848-38081848 on Build GRCh38
rs753429177 Chr.19: 38081719-38081719 on Build GRCh38
rs768566893 Chr.19: 38081752-38081752 on Build GRCh38
rs1038301677 Chr.19: 38081773-38081773 on Build GRCh38
rs764361441 Chr.19: 38081733-38081733 on Build GRCh38
rs562186095 Chr.19: 38081734-38081739 on Build GRCh38
rs750904543 Chr.19: 38081805-38081805 on Build GRCh38
rs768141191 Chr.19: 38081802-38081802 on Build GRCh38
rs777744055 Chr.19: 38081859-38081859 on Build GRCh38
rs994241132 Chr.19: 38081783-38081783 on Build GRCh38
rs376863637 Chr.19: 38081853-38081853 on Build GRCh38
rs756194614 Chr.19: 38081728-38081733 on Build GRCh38
rs950567154 Chr.19: 38081804-38081804 on Build GRCh38
rs774409136 Chr.19: 38081678-38081678 on Build GRCh38
rs955935278 Chr.19: 38081828-38081828 on Build GRCh38
rs773096326 Chr.19: 38081777-38081777 on Build GRCh38
rs766566162 Chr.19: 38081811-38081811 on Build GRCh38
rs902069290 Chr.19: 38081696-38081696 on Build GRCh38
rs375968684 Chr.19: 38081734-38081734 on Build GRCh38
rs201480085 Chr.19: 38081718-38081718 on Build GRCh38
rs71354966 Chr.19: 38081753-38081753 on Build GRCh38
rs749461630 Chr.19: 38081751-38081751 on Build GRCh38
rs767657585 Chr.19: 38081726-38081727 on Build GRCh38
rs770467951 Chr.19: 38081869-38081869 on Build GRCh38
rs1006840238 Chr.19: 38081762-38081762 on Build GRCh38
rs981072768 Chr.19: 38081799-38081799 on Build GRCh38
rs151071468 Chr.19: 38081827-38081827 on Build GRCh38
rs779378262 Chr.19: 38081752-38081753 on Build GRCh38
rs890078136 Chr.19: 38081746-38081746 on Build GRCh38
rs113889464 Chr.19: 38081747-38081747 on Build GRCh38
rs891772280 Chr.19: 38081758-38081758 on Build GRCh38
rs201746414 Chr.19: 38081697-38081697 on Build GRCh38
rs781743909 Chr.19: 38081739-38081739 on Build GRCh38
rs757550778 Chr.19: 38081725-38081726 on Build GRCh38
rs71354965 Chr.19: 38081741-38081741 on Build GRCh38
rs368966925 Chr.19: 38081785-38081785 on Build GRCh38
rs764565234 Chr.19: 38081782-38081782 on Build GRCh38
rs776331340 Chr.19: 38081781-38081781 on Build GRCh38
rs772688597 Chr.19: 38081703-38081703 on Build GRCh38
rs563320778 Chr.19: 38081800-38081800 on Build GRCh38
rs1035360929 Chr.19: 38081818-38081818 on Build GRCh38
rs756323883 Chr.19: 38081809-38081809 on Build GRCh38
rs746533364 Chr.19: 38081778-38081778 on Build GRCh38
rs771652567 Chr.19: 38081693-38081693 on Build GRCh38

View Full Product Details