Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26794887-26795148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIGV
Entrez Gene ID
55650
Gene Name
phosphatidylinositol glycan anchor biosynthesis class V
Gene Aliases
GPI-MT-II, HPMRS1, PIG-V
UniGene
Hs.259605
Gene Chromosome Location
Chr.1: 26787963-26798403 on Build GRCh38
Transcripts
NM_001202554.1 NM_017837.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4031036 c.856C>T p.R286W Chr.1: 26794890-26794890 on Build GRCh38
7263886 c.940G>A p.V314I Chr.1: 26794974-26794974 on Build GRCh38
7177457 c.1058G>T p.C353F Chr.1: 26795092-26795092 on Build GRCh38
5846230 c.1111G>A p.D371N Chr.1: 26795145-26795145 on Build GRCh38
3487596 c.1109C>T p.P370L Chr.1: 26795143-26795143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs796445275 Chr.1: 26795080-26795080 on Build GRCh38
rs201084787 Chr.1: 26795007-26795007 on Build GRCh38
rs35943598 Chr.1: 26794984-26794984 on Build GRCh38
rs746747719 Chr.1: 26795064-26795064 on Build GRCh38
rs768513225 Chr.1: 26794944-26794944 on Build GRCh38
rs144053014 Chr.1: 26795077-26795077 on Build GRCh38
rs772709756 Chr.1: 26794968-26794968 on Build GRCh38
rs753991047 Chr.1: 26795037-26795037 on Build GRCh38
rs148610857 Chr.1: 26794993-26794993 on Build GRCh38
rs199833588 Chr.1: 26794978-26794978 on Build GRCh38
rs376257064 Chr.1: 26795118-26795118 on Build GRCh38
rs904668661 Chr.1: 26795123-26795123 on Build GRCh38
rs758104099 Chr.1: 26794937-26794937 on Build GRCh38
rs774037693 Chr.1: 26795125-26795125 on Build GRCh38
rs370558069 Chr.1: 26794954-26794954 on Build GRCh38
rs748890495 Chr.1: 26795116-26795116 on Build GRCh38
rs764422244 Chr.1: 26795033-26795033 on Build GRCh38
rs768120577 Chr.1: 26795070-26795070 on Build GRCh38
rs769517639 Chr.1: 26794966-26794966 on Build GRCh38
rs988022455 Chr.1: 26795101-26795101 on Build GRCh38
rs774991514 Chr.1: 26795002-26795002 on Build GRCh38
rs139073416 Chr.1: 26795056-26795056 on Build GRCh38
rs772977347 Chr.1: 26795079-26795079 on Build GRCh38
rs746942271 Chr.1: 26794940-26794940 on Build GRCh38
rs147229452 Chr.1: 26794913-26794913 on Build GRCh38
rs997028891 Chr.1: 26795020-26795020 on Build GRCh38
rs555597231 Chr.1: 26794967-26794967 on Build GRCh38
rs1057515437 Chr.1: 26795042-26795042 on Build GRCh38
rs373315999 Chr.1: 26795004-26795004 on Build GRCh38
rs1010432758 Chr.1: 26795114-26795114 on Build GRCh38
rs754929675 Chr.1: 26795066-26795066 on Build GRCh38
rs763425078 Chr.1: 26794975-26794975 on Build GRCh38
rs142884955 Chr.1: 26795014-26795014 on Build GRCh38
rs756422269 Chr.1: 26794910-26794910 on Build GRCh38
rs756227754 Chr.1: 26795030-26795030 on Build GRCh38
rs780094407 Chr.1: 26794939-26794939 on Build GRCh38
rs781067583 Chr.1: 26795069-26795069 on Build GRCh38
rs769166258 Chr.1: 26795073-26795073 on Build GRCh38
rs771852496 Chr.1: 26794930-26794930 on Build GRCh38
rs1052062433 Chr.1: 26795067-26795067 on Build GRCh38
rs1057518270 Chr.1: 26794925-26794925 on Build GRCh38
rs758278375 Chr.1: 26795040-26795040 on Build GRCh38
rs746610869 Chr.1: 26794975-26794980 on Build GRCh38
rs200014772 Chr.1: 26794912-26794912 on Build GRCh38

View Full Product Details