Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 151338133-151338406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED12L
Entrez Gene ID
116931
Gene Name
mediator complex subunit 12 like
Gene Aliases
NOPAR, TNRC11L, TRALP, TRALPUSH
UniGene
Hs.744234
Gene Chromosome Location
Chr.3: 151085665-151436677 on Build GRCh38
Transcripts
NM_053002.5 XM_006713487.3 XM_011512390.2 XM_011512394.2 XM_011512399.2 XM_017005676.1 XM_017005677.1 XM_017005678.1 XM_017005679.1 XM_017005680.1 NM_022788.4 NM_176876.2 XM_017007069.1

Gene Symbol
P2RY12
Entrez Gene ID
64805
Gene Name
purinergic receptor P2Y12
Gene Aliases
ADPG-R, BDPLT8, HORK3, P2T(AC), P2Y(12)R, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12, SP1999
UniGene
Hs.591281
Gene Chromosome Location
Chr.3: 151336843-151384812 on Build GRCh38
Transcripts
NM_053002.5 XM_006713487.3 XM_011512390.2 XM_011512394.2 XM_011512399.2 XM_017005676.1 XM_017005677.1 XM_017005678.1 XM_017005679.1 XM_017005680.1 NM_022788.4 NM_176876.2 XM_017007069.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6736468 c.660C>T p.Y220Y Chr.3: 151338186-151338186 on Build GRCh38
6736469 c.650A>G p.Y217C Chr.3: 151338196-151338196 on Build GRCh38
6481589 c.595T>A p.W199R Chr.3: 151338251-151338251 on Build GRCh38
3003420 c.458T>C p.F153S Chr.3: 151338388-151338388 on Build GRCh38
4420873 c.597G>C p.W199C Chr.3: 151338249-151338249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773010680 Chr.3: 151338228-151338228 on Build GRCh38
rs767970539 Chr.3: 151338173-151338173 on Build GRCh38
rs976417425 Chr.3: 151338298-151338298 on Build GRCh38
rs370983746 Chr.3: 151338226-151338226 on Build GRCh38
rs774480391 Chr.3: 151338350-151338350 on Build GRCh38
rs148966366 Chr.3: 151338178-151338178 on Build GRCh38
rs751218750 Chr.3: 151338303-151338303 on Build GRCh38
rs754485666 Chr.3: 151338307-151338307 on Build GRCh38
rs199746754 Chr.3: 151338374-151338374 on Build GRCh38
rs781058297 Chr.3: 151338219-151338219 on Build GRCh38
rs756074078 Chr.3: 151338336-151338336 on Build GRCh38
rs758020400 Chr.3: 151338194-151338194 on Build GRCh38
rs746513347 Chr.3: 151338206-151338206 on Build GRCh38
rs755439126 Chr.3: 151338217-151338217 on Build GRCh38
rs764709900 Chr.3: 151338369-151338369 on Build GRCh38
rs760773017 Chr.3: 151338367-151338367 on Build GRCh38
rs753306510 Chr.3: 151338265-151338265 on Build GRCh38
rs746084742 Chr.3: 151338354-151338354 on Build GRCh38
rs770759132 Chr.3: 151338348-151338348 on Build GRCh38
rs976052391 Chr.3: 151338275-151338275 on Build GRCh38
rs943960274 Chr.3: 151338246-151338246 on Build GRCh38
rs911824376 Chr.3: 151338193-151338193 on Build GRCh38
rs533622869 Chr.3: 151338174-151338174 on Build GRCh38
rs774623268 Chr.3: 151338240-151338240 on Build GRCh38
rs923308300 Chr.3: 151338285-151338285 on Build GRCh38
rs368508157 Chr.3: 151338345-151338345 on Build GRCh38
rs754207964 Chr.3: 151338186-151338186 on Build GRCh38
rs535856604 Chr.3: 151338249-151338249 on Build GRCh38
rs775744997 Chr.3: 151338365-151338365 on Build GRCh38
rs879192182 Chr.3: 151338259-151338259 on Build GRCh38
rs951857095 Chr.3: 151338175-151338175 on Build GRCh38
rs148095261 Chr.3: 151338222-151338222 on Build GRCh38
rs759605986 Chr.3: 151338171-151338171 on Build GRCh38
rs779688496 Chr.3: 151338204-151338204 on Build GRCh38
rs762694917 Chr.3: 151338229-151338229 on Build GRCh38
rs375189871 Chr.3: 151338248-151338248 on Build GRCh38
rs749142270 Chr.3: 151338346-151338346 on Build GRCh38
rs771075348 Chr.3: 151338235-151338235 on Build GRCh38
rs878872418 Chr.3: 151338258-151338258 on Build GRCh38
rs780654645 Chr.3: 151338311-151338311 on Build GRCh38
rs16863320 Chr.3: 151338300-151338300 on Build GRCh38
rs143655651 Chr.3: 151338205-151338205 on Build GRCh38
rs566389626 Chr.3: 151338358-151338358 on Build GRCh38
rs768198990 Chr.3: 151338212-151338212 on Build GRCh38
rs112155006 Chr.3: 151338177-151338177 on Build GRCh38
rs761286961 Chr.3: 151338289-151338289 on Build GRCh38
rs146203653 Chr.3: 151338378-151338378 on Build GRCh38
rs747604391 Chr.3: 151338333-151338333 on Build GRCh38
rs769622362 Chr.3: 151338227-151338227 on Build GRCh38
rs764657007 Chr.3: 151338299-151338299 on Build GRCh38

View Full Product Details