Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75503987-75504232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRR1
Entrez Gene ID
11103
Gene Name
KRR1, small subunit processome component homolog
Gene Aliases
HRB2, RIP-1
UniGene
Hs.645517
Gene Chromosome Location
Chr.12: 75497639-75511638 on Build GRCh38
Transcripts
NM_007043.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2102509 c.712A>T p.S238C Chr.12: 75504023-75504023 on Build GRCh38
1242446 c.736T>G p.F246V Chr.12: 75503999-75503999 on Build GRCh38
6433973 c.681G>T p.E227D Chr.12: 75504054-75504054 on Build GRCh38
4655621 c.666A>C p.L222F Chr.12: 75504069-75504069 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs892211632 Chr.12: 75504101-75504101 on Build GRCh38
rs747031287 Chr.12: 75504015-75504016 on Build GRCh38
rs762416932 Chr.12: 75504097-75504097 on Build GRCh38
rs367586880 Chr.12: 75504078-75504078 on Build GRCh38
rs901056089 Chr.12: 75504032-75504032 on Build GRCh38
rs749302630 Chr.12: 75504075-75504075 on Build GRCh38
rs571236018 Chr.12: 75504176-75504176 on Build GRCh38
rs1009305113 Chr.12: 75504120-75504120 on Build GRCh38
rs997029796 Chr.12: 75504156-75504156 on Build GRCh38
rs781710988 Chr.12: 75504073-75504074 on Build GRCh38
rs767374508 Chr.12: 75504048-75504048 on Build GRCh38
rs763693574 Chr.12: 75504080-75504081 on Build GRCh38
rs372563816 Chr.12: 75504042-75504042 on Build GRCh38
rs188885620 Chr.12: 75504031-75504031 on Build GRCh38
rs565535318 Chr.12: 75504084-75504084 on Build GRCh38
rs773580860 Chr.12: 75504076-75504077 on Build GRCh38
rs376593401 Chr.12: 75504055-75504055 on Build GRCh38
rs745463656 Chr.12: 75504080-75504080 on Build GRCh38
rs765352026 Chr.12: 75504099-75504099 on Build GRCh38
rs551345271 Chr.12: 75504143-75504143 on Build GRCh38
rs1023303797 Chr.12: 75504086-75504086 on Build GRCh38
rs777000919 Chr.12: 75504096-75504096 on Build GRCh38
rs372122256 Chr.12: 75504079-75504079 on Build GRCh38
rs768705624 Chr.12: 75504044-75504044 on Build GRCh38
rs770108836 Chr.12: 75504075-75504076 on Build GRCh38
rs536612778 Chr.12: 75504190-75504190 on Build GRCh38
rs375763606 Chr.12: 75504089-75504089 on Build GRCh38
rs201035934 Chr.12: 75504068-75504068 on Build GRCh38
rs759146099 Chr.12: 75504077-75504078 on Build GRCh38
rs771733195 Chr.12: 75504081-75504081 on Build GRCh38
rs1005257012 Chr.12: 75504082-75504082 on Build GRCh38
rs75910030 Chr.12: 75504126-75504126 on Build GRCh38
rs748618258 Chr.12: 75504074-75504075 on Build GRCh38

View Full Product Details