Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8288969-8289240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANGRF
Entrez Gene ID
29098
Gene Name
RAN guanine nucleotide release factor
Gene Aliases
HSPC165, HSPC236, MOG1, RANGNRF
UniGene
Hs.408233
Gene Chromosome Location
Chr.17: 8287150-8290091 on Build GRCh38
Transcripts
NM_001177801.1 NM_001177802.1 NM_016492.4 XM_005256618.4 NM_001320871.1 NM_001320872.1 NM_201520.2 NR_135484.1

Gene Symbol
SLC25A35
Entrez Gene ID
399512
Gene Name
solute carrier family 25 member 35
Gene Aliases
-
UniGene
Hs.118918
Gene Chromosome Location
Chr.17: 8287763-8295694 on Build GRCh38
Transcripts
NM_001177801.1 NM_001177802.1 NM_016492.4 XM_005256618.4 NM_001320871.1 NM_001320872.1 NM_201520.2 NR_135484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6243972 c.137G>T p.S46I Chr.17: 8289015-8289015 on Build GRCh38
6243971 c.137G>T p.S46I Chr.17: 8289015-8289015 on Build GRCh38
5655806 c.134A>T p.Q45L Chr.17: 8289012-8289012 on Build GRCh38
5028217 c.156C>G p.L52L Chr.17: 8289034-8289034 on Build GRCh38
1318419 c.181G>T p.E61* Chr.17: 8289059-8289059 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957255041 Chr.17: 8289196-8289197 on Build GRCh38
rs999135974 Chr.17: 8289198-8289198 on Build GRCh38
rs534749055 Chr.17: 8289194-8289194 on Build GRCh38
rs759538182 Chr.17: 8289039-8289039 on Build GRCh38
rs753632700 Chr.17: 8289109-8289109 on Build GRCh38
rs968978416 Chr.17: 8289031-8289031 on Build GRCh38
rs878959333 Chr.17: 8289132-8289132 on Build GRCh38
rs780082409 Chr.17: 8289111-8289111 on Build GRCh38
rs1010842675 Chr.17: 8289193-8289193 on Build GRCh38
rs1014428533 Chr.17: 8289186-8289186 on Build GRCh38
rs1027901416 Chr.17: 8289141-8289141 on Build GRCh38
rs781291944 Chr.17: 8289120-8289120 on Build GRCh38
rs373083572 Chr.17: 8289085-8289085 on Build GRCh38
rs902591326 Chr.17: 8289196-8289196 on Build GRCh38
rs939582974 Chr.17: 8289176-8289176 on Build GRCh38
rs770560485 Chr.17: 8289079-8289079 on Build GRCh38
rs1031466951 Chr.17: 8289204-8289204 on Build GRCh38
rs772884285 Chr.17: 8289035-8289035 on Build GRCh38
rs748634671 Chr.17: 8289121-8289121 on Build GRCh38
rs772883477 Chr.17: 8289053-8289053 on Build GRCh38
rs900678508 Chr.17: 8289089-8289089 on Build GRCh38
rs755920517 Chr.17: 8289048-8289048 on Build GRCh38
rs1053514842 Chr.17: 8289064-8289064 on Build GRCh38
rs774053920 Chr.17: 8289080-8289080 on Build GRCh38
rs372709645 Chr.17: 8289191-8289191 on Build GRCh38
rs377717671 Chr.17: 8289001-8289001 on Build GRCh38
rs786205827 Chr.17: 8289029-8289029 on Build GRCh38
rs746995613 Chr.17: 8289058-8289058 on Build GRCh38
rs140704891 Chr.17: 8289059-8289059 on Build GRCh38
rs762656956 Chr.17: 8289078-8289078 on Build GRCh38
rs770266134 Chr.17: 8289213-8289213 on Build GRCh38
rs569872275 Chr.17: 8289038-8289038 on Build GRCh38
rs895125242 Chr.17: 8289192-8289192 on Build GRCh38
rs755013165 Chr.17: 8289114-8289114 on Build GRCh38
rs781629730 Chr.17: 8289068-8289068 on Build GRCh38
rs758596981 Chr.17: 8289055-8289055 on Build GRCh38
rs60272214 Chr.17: 8289172-8289172 on Build GRCh38
rs778019209 Chr.17: 8289119-8289119 on Build GRCh38
rs977271397 Chr.17: 8289032-8289032 on Build GRCh38
rs1047887709 Chr.17: 8289166-8289166 on Build GRCh38
rs149958174 Chr.17: 8289069-8289069 on Build GRCh38
rs769218673 Chr.17: 8289006-8289006 on Build GRCh38
rs147679793 Chr.17: 8289050-8289050 on Build GRCh38
rs865858807 Chr.17: 8289135-8289135 on Build GRCh38
rs765614285 Chr.17: 8289022-8289022 on Build GRCh38
rs760732462 Chr.17: 8289087-8289087 on Build GRCh38
rs753655214 Chr.17: 8289123-8289124 on Build GRCh38
rs375629912 Chr.17: 8289086-8289086 on Build GRCh38
rs772149331 Chr.17: 8288994-8288994 on Build GRCh38
rs763926414 Chr.17: 8289094-8289094 on Build GRCh38
rs767491025 Chr.17: 8289043-8289043 on Build GRCh38
rs762109580 Chr.17: 8289010-8289010 on Build GRCh38
rs548428743 Chr.17: 8289002-8289002 on Build GRCh38
rs961041397 Chr.17: 8289177-8289177 on Build GRCh38
rs375282172 Chr.17: 8289082-8289082 on Build GRCh38
rs973126905 Chr.17: 8289131-8289131 on Build GRCh38
rs762555619 Chr.17: 8289036-8289036 on Build GRCh38
rs1024017853 Chr.17: 8289188-8289188 on Build GRCh38
rs758641452 Chr.17: 8289110-8289110 on Build GRCh38
rs752693132 Chr.17: 8289044-8289044 on Build GRCh38
rs750625803 Chr.17: 8289054-8289054 on Build GRCh38
rs948501530 Chr.17: 8289046-8289046 on Build GRCh38
rs780167893 Chr.17: 8289056-8289056 on Build GRCh38
rs751803347 Chr.17: 8289113-8289113 on Build GRCh38
rs876661169 Chr.17: 8289073-8289073 on Build GRCh38

View Full Product Details