Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 36881881-36882142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAX5
Entrez Gene ID
5079
Gene Name
paired box 5
Gene Aliases
ALL3, BSAP
UniGene
Hs.654464
Gene Chromosome Location
Chr.9: 36833274-37035949 on Build GRCh38
Transcripts
NM_001280547.1 NM_001280548.1 NM_001280549.1 NM_001280550.1 NM_001280551.1 NM_001280552.1 NM_001280553.1 NM_001280554.1 NM_001280555.1 NM_001280556.1 NM_016734.2 NR_103999.1 NR_104000.1 XM_005251481.4 XM_011517896.2 XM_011517897.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4993360 c.947C>T p.P316L Chr.9: 36882069-36882069 on Build GRCh38
7427499 c.933C>A p.T311T Chr.9: 36882083-36882083 on Build GRCh38
7382506 c.959C>G p.P320R Chr.9: 36882057-36882057 on Build GRCh38
6738241 c.947C>A p.P316H Chr.9: 36882069-36882069 on Build GRCh38
87165 c.47-?_1176+?del p.? Chr.9: 36840560-37020801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572234529 Chr.9: 36881988-36881988 on Build GRCh38
rs999856789 Chr.9: 36881963-36881963 on Build GRCh38
rs199646937 Chr.9: 36882102-36882102 on Build GRCh38
rs1050371686 Chr.9: 36881941-36881941 on Build GRCh38
rs750194822 Chr.9: 36882049-36882049 on Build GRCh38
rs779972349 Chr.9: 36882053-36882053 on Build GRCh38
rs571144392 Chr.9: 36881912-36881912 on Build GRCh38
rs760165632 Chr.9: 36881994-36881994 on Build GRCh38
rs150639275 Chr.9: 36881936-36881936 on Build GRCh38
rs543302698 Chr.9: 36882055-36882055 on Build GRCh38
rs753245650 Chr.9: 36882083-36882083 on Build GRCh38
rs763722779 Chr.9: 36882023-36882023 on Build GRCh38
rs371731824 Chr.9: 36881978-36881978 on Build GRCh38
rs375051266 Chr.9: 36881986-36881986 on Build GRCh38
rs369023231 Chr.9: 36882119-36882119 on Build GRCh38
rs577863510 Chr.9: 36882054-36882054 on Build GRCh38
rs35077960 Chr.9: 36882093-36882093 on Build GRCh38
rs779040709 Chr.9: 36881955-36881955 on Build GRCh38
rs563606361 Chr.9: 36882059-36882059 on Build GRCh38
rs1023906916 Chr.9: 36881997-36881997 on Build GRCh38
rs944559256 Chr.9: 36881951-36881951 on Build GRCh38
rs970894697 Chr.9: 36881911-36881911 on Build GRCh38
rs766894200 Chr.9: 36881935-36881935 on Build GRCh38
rs762138776 Chr.9: 36881984-36881984 on Build GRCh38
rs1015005579 Chr.9: 36882036-36882036 on Build GRCh38
rs34810717 Chr.9: 36882052-36882052 on Build GRCh38
rs556519070 Chr.9: 36881942-36881942 on Build GRCh38
rs952553118 Chr.9: 36881923-36881923 on Build GRCh38
rs982302460 Chr.9: 36882024-36882024 on Build GRCh38
rs141574393 Chr.9: 36882061-36882061 on Build GRCh38
rs913385985 Chr.9: 36881946-36881946 on Build GRCh38
rs144574685 Chr.9: 36882062-36882062 on Build GRCh38
rs367613041 Chr.9: 36881954-36881954 on Build GRCh38
rs369633801 Chr.9: 36882032-36882032 on Build GRCh38
rs761579249 Chr.9: 36882086-36882086 on Build GRCh38
rs940685514 Chr.9: 36881928-36881928 on Build GRCh38
rs907976546 Chr.9: 36881919-36881919 on Build GRCh38
rs763627221 Chr.9: 36882070-36882070 on Build GRCh38
rs200743794 Chr.9: 36881977-36881977 on Build GRCh38
rs145306022 Chr.9: 36882026-36882026 on Build GRCh38
rs973843299 Chr.9: 36881934-36881934 on Build GRCh38
rs772094563 Chr.9: 36881966-36881966 on Build GRCh38
rs778880926 Chr.9: 36882033-36882033 on Build GRCh38
rs145283614 Chr.9: 36882092-36882092 on Build GRCh38
rs370063069 Chr.9: 36881987-36881987 on Build GRCh38
rs780301852 Chr.9: 36881967-36881967 on Build GRCh38
rs372764912 Chr.9: 36881992-36881992 on Build GRCh38
rs760162526 Chr.9: 36882069-36882069 on Build GRCh38
rs540811536 Chr.9: 36882039-36882039 on Build GRCh38
rs773593708 Chr.9: 36882058-36882058 on Build GRCh38
rs780753361 Chr.9: 36882052-36882053 on Build GRCh38
rs779911559 Chr.9: 36882105-36882105 on Build GRCh38
rs201523208 Chr.9: 36881960-36881960 on Build GRCh38
rs982157908 Chr.9: 36882088-36882088 on Build GRCh38
rs552566484 Chr.9: 36881933-36881934 on Build GRCh38
rs376322567 Chr.9: 36882120-36882120 on Build GRCh38

View Full Product Details