Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 25279669-25279938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARMIL1
Entrez Gene ID
55604
Gene Name
capping protein regulator and myosin 1 linker 1
Gene Aliases
CARMIL, CARMIL1a, LRRC16, LRRC16A, dJ501N12.1, dJ501N12.5
UniGene
Hs.649550
Gene Chromosome Location
Chr.6: 25279428-25620530 on Build GRCh38
Transcripts
NM_001173977.1 NM_017640.5 XM_005249218.1 XM_017011008.1 XM_017011009.1 XM_017011010.1 XM_017011011.1 XM_017011012.1 XM_017011013.1 XM_017011014.1 XM_017011015.1 XM_017011016.1 XM_017011017.1

SNP Detail

SNP ID Chromosome Location
rs368677868 Chr.6: 25279844-25279844 on Build GRCh38
rs114684174 Chr.6: 25279695-25279695 on Build GRCh38
rs776610551 Chr.6: 25279842-25279842 on Build GRCh38
rs759422508 Chr.6: 25279849-25279849 on Build GRCh38
rs369678441 Chr.6: 25279764-25279764 on Build GRCh38
rs747772837 Chr.6: 25279825-25279825 on Build GRCh38
rs751623092 Chr.6: 25279781-25279781 on Build GRCh38
rs372041478 Chr.6: 25279798-25279798 on Build GRCh38
rs6914799 Chr.6: 25279891-25279891 on Build GRCh38
rs544149691 Chr.6: 25279911-25279911 on Build GRCh38
rs757300218 Chr.6: 25279869-25279869 on Build GRCh38
rs377466260 Chr.6: 25279870-25279870 on Build GRCh38
rs575845777 Chr.6: 25279767-25279767 on Build GRCh38
rs1011500184 Chr.6: 25279784-25279784 on Build GRCh38
rs35641823 Chr.6: 25279729-25279730 on Build GRCh38
rs770606353 Chr.6: 25279841-25279841 on Build GRCh38
rs375333900 Chr.6: 25279817-25279817 on Build GRCh38
rs183594257 Chr.6: 25279904-25279904 on Build GRCh38
rs2206196 Chr.6: 25279895-25279895 on Build GRCh38
rs771867706 Chr.6: 25279828-25279828 on Build GRCh38
rs764292631 Chr.6: 25279866-25279866 on Build GRCh38
rs1032151148 Chr.6: 25279748-25279748 on Build GRCh38
rs757391659 Chr.6: 25279810-25279810 on Build GRCh38
rs764817374 Chr.6: 25279773-25279774 on Build GRCh38
rs781638799 Chr.6: 25279812-25279812 on Build GRCh38
rs753787276 Chr.6: 25279752-25279752 on Build GRCh38
rs1051307880 Chr.6: 25279726-25279726 on Build GRCh38
rs969987400 Chr.6: 25279840-25279840 on Build GRCh38
rs775159478 Chr.6: 25279856-25279856 on Build GRCh38
rs908933226 Chr.6: 25279887-25279887 on Build GRCh38
rs556009624 Chr.6: 25279712-25279712 on Build GRCh38
rs773058324 Chr.6: 25279829-25279829 on Build GRCh38
rs1046554194 Chr.6: 25279821-25279821 on Build GRCh38
rs764101888 Chr.6: 25279777-25279777 on Build GRCh38
rs998976482 Chr.6: 25279735-25279735 on Build GRCh38
rs1055685905 Chr.6: 25279862-25279862 on Build GRCh38
rs746407221 Chr.6: 25279814-25279814 on Build GRCh38
rs1022842027 Chr.6: 25279787-25279787 on Build GRCh38
rs368994643 Chr.6: 25279727-25279727 on Build GRCh38
rs931475987 Chr.6: 25279775-25279775 on Build GRCh38
rs199737976 Chr.6: 25279756-25279756 on Build GRCh38
rs756168648 Chr.6: 25279879-25279879 on Build GRCh38
rs891279642 Chr.6: 25279733-25279733 on Build GRCh38
rs369908647 Chr.6: 25279882-25279882 on Build GRCh38
rs900051201 Chr.6: 25279710-25279710 on Build GRCh38
rs759344541 Chr.6: 25279755-25279755 on Build GRCh38
rs765015957 Chr.6: 25279850-25279850 on Build GRCh38
rs780318462 Chr.6: 25279819-25279819 on Build GRCh38
rs752082729 Chr.6: 25279873-25279874 on Build GRCh38
rs373297487 Chr.6: 25279863-25279863 on Build GRCh38
rs746515822 Chr.6: 25279835-25279835 on Build GRCh38
rs986547733 Chr.6: 25279725-25279725 on Build GRCh38

View Full Product Details