Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 165406753-165407022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RXRG
Entrez Gene ID
6258
Gene Name
retinoid X receptor gamma
Gene Aliases
NR2B3, RXRC
UniGene
Hs.26550
Gene Chromosome Location
Chr.1: 165400922-165445355 on Build GRCh38
Transcripts
NM_001256570.1 NM_001256571.1 NM_006917.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4926817 c.1231G>A p.E411K Chr.1: 165406825-165406825 on Build GRCh38
6770889 c.1229C>T p.P410L Chr.1: 165406827-165406827 on Build GRCh38
6122409 c.1224G>C p.K408N Chr.1: 165406832-165406832 on Build GRCh38
3976452 c.1186A>C p.K396Q Chr.1: 165406870-165406870 on Build GRCh38
121781 c.1230G>A p.P410P Chr.1: 165406826-165406826 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879814847 Chr.1: 165406991-165406992 on Build GRCh38
rs777511859 Chr.1: 165406794-165406794 on Build GRCh38
rs762921577 Chr.1: 165406876-165406876 on Build GRCh38
rs938261675 Chr.1: 165406973-165406973 on Build GRCh38
rs769870483 Chr.1: 165406862-165406862 on Build GRCh38
rs3737003 Chr.1: 165406991-165406991 on Build GRCh38
rs776927068 Chr.1: 165406827-165406827 on Build GRCh38
rs283696 Chr.1: 165406806-165406806 on Build GRCh38
rs764778331 Chr.1: 165406995-165406996 on Build GRCh38
rs1007995133 Chr.1: 165406781-165406781 on Build GRCh38
rs755414024 Chr.1: 165406956-165406956 on Build GRCh38
rs370161886 Chr.1: 165406940-165406940 on Build GRCh38
rs762173130 Chr.1: 165406844-165406844 on Build GRCh38
rs113398546 Chr.1: 165406996-165406997 on Build GRCh38
rs767566389 Chr.1: 165406921-165406921 on Build GRCh38
rs748465039 Chr.1: 165406967-165406967 on Build GRCh38
rs748696144 Chr.1: 165406803-165406803 on Build GRCh38
rs753564192 Chr.1: 165406941-165406941 on Build GRCh38
rs778539140 Chr.1: 165406950-165406950 on Build GRCh38
rs781199633 Chr.1: 165406786-165406786 on Build GRCh38
rs972854057 Chr.1: 165406847-165406847 on Build GRCh38
rs151269614 Chr.1: 165406826-165406826 on Build GRCh38
rs777044516 Chr.1: 165406972-165406972 on Build GRCh38
rs752499164 Chr.1: 165406927-165406927 on Build GRCh38
rs386636326 Chr.1: 165406991-165406992 on Build GRCh38
rs751447894 Chr.1: 165406901-165406901 on Build GRCh38
rs756713103 Chr.1: 165406947-165406947 on Build GRCh38
rs759488991 Chr.1: 165406920-165406920 on Build GRCh38
rs765694003 Chr.1: 165406784-165406786 on Build GRCh38
rs111545739 Chr.1: 165406990-165406991 on Build GRCh38
rs781689449 Chr.1: 165406964-165406964 on Build GRCh38
rs374191437 Chr.1: 165406951-165406951 on Build GRCh38
rs10800095 Chr.1: 165406992-165406992 on Build GRCh38
rs144624781 Chr.1: 165406789-165406789 on Build GRCh38
rs754790316 Chr.1: 165406779-165406779 on Build GRCh38
rs755760297 Chr.1: 165406790-165406790 on Build GRCh38
rs780391845 Chr.1: 165406825-165406825 on Build GRCh38
rs766520723 Chr.1: 165406892-165406892 on Build GRCh38
rs770244326 Chr.1: 165406968-165406968 on Build GRCh38
rs10800096 Chr.1: 165406996-165406996 on Build GRCh38
rs773507609 Chr.1: 165406872-165406872 on Build GRCh38

View Full Product Details