Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12774993-12775218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRAMEF12
Entrez Gene ID
390999
Gene Name
PRAME family member 12
Gene Aliases
-
UniGene
Hs.156406
Gene Chromosome Location
Chr.1: 12774848-12777906 on Build GRCh38
Transcripts
NM_001080830.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
894893 c.281G>A p.R94H Chr.1: 12775148-12775148 on Build GRCh38
229855 c.193C>T p.P65S Chr.1: 12775060-12775060 on Build GRCh38
894891 c.236G>A p.R79Q Chr.1: 12775103-12775103 on Build GRCh38
1237897 c.169C>G p.Q57E Chr.1: 12775036-12775036 on Build GRCh38
894892 c.276G>A p.K92K Chr.1: 12775143-12775143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771521300 Chr.1: 12775184-12775184 on Build GRCh38
rs371925896 Chr.1: 12775148-12775148 on Build GRCh38
rs980527168 Chr.1: 12775072-12775072 on Build GRCh38
rs77870074 Chr.1: 12775015-12775015 on Build GRCh38
rs774801185 Chr.1: 12775167-12775167 on Build GRCh38
rs760289612 Chr.1: 12775127-12775127 on Build GRCh38
rs781305529 Chr.1: 12775157-12775157 on Build GRCh38
rs545355065 Chr.1: 12775069-12775069 on Build GRCh38
rs757292758 Chr.1: 12775096-12775096 on Build GRCh38
rs761245297 Chr.1: 12775124-12775124 on Build GRCh38
rs912009742 Chr.1: 12775020-12775020 on Build GRCh38
rs374883271 Chr.1: 12775193-12775193 on Build GRCh38
rs774211948 Chr.1: 12775177-12775177 on Build GRCh38
rs769003046 Chr.1: 12775157-12775157 on Build GRCh38
rs769755107 Chr.1: 12775103-12775103 on Build GRCh38
rs747299902 Chr.1: 12775013-12775013 on Build GRCh38
rs770598552 Chr.1: 12775192-12775192 on Build GRCh38
rs572738695 Chr.1: 12775179-12775179 on Build GRCh38
rs150914377 Chr.1: 12775097-12775097 on Build GRCh38
rs367786450 Chr.1: 12775116-12775116 on Build GRCh38
rs746102731 Chr.1: 12775102-12775102 on Build GRCh38
rs780385217 Chr.1: 12775164-12775164 on Build GRCh38
rs375245601 Chr.1: 12775131-12775131 on Build GRCh38
rs775407687 Chr.1: 12775111-12775111 on Build GRCh38
rs75099356 Chr.1: 12775014-12775014 on Build GRCh38
rs372248134 Chr.1: 12775126-12775126 on Build GRCh38
rs376821629 Chr.1: 12775132-12775132 on Build GRCh38
rs1003256734 Chr.1: 12775117-12775117 on Build GRCh38
rs761347326 Chr.1: 12775047-12775047 on Build GRCh38
rs773756461 Chr.1: 12775046-12775046 on Build GRCh38
rs747087551 Chr.1: 12775088-12775088 on Build GRCh38
rs763746767 Chr.1: 12775044-12775044 on Build GRCh38
rs913441810 Chr.1: 12775150-12775150 on Build GRCh38
rs35799100 Chr.1: 12775156-12775157 on Build GRCh38
rs929889334 Chr.1: 12775063-12775063 on Build GRCh38
rs934391406 Chr.1: 12775129-12775129 on Build GRCh38
rs765581572 Chr.1: 12775070-12775070 on Build GRCh38
rs775639002 Chr.1: 12775036-12775036 on Build GRCh38
rs758877294 Chr.1: 12775083-12775083 on Build GRCh38
rs767039459 Chr.1: 12775125-12775125 on Build GRCh38
rs746145515 Chr.1: 12775160-12775160 on Build GRCh38
rs1052656015 Chr.1: 12775197-12775197 on Build GRCh38
rs746053480 Chr.1: 12775045-12775045 on Build GRCh38
rs769702820 Chr.1: 12775031-12775031 on Build GRCh38
rs17346571 Chr.1: 12775025-12775025 on Build GRCh38
rs753242708 Chr.1: 12775079-12775079 on Build GRCh38
rs369581729 Chr.1: 12775136-12775136 on Build GRCh38
rs752095583 Chr.1: 12775147-12775147 on Build GRCh38
rs768442397 Chr.1: 12775173-12775173 on Build GRCh38
rs772755955 Chr.1: 12775186-12775186 on Build GRCh38
rs1057260574 Chr.1: 12775106-12775106 on Build GRCh38
rs767083076 Chr.1: 12775053-12775053 on Build GRCh38
rs760124483 Chr.1: 12775190-12775190 on Build GRCh38
rs371502542 Chr.1: 12775056-12775056 on Build GRCh38
rs752655482 Chr.1: 12775120-12775120 on Build GRCh38
rs200879331 Chr.1: 12775043-12775043 on Build GRCh38
rs780170279 Chr.1: 12775161-12775161 on Build GRCh38
rs1048331355 Chr.1: 12775067-12775067 on Build GRCh38

View Full Product Details