Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 103786498-103786757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLP1
Entrez Gene ID
5354
Gene Name
proteolipid protein 1
Gene Aliases
GPM6C, HLD1, MMPL, PLP, PLP/DM20, PMD, SPG2
UniGene
Hs.1787
Gene Chromosome Location
Chr.X: 103776506-103792619 on Build GRCh38
Transcripts
NM_000533.4 NM_001128834.2 NM_001305004.1 NM_199478.2

SNP Detail

SNP ID Chromosome Location
rs112667596 Chr.X: 103786729-103786729 on Build GRCh38
rs140047941 Chr.X: 103786528-103786528 on Build GRCh38
rs779258535 Chr.X: 103786671-103786671 on Build GRCh38
rs749278720 Chr.X: 103786620-103786620 on Build GRCh38
rs763892975 Chr.X: 103786555-103786555 on Build GRCh38
rs886044450 Chr.X: 103786726-103786726 on Build GRCh38
rs132630287 Chr.X: 103786691-103786691 on Build GRCh38
rs746081433 Chr.X: 103786576-103786576 on Build GRCh38
rs772312288 Chr.X: 103786721-103786721 on Build GRCh38
rs11543024 Chr.X: 103786540-103786540 on Build GRCh38
rs1013902509 Chr.X: 103786703-103786703 on Build GRCh38
rs973903346 Chr.X: 103786591-103786591 on Build GRCh38
rs132630292 Chr.X: 103786707-103786707 on Build GRCh38
rs878853076 Chr.X: 103786661-103786661 on Build GRCh38
rs756976500 Chr.X: 103786561-103786561 on Build GRCh38
rs769235659 Chr.X: 103786663-103786663 on Build GRCh38
rs748595198 Chr.X: 103786681-103786681 on Build GRCh38
rs780293374 Chr.X: 103786618-103786618 on Build GRCh38
rs132630295 Chr.X: 103786682-103786682 on Build GRCh38
rs756334357 Chr.X: 103786615-103786615 on Build GRCh38
rs113345335 Chr.X: 103786727-103786727 on Build GRCh38
rs751404730 Chr.X: 103786558-103786558 on Build GRCh38
rs758372406 Chr.X: 103786532-103786532 on Build GRCh38

View Full Product Details