Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 142848264-142848525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCOLCE2
Entrez Gene ID
26577
Gene Name
procollagen C-endopeptidase enhancer 2
Gene Aliases
PCPE2
UniGene
Hs.8944
Gene Chromosome Location
Chr.3: 142817860-142889203 on Build GRCh38
Transcripts
NM_013363.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6892947 c.269T>G p.F90C Chr.3: 142848396-142848396 on Build GRCh38
5690630 c.244A>T p.S82C Chr.3: 142848421-142848421 on Build GRCh38
6741981 c.306C>T p.R102R Chr.3: 142848359-142848359 on Build GRCh38
3588322 c.366G>A p.M122I Chr.3: 142848299-142848299 on Build GRCh38
3588323 c.217C>T p.L73F Chr.3: 142848448-142848448 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139562886 Chr.3: 142848358-142848358 on Build GRCh38
rs778865512 Chr.3: 142848384-142848384 on Build GRCh38
rs767590952 Chr.3: 142848416-142848416 on Build GRCh38
rs774897446 Chr.3: 142848330-142848330 on Build GRCh38
rs41267847 Chr.3: 142848433-142848433 on Build GRCh38
rs749519201 Chr.3: 142848378-142848378 on Build GRCh38
rs763892062 Chr.3: 142848483-142848483 on Build GRCh38
rs41267845 Chr.3: 142848361-142848361 on Build GRCh38
rs756422671 Chr.3: 142848452-142848452 on Build GRCh38
rs143728484 Chr.3: 142848304-142848304 on Build GRCh38
rs762581941 Chr.3: 142848405-142848405 on Build GRCh38
rs373201044 Chr.3: 142848439-142848439 on Build GRCh38
rs897195384 Chr.3: 142848397-142848397 on Build GRCh38
rs201403616 Chr.3: 142848425-142848425 on Build GRCh38
rs749128310 Chr.3: 142848490-142848490 on Build GRCh38
rs770509532 Chr.3: 142848466-142848466 on Build GRCh38
rs917519830 Chr.3: 142848444-142848444 on Build GRCh38
rs547465850 Chr.3: 142848324-142848324 on Build GRCh38
rs189139903 Chr.3: 142848380-142848380 on Build GRCh38
rs760523969 Chr.3: 142848422-142848422 on Build GRCh38
rs374060736 Chr.3: 142848387-142848387 on Build GRCh38
rs781411140 Chr.3: 142848449-142848449 on Build GRCh38
rs759493410 Chr.3: 142848414-142848414 on Build GRCh38
rs781415884 Chr.3: 142848377-142848377 on Build GRCh38
rs752558576 Chr.3: 142848368-142848368 on Build GRCh38
rs775845872 Chr.3: 142848341-142848341 on Build GRCh38
rs545499868 Chr.3: 142848486-142848486 on Build GRCh38
rs931692154 Chr.3: 142848347-142848347 on Build GRCh38
rs569272508 Chr.3: 142848457-142848457 on Build GRCh38
rs759085276 Chr.3: 142848467-142848467 on Build GRCh38
rs200603685 Chr.3: 142848388-142848388 on Build GRCh38
rs756364642 Chr.3: 142848373-142848373 on Build GRCh38
rs780179321 Chr.3: 142848492-142848492 on Build GRCh38
rs893883 Chr.3: 142848485-142848485 on Build GRCh38
rs767358243 Chr.3: 142848365-142848365 on Build GRCh38
rs749416279 Chr.3: 142848305-142848305 on Build GRCh38
rs202177059 Chr.3: 142848333-142848333 on Build GRCh38
rs751653213 Chr.3: 142848334-142848334 on Build GRCh38
rs760760969 Chr.3: 142848475-142848475 on Build GRCh38
rs769008057 Chr.3: 142848446-142848446 on Build GRCh38
rs770956817 Chr.3: 142848319-142848319 on Build GRCh38
rs909525456 Chr.3: 142848473-142848473 on Build GRCh38
rs758461643 Chr.3: 142848491-142848491 on Build GRCh38
rs777947858 Chr.3: 142848302-142848302 on Build GRCh38
rs369893796 Chr.3: 142848357-142848357 on Build GRCh38
rs769061409 Chr.3: 142848399-142848399 on Build GRCh38
rs752721834 Chr.3: 142848420-142848420 on Build GRCh38
rs747577646 Chr.3: 142848445-142848445 on Build GRCh38
rs375923649 Chr.3: 142848345-142848345 on Build GRCh38
rs747346478 Chr.3: 142848391-142848391 on Build GRCh38
rs748339219 Chr.3: 142848330-142848330 on Build GRCh38
rs200433586 Chr.3: 142848406-142848406 on Build GRCh38
rs775554921 Chr.3: 142848468-142848468 on Build GRCh38
rs147227072 Chr.3: 142848351-142848351 on Build GRCh38
rs956193292 Chr.3: 142848427-142848427 on Build GRCh38
rs143959509 Chr.3: 142848438-142848438 on Build GRCh38
rs770014911 Chr.3: 142848472-142848472 on Build GRCh38
rs537602842 Chr.3: 142848403-142848403 on Build GRCh38
rs765346956 Chr.3: 142848487-142848487 on Build GRCh38
rs748909749 Chr.3: 142848451-142848451 on Build GRCh38
rs199789465 Chr.3: 142848437-142848437 on Build GRCh38

View Full Product Details