Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20126909-20127182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANBP1
Entrez Gene ID
5902
Gene Name
RAN binding protein 1
Gene Aliases
HTF9A
UniGene
Hs.24763
Gene Chromosome Location
Chr.22: 20115938-20127357 on Build GRCh38
Transcripts
NM_001278639.1 NM_001278640.1 NM_001278641.1 NM_002882.3 XM_011530290.2 XM_011530291.2 XM_017028890.1 XM_017028891.1 XM_017028892.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1751731 c.530C>T p.A177V Chr.22: 20126976-20126976 on Build GRCh38
277000 c.606A>C p.*202Y Chr.22: 20127052-20127052 on Build GRCh38
1641502 c.564G>A p.S188S Chr.22: 20127010-20127010 on Build GRCh38
5798062 c.506-10G>C p.? Chr.22: 20126942-20126942 on Build GRCh38
6338293 c.522T>C p.N174N Chr.22: 20126968-20126968 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536566752 Chr.22: 20127056-20127056 on Build GRCh38
rs758153400 Chr.22: 20127127-20127127 on Build GRCh38
rs767041773 Chr.22: 20127072-20127072 on Build GRCh38
rs778276585 Chr.22: 20126937-20126937 on Build GRCh38
rs752062270 Chr.22: 20127071-20127071 on Build GRCh38
rs189965701 Chr.22: 20127117-20127117 on Build GRCh38
rs891175953 Chr.22: 20127015-20127015 on Build GRCh38
rs374874705 Chr.22: 20127081-20127081 on Build GRCh38
rs773220129 Chr.22: 20127033-20127033 on Build GRCh38
rs749700582 Chr.22: 20127004-20127004 on Build GRCh38
rs147326480 Chr.22: 20126995-20126995 on Build GRCh38
rs757616541 Chr.22: 20127081-20127085 on Build GRCh38
rs150530613 Chr.22: 20126960-20126960 on Build GRCh38
rs759533670 Chr.22: 20126955-20126955 on Build GRCh38
rs772091421 Chr.22: 20126945-20126945 on Build GRCh38
rs763847750 Chr.22: 20127058-20127058 on Build GRCh38
rs749227604 Chr.22: 20127099-20127099 on Build GRCh38
rs150814421 Chr.22: 20126989-20126989 on Build GRCh38
rs376753468 Chr.22: 20127049-20127049 on Build GRCh38
rs930941473 Chr.22: 20127035-20127035 on Build GRCh38
rs1015098169 Chr.22: 20127098-20127099 on Build GRCh38
rs775387952 Chr.22: 20127098-20127098 on Build GRCh38
rs756622658 Chr.22: 20127080-20127080 on Build GRCh38
rs868441154 Chr.22: 20127055-20127055 on Build GRCh38
rs943548168 Chr.22: 20127070-20127070 on Build GRCh38
rs755337721 Chr.22: 20127085-20127085 on Build GRCh38
rs769675190 Chr.22: 20127093-20127093 on Build GRCh38
rs757850731 Chr.22: 20127015-20127017 on Build GRCh38
rs148747950 Chr.22: 20126956-20126956 on Build GRCh38
rs768710680 Chr.22: 20127009-20127009 on Build GRCh38
rs780610564 Chr.22: 20126988-20126988 on Build GRCh38
rs749968013 Chr.22: 20127076-20127076 on Build GRCh38
rs747119279 Chr.22: 20127092-20127092 on Build GRCh38
rs369996254 Chr.22: 20127118-20127118 on Build GRCh38
rs375149255 Chr.22: 20126931-20126931 on Build GRCh38
rs751195972 Chr.22: 20127062-20127062 on Build GRCh38
rs772185902 Chr.22: 20127031-20127031 on Build GRCh38
rs956865616 Chr.22: 20127141-20127141 on Build GRCh38
rs910689012 Chr.22: 20127066-20127066 on Build GRCh38
rs765014367 Chr.22: 20126942-20126942 on Build GRCh38
rs752498683 Chr.22: 20126963-20126963 on Build GRCh38
rs371388330 Chr.22: 20127071-20127071 on Build GRCh38
rs754445052 Chr.22: 20127084-20127084 on Build GRCh38
rs372271212 Chr.22: 20127027-20127027 on Build GRCh38
rs944069567 Chr.22: 20127079-20127079 on Build GRCh38
rs1033350399 Chr.22: 20127115-20127115 on Build GRCh38
rs1012720926 Chr.22: 20127126-20127126 on Build GRCh38
rs969657664 Chr.22: 20127120-20127120 on Build GRCh38
rs919494336 Chr.22: 20127021-20127021 on Build GRCh38
rs138409144 Chr.22: 20126977-20126977 on Build GRCh38
rs887633027 Chr.22: 20127092-20127092 on Build GRCh38
rs372174173 Chr.22: 20126967-20126967 on Build GRCh38
rs775229896 Chr.22: 20127053-20127053 on Build GRCh38
rs182031737 Chr.22: 20127110-20127110 on Build GRCh38
rs971346070 Chr.22: 20127002-20127002 on Build GRCh38
rs781574041 Chr.22: 20127091-20127092 on Build GRCh38
rs368678998 Chr.22: 20127018-20127018 on Build GRCh38
rs756772977 Chr.22: 20126979-20126979 on Build GRCh38
rs185846819 Chr.22: 20127111-20127111 on Build GRCh38
rs764432545 Chr.22: 20127059-20127063 on Build GRCh38
rs368048123 Chr.22: 20126933-20126933 on Build GRCh38
rs776598352 Chr.22: 20126951-20126951 on Build GRCh38
rs748386685 Chr.22: 20127090-20127090 on Build GRCh38
rs925486097 Chr.22: 20127128-20127128 on Build GRCh38
rs752196675 Chr.22: 20126949-20126950 on Build GRCh38

View Full Product Details