Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55785632-55785905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRP11
Entrez Gene ID
204801
Gene Name
NLR family pyrin domain containing 11
Gene Aliases
CLR19.6, NALP11, NOD17, PAN10, PYPAF6, PYPAF7
UniGene
Hs.375039
Gene Chromosome Location
Chr.19: 55785397-55836762 on Build GRCh38
Transcripts
NM_001297743.1 NM_145007.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2969485 c.3046C>T p.P1016S Chr.19: 55785681-55785681 on Build GRCh38
1712868 c.3025G>A p.D1009N Chr.19: 55785702-55785702 on Build GRCh38
1712869 c.2959G>A p.E987K Chr.19: 55785768-55785768 on Build GRCh38
2969487 c.2995C>T p.P999S Chr.19: 55785732-55785732 on Build GRCh38
3735557 c.3047C>T p.P1016L Chr.19: 55785680-55785680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761117440 Chr.19: 55785743-55785743 on Build GRCh38
rs768139801 Chr.19: 55785714-55785714 on Build GRCh38
rs775377221 Chr.19: 55785661-55785661 on Build GRCh38
rs761789117 Chr.19: 55785710-55785710 on Build GRCh38
rs750399264 Chr.19: 55785796-55785796 on Build GRCh38
rs755811062 Chr.19: 55785884-55785885 on Build GRCh38
rs972671262 Chr.19: 55785753-55785753 on Build GRCh38
rs759879184 Chr.19: 55785864-55785864 on Build GRCh38
rs925645925 Chr.19: 55785662-55785662 on Build GRCh38
rs373925447 Chr.19: 55785848-55785848 on Build GRCh38
rs193180010 Chr.19: 55785656-55785656 on Build GRCh38
rs749071062 Chr.19: 55785807-55785807 on Build GRCh38
rs780686846 Chr.19: 55785792-55785792 on Build GRCh38
rs769714987 Chr.19: 55785660-55785660 on Build GRCh38
rs868013577 Chr.19: 55785799-55785799 on Build GRCh38
rs1052909714 Chr.19: 55785698-55785698 on Build GRCh38
rs761693639 Chr.19: 55785822-55785822 on Build GRCh38
rs1014066502 Chr.19: 55785783-55785783 on Build GRCh38
rs926576214 Chr.19: 55785849-55785849 on Build GRCh38
rs773290992 Chr.19: 55785828-55785828 on Build GRCh38
rs766748570 Chr.19: 55785744-55785744 on Build GRCh38
rs868582143 Chr.19: 55785755-55785755 on Build GRCh38
rs768970824 Chr.19: 55785682-55785682 on Build GRCh38
rs758084844 Chr.19: 55785760-55785760 on Build GRCh38
rs772062539 Chr.19: 55785823-55785823 on Build GRCh38
rs982205774 Chr.19: 55785833-55785833 on Build GRCh38
rs763871559 Chr.19: 55785874-55785874 on Build GRCh38
rs1016511812 Chr.19: 55785679-55785679 on Build GRCh38
rs777026964 Chr.19: 55785863-55785863 on Build GRCh38
rs919332181 Chr.19: 55785791-55785791 on Build GRCh38
rs115847229 Chr.19: 55785668-55785668 on Build GRCh38
rs763628855 Chr.19: 55785774-55785774 on Build GRCh38
rs370557210 Chr.19: 55785787-55785787 on Build GRCh38
rs753740562 Chr.19: 55785815-55785815 on Build GRCh38
rs780035413 Chr.19: 55785801-55785801 on Build GRCh38
rs59238812 Chr.19: 55785883-55785883 on Build GRCh38
rs751137056 Chr.19: 55785777-55785777 on Build GRCh38
rs370823183 Chr.19: 55785655-55785655 on Build GRCh38
rs116820715 Chr.19: 55785798-55785798 on Build GRCh38
rs764263811 Chr.19: 55785884-55785887 on Build GRCh38
rs754157491 Chr.19: 55785757-55785757 on Build GRCh38
rs751788635 Chr.19: 55785883-55785884 on Build GRCh38
rs779002503 Chr.19: 55785820-55785820 on Build GRCh38
rs748337702 Chr.19: 55785821-55785821 on Build GRCh38
rs536704098 Chr.19: 55785862-55785862 on Build GRCh38
rs368312156 Chr.19: 55785693-55785693 on Build GRCh38
rs144909019 Chr.19: 55785717-55785717 on Build GRCh38
rs866074331 Chr.19: 55785768-55785768 on Build GRCh38

View Full Product Details