Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76329944-76330203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPSAP1
Entrez Gene ID
5635
Gene Name
phosphoribosyl pyrophosphate synthetase associated protein 1
Gene Aliases
PAP39
UniGene
Hs.77498
Gene Chromosome Location
Chr.17: 76310787-76354149 on Build GRCh38
Transcripts
NM_002766.2 XM_011525034.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6349072 c.516C>A p.V172V Chr.17: 76330075-76330075 on Build GRCh38
276917 c.502T>C p.Y168H Chr.17: 76330089-76330089 on Build GRCh38
4718049 c.499A>G p.N167D Chr.17: 76330092-76330092 on Build GRCh38
6756740 c.586A>G p.N196D Chr.17: 76330092-76330092 on Build GRCh38
7166112 c.546G>T p.K182N Chr.17: 76330045-76330045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531265730 Chr.17: 76330112-76330112 on Build GRCh38
rs147421725 Chr.17: 76330014-76330014 on Build GRCh38
rs372471355 Chr.17: 76330132-76330132 on Build GRCh38
rs754170194 Chr.17: 76330133-76330133 on Build GRCh38
rs371577670 Chr.17: 76330117-76330117 on Build GRCh38
rs375162743 Chr.17: 76329999-76329999 on Build GRCh38
rs771129032 Chr.17: 76330102-76330102 on Build GRCh38
rs995221851 Chr.17: 76330081-76330081 on Build GRCh38
rs760135444 Chr.17: 76329998-76329998 on Build GRCh38
rs778297572 Chr.17: 76330027-76330027 on Build GRCh38
rs756524506 Chr.17: 76330008-76330008 on Build GRCh38
rs772031475 Chr.17: 76330036-76330036 on Build GRCh38
rs756846521 Chr.17: 76330019-76330019 on Build GRCh38
rs768408827 Chr.17: 76330064-76330064 on Build GRCh38
rs181291596 Chr.17: 76330074-76330074 on Build GRCh38
rs779922322 Chr.17: 76330042-76330042 on Build GRCh38
rs753132828 Chr.17: 76330001-76330001 on Build GRCh38
rs746858380 Chr.17: 76330057-76330057 on Build GRCh38
rs542385758 Chr.17: 76330029-76330029 on Build GRCh38
rs774324405 Chr.17: 76330104-76330104 on Build GRCh38
rs749459779 Chr.17: 76330078-76330078 on Build GRCh38
rs756862124 Chr.17: 76330144-76330144 on Build GRCh38
rs369620317 Chr.17: 76330011-76330011 on Build GRCh38

View Full Product Details