Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 28300572-28300831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGBD1
Entrez Gene ID
84547
Gene Name
piggyBac transposable element derived 1
Gene Aliases
HUCEP-4, SCAND4, dJ874C20.4
UniGene
Hs.144527
Gene Chromosome Location
Chr.6: 28281537-28303681 on Build GRCh38
Transcripts
NM_001184743.1 NM_032507.3 XM_006715228.2 XM_011514942.2 XM_011514943.2 XM_011514944.2 XM_017011361.1 XM_017011362.1 XM_017011363.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
150062 c.870-4C>T p.? Chr.6: 28300720-28300720 on Build GRCh38
5474920 c.949C>T p.R317C Chr.6: 28300803-28300803 on Build GRCh38
1744080 c.965T>C p.L322S Chr.6: 28300819-28300819 on Build GRCh38
1077070 c.973C>T p.R325C Chr.6: 28300827-28300827 on Build GRCh38
4751754 c.950G>A p.R317H Chr.6: 28300804-28300804 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762522752 Chr.6: 28300677-28300677 on Build GRCh38
rs576383391 Chr.6: 28300630-28300630 on Build GRCh38
rs3215466 Chr.6: 28300708-28300708 on Build GRCh38
rs768016797 Chr.6: 28300780-28300780 on Build GRCh38
rs749461439 Chr.6: 28300724-28300724 on Build GRCh38
rs767565866 Chr.6: 28300681-28300681 on Build GRCh38
rs774864891 Chr.6: 28300757-28300757 on Build GRCh38
rs1032120839 Chr.6: 28300619-28300619 on Build GRCh38
rs542116014 Chr.6: 28300648-28300648 on Build GRCh38
rs970227257 Chr.6: 28300670-28300670 on Build GRCh38
rs377211662 Chr.6: 28300801-28300801 on Build GRCh38
rs763638943 Chr.6: 28300766-28300766 on Build GRCh38
rs1024506912 Chr.6: 28300659-28300659 on Build GRCh38
rs751967647 Chr.6: 28300684-28300684 on Build GRCh38
rs373880207 Chr.6: 28300735-28300735 on Build GRCh38
rs751305530 Chr.6: 28300772-28300772 on Build GRCh38
rs966594012 Chr.6: 28300592-28300592 on Build GRCh38
rs756584844 Chr.6: 28300795-28300795 on Build GRCh38
rs1014494815 Chr.6: 28300646-28300646 on Build GRCh38
rs751005753 Chr.6: 28300715-28300715 on Build GRCh38
rs562802961 Chr.6: 28300606-28300606 on Build GRCh38
rs979257205 Chr.6: 28300614-28300614 on Build GRCh38
rs1936365 Chr.6: 28300675-28300675 on Build GRCh38
rs950982359 Chr.6: 28300620-28300620 on Build GRCh38
rs770756381 Chr.6: 28300737-28300737 on Build GRCh38
rs781758578 Chr.6: 28300706-28300706 on Build GRCh38
rs769288394 Chr.6: 28300755-28300755 on Build GRCh38
rs768757599 Chr.6: 28300726-28300726 on Build GRCh38
rs111232284 Chr.6: 28300785-28300785 on Build GRCh38
rs779166431 Chr.6: 28300733-28300733 on Build GRCh38
rs762237088 Chr.6: 28300776-28300776 on Build GRCh38
rs199718652 Chr.6: 28300804-28300804 on Build GRCh38
rs764681940 Chr.6: 28300673-28300673 on Build GRCh38
rs933343311 Chr.6: 28300698-28300698 on Build GRCh38
rs112042918 Chr.6: 28300671-28300671 on Build GRCh38
rs144722937 Chr.6: 28300651-28300651 on Build GRCh38
rs751780526 Chr.6: 28300605-28300605 on Build GRCh38
rs762589471 Chr.6: 28300758-28300758 on Build GRCh38
rs185338871 Chr.6: 28300747-28300747 on Build GRCh38
rs2281043 Chr.6: 28300720-28300720 on Build GRCh38
rs757754250 Chr.6: 28300697-28300697 on Build GRCh38
rs780678888 Chr.6: 28300803-28300803 on Build GRCh38
rs759410392 Chr.6: 28300753-28300753 on Build GRCh38
rs756171018 Chr.6: 28300719-28300719 on Build GRCh38
rs755137447 Chr.6: 28300808-28300808 on Build GRCh38

View Full Product Details