Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 74380935-74381208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EREG
Entrez Gene ID
2069
Gene Name
epiregulin
Gene Aliases
EPR, ER, Ep
UniGene
Hs.115263
Gene Chromosome Location
Chr.4: 74365143-74388760 on Build GRCh38
Transcripts
NM_001432.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2831473 c.238C>G p.Q80E Chr.4: 74381097-74381097 on Build GRCh38
6168072 c.201G>A p.K67K Chr.4: 74381060-74381060 on Build GRCh38
4386775 c.237A>G p.G79G Chr.4: 74381096-74381096 on Build GRCh38
6100979 c.189G>A p.V63V Chr.4: 74381048-74381048 on Build GRCh38
4125931 c.276C>T p.C92C Chr.4: 74381135-74381135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879069008 Chr.4: 74381137-74381137 on Build GRCh38
rs756454537 Chr.4: 74381142-74381142 on Build GRCh38
rs371135213 Chr.4: 74381069-74381069 on Build GRCh38
rs367705319 Chr.4: 74381070-74381070 on Build GRCh38
rs201201335 Chr.4: 74380991-74380991 on Build GRCh38
rs765534458 Chr.4: 74381015-74381015 on Build GRCh38
rs113254970 Chr.4: 74381072-74381072 on Build GRCh38
rs55702318 Chr.4: 74381011-74381011 on Build GRCh38
rs754585265 Chr.4: 74381160-74381160 on Build GRCh38
rs961048154 Chr.4: 74381126-74381126 on Build GRCh38
rs751662634 Chr.4: 74381031-74381031 on Build GRCh38
rs767497744 Chr.4: 74380981-74380981 on Build GRCh38
rs751540177 Chr.4: 74381129-74381129 on Build GRCh38
rs758549905 Chr.4: 74381018-74381018 on Build GRCh38
rs145916659 Chr.4: 74381045-74381045 on Build GRCh38
rs780340135 Chr.4: 74381157-74381157 on Build GRCh38
rs763197664 Chr.4: 74381107-74381107 on Build GRCh38
rs370963182 Chr.4: 74381008-74381008 on Build GRCh38
rs1031227852 Chr.4: 74380968-74380968 on Build GRCh38
rs529424022 Chr.4: 74381035-74381035 on Build GRCh38
rs759052961 Chr.4: 74381174-74381175 on Build GRCh38
rs778220054 Chr.4: 74381077-74381077 on Build GRCh38
rs375592554 Chr.4: 74381034-74381034 on Build GRCh38
rs772846778 Chr.4: 74380994-74380994 on Build GRCh38
rs189413524 Chr.4: 74381093-74381093 on Build GRCh38
rs372206060 Chr.4: 74381124-74381124 on Build GRCh38
rs754208270 Chr.4: 74381159-74381159 on Build GRCh38
rs778649779 Chr.4: 74381163-74381163 on Build GRCh38
rs774016656 Chr.4: 74381154-74381157 on Build GRCh38
rs767878864 Chr.4: 74381133-74381133 on Build GRCh38
rs964195523 Chr.4: 74381118-74381118 on Build GRCh38
rs764211023 Chr.4: 74381024-74381024 on Build GRCh38
rs776578500 Chr.4: 74381104-74381104 on Build GRCh38
rs773255236 Chr.4: 74381092-74381092 on Build GRCh38
rs771860305 Chr.4: 74381086-74381086 on Build GRCh38
rs376675128 Chr.4: 74381066-74381066 on Build GRCh38
rs376767812 Chr.4: 74381130-74381130 on Build GRCh38
rs764438350 Chr.4: 74381113-74381113 on Build GRCh38
rs751004908 Chr.4: 74381138-74381138 on Build GRCh38
rs756509590 Chr.4: 74381046-74381046 on Build GRCh38
rs747592717 Chr.4: 74381166-74381166 on Build GRCh38
rs747725770 Chr.4: 74381080-74381080 on Build GRCh38

View Full Product Details