Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 57014982-57015255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEND6
Entrez Gene ID
221336
Gene Name
BEN domain containing 6
Gene Aliases
C6orf65
UniGene
Hs.582993
Gene Chromosome Location
Chr.6: 56954975-57027652 on Build GRCh38
Transcripts
NM_152731.2 XM_005248889.1 XM_011514346.2 XM_017010406.1 XM_017010407.1 XM_017010408.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5201568 c.379C>T p.H127Y Chr.6: 57015149-57015149 on Build GRCh38
3830900 c.331T>A p.L111M Chr.6: 57015165-57015165 on Build GRCh38
5414521 c.368G>A p.G123E Chr.6: 57015202-57015202 on Build GRCh38
7181136 c.383C>G p.S128C Chr.6: 57015217-57015217 on Build GRCh38
331507 c.338G>T p.G113V Chr.6: 57015172-57015172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138064771 Chr.6: 57015063-57015064 on Build GRCh38
rs78910641 Chr.6: 57015014-57015014 on Build GRCh38
rs546241637 Chr.6: 57015215-57015215 on Build GRCh38
rs575563406 Chr.6: 57015200-57015200 on Build GRCh38
rs370271833 Chr.6: 57015133-57015133 on Build GRCh38
rs372231731 Chr.6: 57015188-57015188 on Build GRCh38
rs889291684 Chr.6: 57015063-57015063 on Build GRCh38
rs769975234 Chr.6: 57015162-57015162 on Build GRCh38
rs1033450509 Chr.6: 57015204-57015204 on Build GRCh38
rs551551087 Chr.6: 57015194-57015194 on Build GRCh38
rs1051901700 Chr.6: 57015052-57015052 on Build GRCh38
rs80252771 Chr.6: 57015073-57015073 on Build GRCh38
rs752489669 Chr.6: 57015218-57015218 on Build GRCh38
rs368474009 Chr.6: 57015206-57015206 on Build GRCh38
rs755909139 Chr.6: 57015219-57015219 on Build GRCh38
rs375335970 Chr.6: 57015066-57015066 on Build GRCh38
rs375033965 Chr.6: 57015165-57015165 on Build GRCh38
rs554763248 Chr.6: 57015179-57015179 on Build GRCh38
rs71675764 Chr.6: 57015027-57015028 on Build GRCh38
rs749961367 Chr.6: 57015085-57015085 on Build GRCh38
rs548090496 Chr.6: 57015017-57015017 on Build GRCh38
rs757958602 Chr.6: 57015093-57015093 on Build GRCh38
rs115919055 Chr.6: 57015037-57015037 on Build GRCh38
rs750059493 Chr.6: 57015201-57015201 on Build GRCh38
rs777890465 Chr.6: 57015224-57015224 on Build GRCh38
rs774865989 Chr.6: 57015185-57015185 on Build GRCh38
rs764940149 Chr.6: 57015084-57015084 on Build GRCh38
rs536652097 Chr.6: 57015018-57015018 on Build GRCh38
rs553468125 Chr.6: 57015019-57015026 on Build GRCh38
rs780788773 Chr.6: 57015115-57015115 on Build GRCh38
rs888232354 Chr.6: 57015094-57015094 on Build GRCh38
rs765939236 Chr.6: 57015205-57015205 on Build GRCh38
rs541651390 Chr.6: 57015176-57015176 on Build GRCh38
rs574624527 Chr.6: 57015113-57015113 on Build GRCh38
rs550560057 Chr.6: 57015015-57015015 on Build GRCh38
rs746902395 Chr.6: 57015111-57015111 on Build GRCh38
rs759983043 Chr.6: 57015190-57015190 on Build GRCh38
rs1033998764 Chr.6: 57015172-57015172 on Build GRCh38
rs749466881 Chr.6: 57015227-57015227 on Build GRCh38
rs149690283 Chr.6: 57015026-57015027 on Build GRCh38
rs761089380 Chr.6: 57015198-57015198 on Build GRCh38
rs552768315 Chr.6: 57015095-57015095 on Build GRCh38
rs774004085 Chr.6: 57015091-57015093 on Build GRCh38
rs970416397 Chr.6: 57015196-57015196 on Build GRCh38
rs947401595 Chr.6: 57015009-57015009 on Build GRCh38
rs199545074 Chr.6: 57015192-57015192 on Build GRCh38
rs182332221 Chr.6: 57015031-57015031 on Build GRCh38
rs368001280 Chr.6: 57015180-57015180 on Build GRCh38

View Full Product Details