Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 22589182-22589429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA8
Entrez Gene ID
2046
Gene Name
EPH receptor A8
Gene Aliases
EEK, EK3, HEK3
UniGene
Hs.283613
Gene Chromosome Location
Chr.1: 22563508-22603594 on Build GRCh38
Transcripts
NM_001006943.1 XM_011540969.1 XM_011540970.1 XM_011540972.1 XM_011540973.1 XM_011540975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6655139 c.1400G>T p.G467V Chr.1: 22589291-22589291 on Build GRCh38
4927537 c.1301C>T p.T434I Chr.1: 22589192-22589192 on Build GRCh38
1180273 c.1422G>T p.R474S Chr.1: 22589313-22589313 on Build GRCh38
5762699 c.1337C>T p.P446L Chr.1: 22589228-22589228 on Build GRCh38
99210 c.1474C>T p.L492F Chr.1: 22589365-22589365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148808910 Chr.1: 22589363-22589363 on Build GRCh38
rs79330282 Chr.1: 22589398-22589398 on Build GRCh38
rs752807373 Chr.1: 22589267-22589267 on Build GRCh38
rs775010299 Chr.1: 22589223-22589223 on Build GRCh38
rs755696598 Chr.1: 22589310-22589310 on Build GRCh38
rs111448114 Chr.1: 22589295-22589295 on Build GRCh38
rs779185075 Chr.1: 22589373-22589373 on Build GRCh38
rs187066665 Chr.1: 22589323-22589323 on Build GRCh38
rs150728777 Chr.1: 22589253-22589253 on Build GRCh38
rs1023539465 Chr.1: 22589389-22589389 on Build GRCh38
rs1010783798 Chr.1: 22589384-22589384 on Build GRCh38
rs76452513 Chr.1: 22589365-22589365 on Build GRCh38
rs539053113 Chr.1: 22589396-22589396 on Build GRCh38
rs761428571 Chr.1: 22589313-22589313 on Build GRCh38
rs746707886 Chr.1: 22589307-22589307 on Build GRCh38
rs111930583 Chr.1: 22589290-22589290 on Build GRCh38
rs759290601 Chr.1: 22589358-22589358 on Build GRCh38
rs753688757 Chr.1: 22589216-22589216 on Build GRCh38
rs775864262 Chr.1: 22589363-22589364 on Build GRCh38
rs143644743 Chr.1: 22589228-22589228 on Build GRCh38
rs750322468 Chr.1: 22589233-22589233 on Build GRCh38
rs768339450 Chr.1: 22589225-22589225 on Build GRCh38
rs267598407 Chr.1: 22589275-22589275 on Build GRCh38
rs751791909 Chr.1: 22589249-22589249 on Build GRCh38
rs199679973 Chr.1: 22589224-22589224 on Build GRCh38
rs540588884 Chr.1: 22589231-22589231 on Build GRCh38
rs749738392 Chr.1: 22589292-22589292 on Build GRCh38
rs781002626 Chr.1: 22589390-22589390 on Build GRCh38
rs199965008 Chr.1: 22589212-22589212 on Build GRCh38
rs969158306 Chr.1: 22589248-22589248 on Build GRCh38
rs756141499 Chr.1: 22589289-22589289 on Build GRCh38
rs769286675 Chr.1: 22589318-22589318 on Build GRCh38
rs1023429699 Chr.1: 22589246-22589246 on Build GRCh38
rs547155819 Chr.1: 22589344-22589344 on Build GRCh38
rs766127390 Chr.1: 22589242-22589242 on Build GRCh38
rs200753371 Chr.1: 22589399-22589402 on Build GRCh38
rs146096622 Chr.1: 22589229-22589229 on Build GRCh38
rs1016134236 Chr.1: 22589202-22589202 on Build GRCh38
rs754360957 Chr.1: 22589370-22589370 on Build GRCh38
rs748031383 Chr.1: 22589406-22589406 on Build GRCh38
rs771272070 Chr.1: 22589215-22589215 on Build GRCh38
rs771267594 Chr.1: 22589294-22589294 on Build GRCh38
rs146978261 Chr.1: 22589213-22589213 on Build GRCh38
rs762667401 Chr.1: 22589234-22589234 on Build GRCh38
rs762939441 Chr.1: 22589334-22589334 on Build GRCh38
rs776382045 Chr.1: 22589312-22589312 on Build GRCh38
rs746264367 Chr.1: 22589374-22589374 on Build GRCh38
rs774143169 Chr.1: 22589403-22589403 on Build GRCh38
rs112975371 Chr.1: 22589362-22589362 on Build GRCh38
rs606002 Chr.1: 22589260-22589260 on Build GRCh38
rs766141447 Chr.1: 22589339-22589339 on Build GRCh38
rs903297087 Chr.1: 22589359-22589359 on Build GRCh38
rs772761765 Chr.1: 22589404-22589404 on Build GRCh38

View Full Product Details