Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65086717-65086964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFPL1
Entrez Gene ID
7542
Gene Name
zinc finger protein like 1
Gene Aliases
D11S750, MCG4
UniGene
Hs.712317
Gene Chromosome Location
Chr.11: 65084222-65088402 on Build GRCh38
Transcripts
NM_006782.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7128280 c.452C>T p.S151F Chr.11: 65086763-65086763 on Build GRCh38
429582 c.459C>T p.F153F Chr.11: 65086770-65086770 on Build GRCh38
7154029 c.454G>C p.D152H Chr.11: 65086765-65086765 on Build GRCh38
5938305 c.412G>A p.D138N Chr.11: 65086723-65086723 on Build GRCh38
4742688 c.434C>T p.P145L Chr.11: 65086745-65086745 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770194000 Chr.11: 65086802-65086802 on Build GRCh38
rs746612299 Chr.11: 65086761-65086761 on Build GRCh38
rs1019089364 Chr.11: 65086819-65086819 on Build GRCh38
rs765186357 Chr.11: 65086886-65086886 on Build GRCh38
rs199991849 Chr.11: 65086814-65086814 on Build GRCh38
rs976439765 Chr.11: 65086943-65086943 on Build GRCh38
rs145973436 Chr.11: 65086760-65086760 on Build GRCh38
rs780342788 Chr.11: 65086927-65086927 on Build GRCh38
rs779082095 Chr.11: 65086798-65086798 on Build GRCh38
rs763596298 Chr.11: 65086891-65086891 on Build GRCh38
rs767873757 Chr.11: 65086834-65086834 on Build GRCh38
rs749748214 Chr.11: 65086941-65086941 on Build GRCh38
rs373141594 Chr.11: 65086812-65086812 on Build GRCh38
rs778971131 Chr.11: 65086753-65086753 on Build GRCh38
rs267603112 Chr.11: 65086750-65086750 on Build GRCh38
rs913244212 Chr.11: 65086938-65086938 on Build GRCh38
rs756858400 Chr.11: 65086907-65086907 on Build GRCh38
rs1015192358 Chr.11: 65086880-65086880 on Build GRCh38
rs369925125 Chr.11: 65086751-65086751 on Build GRCh38
rs746257308 Chr.11: 65086800-65086800 on Build GRCh38
rs770578313 Chr.11: 65086777-65086777 on Build GRCh38
rs781498613 Chr.11: 65086779-65086779 on Build GRCh38
rs751065711 Chr.11: 65086905-65086905 on Build GRCh38
rs745560684 Chr.11: 65086924-65086924 on Build GRCh38
rs768769309 Chr.11: 65086816-65086816 on Build GRCh38
rs137924741 Chr.11: 65086756-65086756 on Build GRCh38
rs762052540 Chr.11: 65086829-65086829 on Build GRCh38
rs996429135 Chr.11: 65086862-65086862 on Build GRCh38
rs752864609 Chr.11: 65086815-65086818 on Build GRCh38
rs200454788 Chr.11: 65086747-65086747 on Build GRCh38
rs756466788 Chr.11: 65086925-65086925 on Build GRCh38
rs763439454 Chr.11: 65086813-65086813 on Build GRCh38
rs780804160 Chr.11: 65086908-65086908 on Build GRCh38
rs758279887 Chr.11: 65086890-65086890 on Build GRCh38
rs541397295 Chr.11: 65086884-65086884 on Build GRCh38
rs766209202 Chr.11: 65086746-65086746 on Build GRCh38
rs111719807 Chr.11: 65086743-65086743 on Build GRCh38
rs141258658 Chr.11: 65086757-65086757 on Build GRCh38
rs542277268 Chr.11: 65086889-65086889 on Build GRCh38
rs886678879 Chr.11: 65086856-65086856 on Build GRCh38
rs114865934 Chr.11: 65086870-65086870 on Build GRCh38
rs373973890 Chr.11: 65086844-65086844 on Build GRCh38

View Full Product Details