Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 169594006-169594267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPDL1
Entrez Gene ID
54908
Gene Name
spindle apparatus coiled-coil protein 1
Gene Aliases
CCDC99
UniGene
Hs.368710
Gene Chromosome Location
Chr.5: 169583634-169604778 on Build GRCh38
Transcripts
NM_017785.4 XM_011534585.1 XM_011534586.2 XM_011534587.2 XM_011534588.2 XM_011534589.2 XM_011534590.2 XM_017009601.1 XM_017009602.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5687067 c.566A>G p.Y189C Chr.5: 169594179-169594179 on Build GRCh38
3919657 c.589C>T p.L197F Chr.5: 169594202-169594202 on Build GRCh38
7198017 c.631G>A p.E211K Chr.5: 169594244-169594244 on Build GRCh38
6170156 c.541A>G p.K181E Chr.5: 169594154-169594154 on Build GRCh38
6992413 c.630G>C p.E210D Chr.5: 169594243-169594243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767162543 Chr.5: 169594183-169594183 on Build GRCh38
rs567203657 Chr.5: 169594174-169594174 on Build GRCh38
rs963771300 Chr.5: 169594039-169594039 on Build GRCh38
rs748668253 Chr.5: 169594108-169594109 on Build GRCh38
rs1039930085 Chr.5: 169594164-169594164 on Build GRCh38
rs370771460 Chr.5: 169594243-169594243 on Build GRCh38
rs753781869 Chr.5: 169594220-169594220 on Build GRCh38
rs889785752 Chr.5: 169594069-169594069 on Build GRCh38
rs759219679 Chr.5: 169594100-169594100 on Build GRCh38
rs768912342 Chr.5: 169594150-169594150 on Build GRCh38
rs763821723 Chr.5: 169594214-169594214 on Build GRCh38
rs773895791 Chr.5: 169594176-169594176 on Build GRCh38
rs747263372 Chr.5: 169594146-169594146 on Build GRCh38
rs773770792 Chr.5: 169594233-169594233 on Build GRCh38
rs777721837 Chr.5: 169594132-169594132 on Build GRCh38
rs532499291 Chr.5: 169594037-169594037 on Build GRCh38
rs75861463 Chr.5: 169594175-169594175 on Build GRCh38
rs772647075 Chr.5: 169594154-169594156 on Build GRCh38
rs756081335 Chr.5: 169594124-169594124 on Build GRCh38
rs17644562 Chr.5: 169594094-169594094 on Build GRCh38
rs752302635 Chr.5: 169594195-169594195 on Build GRCh38
rs773869963 Chr.5: 169594173-169594173 on Build GRCh38
rs142049273 Chr.5: 169594229-169594229 on Build GRCh38
rs753730976 Chr.5: 169594136-169594136 on Build GRCh38
rs774814762 Chr.5: 169594103-169594103 on Build GRCh38
rs754711198 Chr.5: 169594141-169594141 on Build GRCh38
rs933291309 Chr.5: 169594120-169594120 on Build GRCh38
rs138841185 Chr.5: 169594221-169594221 on Build GRCh38
rs140051190 Chr.5: 169594106-169594106 on Build GRCh38
rs547263443 Chr.5: 169594142-169594142 on Build GRCh38
rs972152973 Chr.5: 169594079-169594079 on Build GRCh38
rs529963287 Chr.5: 169594163-169594163 on Build GRCh38
rs55679721 Chr.5: 169594122-169594122 on Build GRCh38
rs751638286 Chr.5: 169594222-169594222 on Build GRCh38
rs760489927 Chr.5: 169594203-169594203 on Build GRCh38
rs144060585 Chr.5: 169594161-169594161 on Build GRCh38
rs370986996 Chr.5: 169594165-169594165 on Build GRCh38
rs748320895 Chr.5: 169594231-169594231 on Build GRCh38
rs146344218 Chr.5: 169594232-169594232 on Build GRCh38
rs903708492 Chr.5: 169594175-169594176 on Build GRCh38
rs759158436 Chr.5: 169594177-169594177 on Build GRCh38
rs755197562 Chr.5: 169594225-169594225 on Build GRCh38

View Full Product Details