Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24505871-24506132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMA1
Entrez Gene ID
1215
Gene Name
chymase 1
Gene Aliases
CYH, MCT1, chymase
UniGene
Hs.135626
Gene Chromosome Location
Chr.14: 24505355-24508265 on Build GRCh38
Transcripts
NM_001308083.1 NM_001836.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7415693 c.544G>T p.D182Y Chr.14: 24506084-24506084 on Build GRCh38
5460294 c.521C>A p.A174D Chr.14: 24506107-24506107 on Build GRCh38
5823430 c.521C>T p.A174V Chr.14: 24506107-24506107 on Build GRCh38
3495561 c.528C>T p.S176S Chr.14: 24506100-24506100 on Build GRCh38
400496 c.569G>A p.G190D Chr.14: 24506059-24506059 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775479602 Chr.14: 24506103-24506103 on Build GRCh38
rs772131645 Chr.14: 24506096-24506096 on Build GRCh38
rs760955763 Chr.14: 24506032-24506032 on Build GRCh38
rs1007325607 Chr.14: 24505895-24505895 on Build GRCh38
rs765175474 Chr.14: 24506106-24506106 on Build GRCh38
rs1024717800 Chr.14: 24505906-24505906 on Build GRCh38
rs545333425 Chr.14: 24505945-24505945 on Build GRCh38
rs202117854 Chr.14: 24506065-24506065 on Build GRCh38
rs764063229 Chr.14: 24505984-24505984 on Build GRCh38
rs761749471 Chr.14: 24506104-24506104 on Build GRCh38
rs2073335 Chr.14: 24505955-24505955 on Build GRCh38
rs749623365 Chr.14: 24506077-24506077 on Build GRCh38
rs971949686 Chr.14: 24506013-24506013 on Build GRCh38
rs546717814 Chr.14: 24505909-24505909 on Build GRCh38
rs199514255 Chr.14: 24505975-24505975 on Build GRCh38
rs567503555 Chr.14: 24506070-24506070 on Build GRCh38
rs1053346932 Chr.14: 24506108-24506108 on Build GRCh38
rs971301085 Chr.14: 24505935-24505935 on Build GRCh38
rs558842883 Chr.14: 24506088-24506088 on Build GRCh38
rs751158495 Chr.14: 24506059-24506059 on Build GRCh38
rs201312283 Chr.14: 24506081-24506081 on Build GRCh38
rs754514977 Chr.14: 24506060-24506060 on Build GRCh38
rs750177234 Chr.14: 24506030-24506030 on Build GRCh38
rs753759568 Chr.14: 24505991-24505991 on Build GRCh38
rs994137637 Chr.14: 24505904-24505904 on Build GRCh38
rs148274161 Chr.14: 24506029-24506029 on Build GRCh38
rs919212972 Chr.14: 24506027-24506027 on Build GRCh38
rs373415040 Chr.14: 24505933-24505933 on Build GRCh38
rs754233059 Chr.14: 24506078-24506078 on Build GRCh38
rs200745820 Chr.14: 24506008-24506008 on Build GRCh38
rs758012375 Chr.14: 24506035-24506035 on Build GRCh38
rs898850371 Chr.14: 24505900-24505900 on Build GRCh38
rs756159421 Chr.14: 24506088-24506088 on Build GRCh38
rs778019528 Chr.14: 24506071-24506071 on Build GRCh38
rs1037343171 Chr.14: 24505899-24505899 on Build GRCh38
rs761661153 Chr.14: 24506019-24506019 on Build GRCh38
rs908833857 Chr.14: 24505959-24505959 on Build GRCh38
rs190452117 Chr.14: 24505946-24505946 on Build GRCh38
rs980566264 Chr.14: 24505956-24505956 on Build GRCh38
rs746001351 Chr.14: 24506092-24506092 on Build GRCh38
rs764394472 Chr.14: 24506054-24506054 on Build GRCh38

View Full Product Details