Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 100144964-100145225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO43
Entrez Gene ID
286151
Gene Name
F-box protein 43
Gene Aliases
EMI2, ERP1, FBX43
UniGene
Hs.339577
Gene Chromosome Location
Chr.8: 100133360-100150569 on Build GRCh38
Transcripts
NM_001029860.3 NR_036491.1 XM_011516987.2 XM_011516989.2 XM_011516990.1 XM_011516992.2 XM_017013358.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4996884 c.74G>T p.R25I Chr.8: 100145062-100145062 on Build GRCh38
5686836 c.62C>T p.S21F Chr.8: 100145074-100145074 on Build GRCh38
5359009 c.61T>G p.S21A Chr.8: 100145075-100145075 on Build GRCh38
364108 c.26G>C p.R9T Chr.8: 100145110-100145110 on Build GRCh38
1094311 c.26G>T p.R9I Chr.8: 100145110-100145110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899320244 Chr.8: 100145033-100145033 on Build GRCh38
rs190807307 Chr.8: 100145177-100145177 on Build GRCh38
rs751661086 Chr.8: 100145023-100145023 on Build GRCh38
rs748617183 Chr.8: 100145125-100145125 on Build GRCh38
rs763375843 Chr.8: 100145142-100145142 on Build GRCh38
rs753634464 Chr.8: 100145183-100145183 on Build GRCh38
rs764804996 Chr.8: 100145021-100145023 on Build GRCh38
rs993775516 Chr.8: 100145045-100145045 on Build GRCh38
rs771679413 Chr.8: 100145095-100145095 on Build GRCh38
rs774613362 Chr.8: 100145152-100145152 on Build GRCh38
rs778469182 Chr.8: 100145184-100145184 on Build GRCh38
rs965520090 Chr.8: 100145056-100145056 on Build GRCh38
rs777706539 Chr.8: 100145152-100145153 on Build GRCh38
rs185164902 Chr.8: 100145148-100145148 on Build GRCh38
rs750680749 Chr.8: 100145001-100145001 on Build GRCh38
rs767494590 Chr.8: 100145050-100145050 on Build GRCh38
rs766489675 Chr.8: 100145014-100145014 on Build GRCh38
rs978854471 Chr.8: 100145192-100145192 on Build GRCh38
rs965189366 Chr.8: 100145179-100145179 on Build GRCh38
rs1025824704 Chr.8: 100145123-100145123 on Build GRCh38
rs1032612607 Chr.8: 100145193-100145195 on Build GRCh38
rs763028270 Chr.8: 100145006-100145006 on Build GRCh38
rs200585882 Chr.8: 100145131-100145131 on Build GRCh38
rs765658140 Chr.8: 100145000-100145000 on Build GRCh38
rs777253188 Chr.8: 100144992-100144992 on Build GRCh38
rs143939170 Chr.8: 100145068-100145068 on Build GRCh38
rs752229751 Chr.8: 100145087-100145087 on Build GRCh38
rs755922881 Chr.8: 100145072-100145072 on Build GRCh38
rs2514689 Chr.8: 100144999-100144999 on Build GRCh38
rs755039850 Chr.8: 100145026-100145026 on Build GRCh38
rs752639915 Chr.8: 100145169-100145169 on Build GRCh38
rs748971141 Chr.8: 100145074-100145074 on Build GRCh38
rs773386263 Chr.8: 100145132-100145132 on Build GRCh38
rs1026587294 Chr.8: 100145047-100145047 on Build GRCh38
rs778614662 Chr.8: 100145091-100145091 on Build GRCh38
rs76557796 Chr.8: 100145168-100145168 on Build GRCh38
rs755975849 Chr.8: 100145171-100145171 on Build GRCh38
rs756925561 Chr.8: 100145087-100145087 on Build GRCh38
rs777312050 Chr.8: 100145100-100145100 on Build GRCh38
rs904943801 Chr.8: 100144997-100144997 on Build GRCh38
rs758127396 Chr.8: 100145137-100145137 on Build GRCh38
rs745417810 Chr.8: 100145093-100145093 on Build GRCh38
rs890685002 Chr.8: 100145144-100145144 on Build GRCh38
rs752588384 Chr.8: 100145054-100145054 on Build GRCh38
rs759640054 Chr.8: 100145163-100145163 on Build GRCh38

View Full Product Details