Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27053843-27054114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL5
Entrez Gene ID
60509
Gene Name
ATP/GTP binding protein like 5
Gene Aliases
CCP5, RP75
UniGene
Hs.138207
Gene Chromosome Location
Chr.2: 27051423-27070622 on Build GRCh38
Transcripts
NM_001035507.2 NM_021831.5 NR_104246.1 XM_005264477.2 XM_011533011.2 XM_011533012.2 XM_011533013.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7293878 c.518A>G p.Q173R Chr.2: 27054026-27054026 on Build GRCh38
4660343 c.424C>T p.R142C Chr.2: 27053932-27053932 on Build GRCh38
1592038 c.414C>A p.S138S Chr.2: 27053922-27053922 on Build GRCh38
7118472 c.487G>A p.D163N Chr.2: 27053995-27053995 on Build GRCh38
4660344 c.424C>T p.R142C Chr.2: 27053932-27053932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377519364 Chr.2: 27054064-27054064 on Build GRCh38
rs766782738 Chr.2: 27054087-27054087 on Build GRCh38
rs533283908 Chr.2: 27053907-27053907 on Build GRCh38
rs750887726 Chr.2: 27053950-27053950 on Build GRCh38
rs144135243 Chr.2: 27053947-27053947 on Build GRCh38
rs774398504 Chr.2: 27053985-27053987 on Build GRCh38
rs749127946 Chr.2: 27054012-27054012 on Build GRCh38
rs139319795 Chr.2: 27054036-27054036 on Build GRCh38
rs753974793 Chr.2: 27053935-27053935 on Build GRCh38
rs768583700 Chr.2: 27053941-27053941 on Build GRCh38
rs754521080 Chr.2: 27053976-27053976 on Build GRCh38
rs766150855 Chr.2: 27053968-27053968 on Build GRCh38
rs777483018 Chr.2: 27053920-27053920 on Build GRCh38
rs769641954 Chr.2: 27053906-27053906 on Build GRCh38
rs143002099 Chr.2: 27053937-27053937 on Build GRCh38
rs375924230 Chr.2: 27053885-27053885 on Build GRCh38
rs780508825 Chr.2: 27053982-27053982 on Build GRCh38
rs140994852 Chr.2: 27053992-27053992 on Build GRCh38
rs756068995 Chr.2: 27053999-27053999 on Build GRCh38
rs749075984 Chr.2: 27053934-27053934 on Build GRCh38
rs765762435 Chr.2: 27053948-27053948 on Build GRCh38
rs144657011 Chr.2: 27054003-27054003 on Build GRCh38
rs984315169 Chr.2: 27054060-27054060 on Build GRCh38
rs747968599 Chr.2: 27053887-27053887 on Build GRCh38
rs541762268 Chr.2: 27054069-27054069 on Build GRCh38
rs371065936 Chr.2: 27054089-27054089 on Build GRCh38
rs142859717 Chr.2: 27054035-27054035 on Build GRCh38
rs3739090 Chr.2: 27053938-27053938 on Build GRCh38
rs746726078 Chr.2: 27054085-27054085 on Build GRCh38
rs780288772 Chr.2: 27054024-27054024 on Build GRCh38
rs751215304 Chr.2: 27053970-27053970 on Build GRCh38
rs922300627 Chr.2: 27053874-27053875 on Build GRCh38
rs981921240 Chr.2: 27054028-27054028 on Build GRCh38
rs770666370 Chr.2: 27054013-27054013 on Build GRCh38
rs747172298 Chr.2: 27054029-27054029 on Build GRCh38
rs761948167 Chr.2: 27054037-27054037 on Build GRCh38
rs763380664 Chr.2: 27053964-27053964 on Build GRCh38
rs141068963 Chr.2: 27054054-27054054 on Build GRCh38
rs777654089 Chr.2: 27054009-27054009 on Build GRCh38

View Full Product Details