Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 184936521-184936764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF804A
Entrez Gene ID
91752
Gene Name
zinc finger protein 804A
Gene Aliases
C2orf10
UniGene
Hs.159528
Gene Chromosome Location
Chr.2: 184598366-184939487 on Build GRCh38
Transcripts
NM_194250.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6089900 c.1285C>T p.L429F Chr.2: 184936681-184936681 on Build GRCh38
5569929 c.1229C>A p.T410N Chr.2: 184936625-184936625 on Build GRCh38
5577233 c.1238A>G p.K413R Chr.2: 184936634-184936634 on Build GRCh38
4136756 c.1321T>C p.S441P Chr.2: 184936717-184936717 on Build GRCh38
3981830 c.1207A>C p.N403H Chr.2: 184936603-184936603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750343969 Chr.2: 184936624-184936624 on Build GRCh38
rs752188521 Chr.2: 184936658-184936658 on Build GRCh38
rs746667804 Chr.2: 184936578-184936578 on Build GRCh38
rs568988606 Chr.2: 184936615-184936615 on Build GRCh38
rs769523473 Chr.2: 184936549-184936549 on Build GRCh38
rs746374699 Chr.2: 184936706-184936706 on Build GRCh38
rs749835146 Chr.2: 184936585-184936585 on Build GRCh38
rs141114551 Chr.2: 184936616-184936616 on Build GRCh38
rs754609316 Chr.2: 184936632-184936632 on Build GRCh38
rs750149492 Chr.2: 184936556-184936556 on Build GRCh38
rs903864004 Chr.2: 184936564-184936564 on Build GRCh38
rs762799601 Chr.2: 184936628-184936628 on Build GRCh38
rs772365402 Chr.2: 184936724-184936724 on Build GRCh38
rs772552984 Chr.2: 184936593-184936593 on Build GRCh38
rs151226103 Chr.2: 184936562-184936562 on Build GRCh38
rs371251154 Chr.2: 184936606-184936606 on Build GRCh38
rs780598334 Chr.2: 184936642-184936642 on Build GRCh38
rs892716124 Chr.2: 184936603-184936603 on Build GRCh38
rs191570335 Chr.2: 184936565-184936565 on Build GRCh38
rs985576496 Chr.2: 184936681-184936681 on Build GRCh38
rs751217226 Chr.2: 184936631-184936631 on Build GRCh38
rs757852500 Chr.2: 184936664-184936664 on Build GRCh38
rs774861402 Chr.2: 184936589-184936589 on Build GRCh38
rs62198466 Chr.2: 184936621-184936621 on Build GRCh38
rs779623841 Chr.2: 184936694-184936694 on Build GRCh38
rs766166181 Chr.2: 184936629-184936629 on Build GRCh38
rs535681707 Chr.2: 184936574-184936574 on Build GRCh38
rs866891873 Chr.2: 184936567-184936567 on Build GRCh38
rs760922921 Chr.2: 184936613-184936613 on Build GRCh38
rs746179906 Chr.2: 184936590-184936590 on Build GRCh38
rs34345078 Chr.2: 184936584-184936584 on Build GRCh38

View Full Product Details