Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 134568119-134568364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1B15
Entrez Gene ID
441282
Gene Name
aldo-keto reductase family 1 member B15
Gene Aliases
AKR1B10L, AKR1B10L, AK1R1B7, AKR1R1B7
UniGene
Hs.729418
Gene Chromosome Location
Chr.7: 134549097-134579841 on Build GRCh38
Transcripts
NM_001080538.2 XM_011516241.2 XM_017012224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5884490 c.229A>T p.S77C Chr.7: 134568320-134568320 on Build GRCh38
3698186 c.159C>T p.L53L Chr.7: 134568166-134568166 on Build GRCh38
5822637 c.173C>T p.A58V Chr.7: 134568264-134568264 on Build GRCh38
7257575 c.206T>A p.M69K Chr.7: 134568297-134568297 on Build GRCh38
5959940 c.146A>G p.Y49C Chr.7: 134568237-134568237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767349208 Chr.7: 134568153-134568153 on Build GRCh38
rs770750822 Chr.7: 134568331-134568331 on Build GRCh38
rs763742509 Chr.7: 134568344-134568344 on Build GRCh38
rs370933423 Chr.7: 134568250-134568250 on Build GRCh38
rs781699665 Chr.7: 134568237-134568237 on Build GRCh38
rs370337877 Chr.7: 134568286-134568286 on Build GRCh38
rs760551823 Chr.7: 134568260-134568260 on Build GRCh38
rs778659679 Chr.7: 134568165-134568165 on Build GRCh38
rs368093454 Chr.7: 134568274-134568274 on Build GRCh38
rs201762103 Chr.7: 134568183-134568183 on Build GRCh38
rs778340418 Chr.7: 134568221-134568221 on Build GRCh38
rs373511828 Chr.7: 134568212-134568212 on Build GRCh38
rs775667669 Chr.7: 134568192-134568192 on Build GRCh38
rs766363195 Chr.7: 134568213-134568213 on Build GRCh38
rs200408342 Chr.7: 134568300-134568300 on Build GRCh38
rs1037987094 Chr.7: 134568321-134568321 on Build GRCh38
rs368705788 Chr.7: 134568332-134568332 on Build GRCh38
rs201443523 Chr.7: 134568301-134568301 on Build GRCh38
rs779362790 Chr.7: 134568159-134568159 on Build GRCh38
rs747792903 Chr.7: 134568225-134568225 on Build GRCh38
rs1006481279 Chr.7: 134568160-134568160 on Build GRCh38
rs766128986 Chr.7: 134568261-134568261 on Build GRCh38
rs771300283 Chr.7: 134568235-134568235 on Build GRCh38
rs765268945 Chr.7: 134568191-134568191 on Build GRCh38
rs185318158 Chr.7: 134568184-134568184 on Build GRCh38
rs866264269 Chr.7: 134568155-134568155 on Build GRCh38
rs761991426 Chr.7: 134568329-134568329 on Build GRCh38
rs772945102 Chr.7: 134568259-134568259 on Build GRCh38
rs79024112 Chr.7: 134568167-134568167 on Build GRCh38
rs750221215 Chr.7: 134568210-134568210 on Build GRCh38
rs753247888 Chr.7: 134568215-134568215 on Build GRCh38
rs756109051 Chr.7: 134568157-134568157 on Build GRCh38
rs372007991 Chr.7: 134568336-134568336 on Build GRCh38
rs754480745 Chr.7: 134568217-134568217 on Build GRCh38
rs530258799 Chr.7: 134568317-134568317 on Build GRCh38
rs767294095 Chr.7: 134568207-134568207 on Build GRCh38
rs747847872 Chr.7: 134568166-134568166 on Build GRCh38
rs150714393 Chr.7: 134568337-134568337 on Build GRCh38
rs373958065 Chr.7: 134568299-134568299 on Build GRCh38
rs990125142 Chr.7: 134568263-134568263 on Build GRCh38
rs190141715 Chr.7: 134568205-134568205 on Build GRCh38
rs191960014 Chr.7: 134568162-134568162 on Build GRCh38
rs803473 Chr.7: 134568148-134568148 on Build GRCh38
rs201714104 Chr.7: 134568290-134568290 on Build GRCh38
rs375294848 Chr.7: 134568170-134568170 on Build GRCh38
rs758072997 Chr.7: 134568288-134568288 on Build GRCh38
rs747398680 Chr.7: 134568233-134568233 on Build GRCh38
rs763093453 Chr.7: 134568255-134568255 on Build GRCh38
rs553102909 Chr.7: 134568297-134568297 on Build GRCh38
rs377496966 Chr.7: 134568245-134568245 on Build GRCh38
rs758195535 Chr.7: 134568227-134568227 on Build GRCh38
rs375980254 Chr.7: 134568316-134568316 on Build GRCh38
rs746407140 Chr.7: 134568238-134568238 on Build GRCh38
rs539015857 Chr.7: 134568277-134568277 on Build GRCh38
rs369033836 Chr.7: 134568174-134568174 on Build GRCh38
rs770204962 Chr.7: 134568304-134568304 on Build GRCh38
rs780049138 Chr.7: 134568306-134568306 on Build GRCh38
rs201701802 Chr.7: 134568314-134568314 on Build GRCh38
rs1047738588 Chr.7: 134568158-134568158 on Build GRCh38
rs1025120754 Chr.7: 134568181-134568181 on Build GRCh38
rs368533580 Chr.7: 134568173-134568173 on Build GRCh38
rs753807222 Chr.7: 134568180-134568180 on Build GRCh38
rs778219019 Chr.7: 134568231-134568231 on Build GRCh38
rs369738819 Chr.7: 134568156-134568156 on Build GRCh38
rs762519183 Chr.7: 134568338-134568338 on Build GRCh38
rs10260266 Chr.7: 134568262-134568262 on Build GRCh38

View Full Product Details