Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 9216811-9217036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD12
Entrez Gene ID
23253
Gene Name
ankyrin repeat domain 12
Gene Aliases
ANCO-2, ANCO1, GAC-1, Nbla00144
UniGene
Hs.464585
Gene Chromosome Location
Chr.18: 9136753-9285985 on Build GRCh38
Transcripts
NM_001083625.2 NM_001204056.1 NM_015208.4 XM_005258092.3 XM_005258093.3 XM_005258095.2 XM_005258096.2 XM_011525638.2 XM_011525639.2 XM_017025661.1 XM_017025662.1 XM_017025663.1 XM_017025664.1 XM_017025665.1 XM_017025666.1 XM_017025667.1 XM_017025668.1 XM_017025670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1630677 c.766C>A p.H256N Chr.18: 9216871-9216871 on Build GRCh38
5673572 c.738A>T p.T246T Chr.18: 9216843-9216843 on Build GRCh38
6083928 c.722G>C p.G241A Chr.18: 9216827-9216827 on Build GRCh38
6575291 c.760C>G p.P254A Chr.18: 9216865-9216865 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779423682 Chr.18: 9216876-9216876 on Build GRCh38
rs748590274 Chr.18: 9216910-9216910 on Build GRCh38
rs758715753 Chr.18: 9216946-9216947 on Build GRCh38
rs753458079 Chr.18: 9216837-9216837 on Build GRCh38
rs1047144380 Chr.18: 9216957-9216957 on Build GRCh38
rs896538103 Chr.18: 9216998-9216998 on Build GRCh38
rs555602158 Chr.18: 9216924-9216924 on Build GRCh38
rs768851378 Chr.18: 9216952-9216953 on Build GRCh38
rs1026733938 Chr.18: 9216949-9216949 on Build GRCh38
rs1046051488 Chr.18: 9216954-9216954 on Build GRCh38
rs747077016 Chr.18: 9216933-9216933 on Build GRCh38
rs377340834 Chr.18: 9216944-9216944 on Build GRCh38
rs776861281 Chr.18: 9216939-9216939 on Build GRCh38
rs772693726 Chr.18: 9216922-9216922 on Build GRCh38
rs939329116 Chr.18: 9216974-9216974 on Build GRCh38
rs897675038 Chr.18: 9216911-9216911 on Build GRCh38
rs750820801 Chr.18: 9216919-9216922 on Build GRCh38
rs747171321 Chr.18: 9216948-9216949 on Build GRCh38
rs759760717 Chr.18: 9216942-9216942 on Build GRCh38
rs1037609650 Chr.18: 9216840-9216840 on Build GRCh38
rs906573155 Chr.18: 9216970-9216970 on Build GRCh38
rs755335130 Chr.18: 9216843-9216843 on Build GRCh38
rs1055163382 Chr.18: 9216997-9216997 on Build GRCh38
rs796348697 Chr.18: 9216946-9216947 on Build GRCh38
rs770944646 Chr.18: 9216935-9216935 on Build GRCh38
rs954165888 Chr.18: 9216971-9216971 on Build GRCh38

View Full Product Details