Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37724162-37724421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMPD1
Entrez Gene ID
22844
Gene Name
FERM and PDZ domain containing 1
Gene Aliases
FRMD2
UniGene
Hs.163990
Gene Chromosome Location
Chr.9: 37602979-37746904 on Build GRCh38
Transcripts
NM_014907.2 XM_011517803.2 XM_011517804.2 XM_011517805.2 XM_011517806.2 XM_017014480.1 XM_017014481.1 XM_017014482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5646486 c.543C>T p.N181N Chr.9: 37724251-37724251 on Build GRCh38
1252803 c.587A>G p.K196R Chr.9: 37724295-37724295 on Build GRCh38
6115909 c.129T>C p.N43N Chr.9: 37724230-37724230 on Build GRCh38
6667059 c.136C>T p.L46L Chr.9: 37724237-37724237 on Build GRCh38
6667058 c.529C>T p.L177L Chr.9: 37724237-37724237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774896122 Chr.9: 37724323-37724323 on Build GRCh38
rs370691170 Chr.9: 37724216-37724216 on Build GRCh38
rs777106632 Chr.9: 37724245-37724245 on Build GRCh38
rs745611500 Chr.9: 37724234-37724234 on Build GRCh38
rs989107715 Chr.9: 37724272-37724272 on Build GRCh38
rs558807322 Chr.9: 37724183-37724184 on Build GRCh38
rs769214377 Chr.9: 37724240-37724240 on Build GRCh38
rs773875210 Chr.9: 37724252-37724252 on Build GRCh38
rs144530485 Chr.9: 37724261-37724261 on Build GRCh38
rs375253000 Chr.9: 37724251-37724251 on Build GRCh38
rs377461996 Chr.9: 37724208-37724208 on Build GRCh38
rs374684887 Chr.9: 37724212-37724212 on Build GRCh38
rs758596354 Chr.9: 37724215-37724215 on Build GRCh38
rs1000559664 Chr.9: 37724194-37724194 on Build GRCh38
rs765428888 Chr.9: 37724337-37724337 on Build GRCh38
rs759137843 Chr.9: 37724262-37724262 on Build GRCh38
rs914920668 Chr.9: 37724324-37724324 on Build GRCh38
rs756794597 Chr.9: 37724184-37724184 on Build GRCh38
rs557385868 Chr.9: 37724314-37724314 on Build GRCh38
rs1050792413 Chr.9: 37724373-37724373 on Build GRCh38
rs748593586 Chr.9: 37724247-37724247 on Build GRCh38
rs757400260 Chr.9: 37724205-37724205 on Build GRCh38
rs865879056 Chr.9: 37724246-37724246 on Build GRCh38
rs775189906 Chr.9: 37724295-37724295 on Build GRCh38
rs753842893 Chr.9: 37724315-37724315 on Build GRCh38
rs1024453314 Chr.9: 37724207-37724207 on Build GRCh38
rs764092584 Chr.9: 37724191-37724191 on Build GRCh38
rs267602237 Chr.9: 37724320-37724320 on Build GRCh38
rs751923195 Chr.9: 37724367-37724367 on Build GRCh38
rs767180692 Chr.9: 37724263-37724263 on Build GRCh38
rs758588552 Chr.9: 37724338-37724338 on Build GRCh38
rs370662004 Chr.9: 37724193-37724193 on Build GRCh38
rs940290593 Chr.9: 37724377-37724377 on Build GRCh38
rs113684935 Chr.9: 37724343-37724343 on Build GRCh38
rs111582728 Chr.9: 37724209-37724209 on Build GRCh38
rs760515210 Chr.9: 37724312-37724312 on Build GRCh38
rs867386095 Chr.9: 37724224-37724224 on Build GRCh38
rs10814609 Chr.9: 37724198-37724198 on Build GRCh38
rs956483033 Chr.9: 37724256-37724256 on Build GRCh38
rs747367402 Chr.9: 37724225-37724225 on Build GRCh38

View Full Product Details