Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3818264-3818493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCBP3
Entrez Gene ID
55421
Gene Name
nuclear cap binding subunit 3
Gene Aliases
C17orf85, ELG, HSA277841
UniGene
Hs.120963
Gene Chromosome Location
Chr.17: 3806748-3846251 on Build GRCh38
Transcripts
NM_001114118.2 XM_005256717.2 XM_006721556.2 XM_011523954.2 XM_011523955.2 XM_017024826.1 XM_017024827.1 XM_017024828.1 XM_017024829.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5002339 c.1207G>T p.D403Y Chr.17: 3818366-3818366 on Build GRCh38
5002340 c.367G>T p.D123Y Chr.17: 3818366-3818366 on Build GRCh38
978528 c.258T>C p.D86D Chr.17: 3818475-3818475 on Build GRCh38
7252162 c.1161C>A p.S387R Chr.17: 3818412-3818412 on Build GRCh38
978526 c.442G>A p.E148K Chr.17: 3818291-3818291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145210828 Chr.17: 3818450-3818450 on Build GRCh38
rs954129329 Chr.17: 3818324-3818324 on Build GRCh38
rs1040936110 Chr.17: 3818467-3818467 on Build GRCh38
rs756021801 Chr.17: 3818408-3818408 on Build GRCh38
rs767246507 Chr.17: 3818386-3818386 on Build GRCh38
rs375218323 Chr.17: 3818435-3818435 on Build GRCh38
rs752633062 Chr.17: 3818401-3818401 on Build GRCh38
rs371676057 Chr.17: 3818423-3818423 on Build GRCh38
rs748033281 Chr.17: 3818346-3818346 on Build GRCh38
rs368863088 Chr.17: 3818405-3818405 on Build GRCh38
rs922482645 Chr.17: 3818468-3818468 on Build GRCh38
rs879066882 Chr.17: 3818412-3818412 on Build GRCh38
rs373070322 Chr.17: 3818409-3818409 on Build GRCh38
rs762032858 Chr.17: 3818463-3818463 on Build GRCh38
rs756893326 Chr.17: 3818411-3818411 on Build GRCh38
rs372415582 Chr.17: 3818417-3818417 on Build GRCh38
rs779504421 Chr.17: 3818302-3818303 on Build GRCh38
rs751377137 Chr.17: 3818381-3818381 on Build GRCh38
rs768720440 Chr.17: 3818340-3818340 on Build GRCh38
rs770109767 Chr.17: 3818472-3818472 on Build GRCh38
rs769105284 Chr.17: 3818437-3818437 on Build GRCh38
rs765612836 Chr.17: 3818362-3818362 on Build GRCh38
rs77869954 Chr.17: 3818461-3818461 on Build GRCh38
rs764423300 Chr.17: 3818366-3818366 on Build GRCh38
rs746293908 Chr.17: 3818295-3818295 on Build GRCh38
rs776852488 Chr.17: 3818357-3818357 on Build GRCh38
rs762118736 Chr.17: 3818358-3818358 on Build GRCh38
rs746202360 Chr.17: 3818422-3818422 on Build GRCh38
rs924972929 Chr.17: 3818440-3818440 on Build GRCh38
rs772440375 Chr.17: 3818334-3818334 on Build GRCh38
rs761120701 Chr.17: 3818335-3818335 on Build GRCh38
rs111458199 Chr.17: 3818410-3818410 on Build GRCh38
rs747480883 Chr.17: 3818436-3818436 on Build GRCh38
rs780292169 Chr.17: 3818426-3818426 on Build GRCh38

View Full Product Details