Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 47809517-47809778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO11
Entrez Gene ID
80204
Gene Name
F-box protein 11
Gene Aliases
FBX11, PRMT9, UBR6, UG063H01, VIT1
UniGene
Hs.352677
Gene Chromosome Location
Chr.2: 47806920-47905793 on Build GRCh38
Transcripts
NM_001190274.1 NM_025133.4 XM_005264572.4 XM_005264573.4 XM_017005015.1 XM_017005016.1 XM_017005017.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1318900 c.2178_2179insTC p.N727fs*3 Chr.2: 47809615-47809616 on Build GRCh38
6258903 c.2351C>A p.T784K Chr.2: 47809695-47809695 on Build GRCh38
6258904 c.2099C>A p.T700K Chr.2: 47809695-47809695 on Build GRCh38
1318899 c.2430_2431insTC p.N811fs*3 Chr.2: 47809615-47809616 on Build GRCh38
4777471 c.2348T>A p.I783N Chr.2: 47809698-47809698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761520519 Chr.2: 47809682-47809682 on Build GRCh38
rs780405624 Chr.2: 47809567-47809568 on Build GRCh38
rs373506263 Chr.2: 47809750-47809750 on Build GRCh38
rs774903128 Chr.2: 47809578-47809578 on Build GRCh38
rs542408171 Chr.2: 47809579-47809579 on Build GRCh38
rs770086772 Chr.2: 47809560-47809560 on Build GRCh38
rs148676301 Chr.2: 47809610-47809610 on Build GRCh38
rs113217589 Chr.2: 47809711-47809711 on Build GRCh38
rs756128545 Chr.2: 47809730-47809730 on Build GRCh38
rs143212725 Chr.2: 47809563-47809563 on Build GRCh38
rs780952250 Chr.2: 47809713-47809713 on Build GRCh38
rs776226469 Chr.2: 47809594-47809594 on Build GRCh38
rs377488125 Chr.2: 47809569-47809569 on Build GRCh38
rs763988532 Chr.2: 47809684-47809684 on Build GRCh38
rs921477509 Chr.2: 47809724-47809724 on Build GRCh38
rs768797614 Chr.2: 47809740-47809740 on Build GRCh38
rs543514855 Chr.2: 47809737-47809737 on Build GRCh38
rs747303426 Chr.2: 47809572-47809579 on Build GRCh38
rs769185982 Chr.2: 47809711-47809715 on Build GRCh38
rs1043332222 Chr.2: 47809735-47809735 on Build GRCh38
rs980579180 Chr.2: 47809547-47809547 on Build GRCh38
rs371182710 Chr.2: 47809748-47809748 on Build GRCh38
rs576110433 Chr.2: 47809736-47809736 on Build GRCh38
rs980079107 Chr.2: 47809629-47809629 on Build GRCh38
rs536023138 Chr.2: 47809582-47809582 on Build GRCh38
rs946336761 Chr.2: 47809697-47809697 on Build GRCh38
rs554668738 Chr.2: 47809636-47809636 on Build GRCh38
rs781350474 Chr.2: 47809557-47809557 on Build GRCh38
rs764977191 Chr.2: 47809637-47809637 on Build GRCh38
rs557498118 Chr.2: 47809685-47809685 on Build GRCh38
rs893463495 Chr.2: 47809613-47809613 on Build GRCh38
rs369083837 Chr.2: 47809554-47809554 on Build GRCh38
rs752266773 Chr.2: 47809653-47809653 on Build GRCh38
rs757286262 Chr.2: 47809688-47809688 on Build GRCh38
rs772509139 Chr.2: 47809583-47809583 on Build GRCh38
rs757196357 Chr.2: 47809555-47809555 on Build GRCh38
rs750556419 Chr.2: 47809714-47809714 on Build GRCh38
rs749713463 Chr.2: 47809565-47809565 on Build GRCh38
rs370048586 Chr.2: 47809683-47809683 on Build GRCh38

View Full Product Details