Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56086460-56086731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERBB3
Entrez Gene ID
2065
Gene Name
erb-b2 receptor tyrosine kinase 3
Gene Aliases
ErbB-3, HER3, LCCS2, MDA-BF-1, c-erbB-3, c-erbB3, erbB3-S, p180-ErbB3, p45-sErbB3, p85-sErbB3
UniGene
Hs.118681
Gene Chromosome Location
Chr.12: 56080025-56103507 on Build GRCh38
Transcripts
NM_001982.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5986904 c.442T>C p.Y148H Chr.12: 56086551-56086551 on Build GRCh38
4390482 c.501G>C p.V167V Chr.12: 56086610-56086610 on Build GRCh38
6854103 c.423G>T p.E141D Chr.12: 56086532-56086532 on Build GRCh38
6655784 c.504G>T p.R168S Chr.12: 56086613-56086613 on Build GRCh38
6923585 c.427C>A p.L143M Chr.12: 56086536-56086536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758421385 Chr.12: 56086608-56086608 on Build GRCh38
rs922342968 Chr.12: 56086483-56086483 on Build GRCh38
rs1003704526 Chr.12: 56086521-56086521 on Build GRCh38
rs781058680 Chr.12: 56086541-56086541 on Build GRCh38
rs573798308 Chr.12: 56086485-56086485 on Build GRCh38
rs766619595 Chr.12: 56086589-56086589 on Build GRCh38
rs762130727 Chr.12: 56086489-56086489 on Build GRCh38
rs141700623 Chr.12: 56086578-56086578 on Build GRCh38
rs780753707 Chr.12: 56086623-56086623 on Build GRCh38
rs925749656 Chr.12: 56086583-56086583 on Build GRCh38
rs976572614 Chr.12: 56086657-56086657 on Build GRCh38
rs115311663 Chr.12: 56086524-56086524 on Build GRCh38
rs759585997 Chr.12: 56086593-56086593 on Build GRCh38
rs373476674 Chr.12: 56086495-56086495 on Build GRCh38
rs560422339 Chr.12: 56086600-56086600 on Build GRCh38
rs562044145 Chr.12: 56086630-56086630 on Build GRCh38
rs79222157 Chr.12: 56086690-56086690 on Build GRCh38
rs755643469 Chr.12: 56086647-56086647 on Build GRCh38
rs113267229 Chr.12: 56086501-56086501 on Build GRCh38
rs199616683 Chr.12: 56086496-56086496 on Build GRCh38
rs773530299 Chr.12: 56086675-56086675 on Build GRCh38
rs760733324 Chr.12: 56086494-56086494 on Build GRCh38
rs75038291 Chr.12: 56086685-56086685 on Build GRCh38
rs374643843 Chr.12: 56086605-56086605 on Build GRCh38
rs986135420 Chr.12: 56086626-56086626 on Build GRCh38
rs746368473 Chr.12: 56086545-56086545 on Build GRCh38
rs188795493 Chr.12: 56086590-56086590 on Build GRCh38
rs777551696 Chr.12: 56086504-56086504 on Build GRCh38
rs961328826 Chr.12: 56086514-56086514 on Build GRCh38
rs879027206 Chr.12: 56086607-56086607 on Build GRCh38
rs745349643 Chr.12: 56086633-56086633 on Build GRCh38
rs759646874 Chr.12: 56086703-56086703 on Build GRCh38
rs780394534 Chr.12: 56086498-56086498 on Build GRCh38
rs747963339 Chr.12: 56086657-56086658 on Build GRCh38
rs147905731 Chr.12: 56086566-56086566 on Build GRCh38
rs772486476 Chr.12: 56086556-56086556 on Build GRCh38
rs774723821 Chr.12: 56086486-56086486 on Build GRCh38
rs929711174 Chr.12: 56086692-56086692 on Build GRCh38
rs769417912 Chr.12: 56086686-56086686 on Build GRCh38
rs751430378 Chr.12: 56086617-56086617 on Build GRCh38
rs770712528 Chr.12: 56086510-56086510 on Build GRCh38
rs748548967 Chr.12: 56086664-56086664 on Build GRCh38
rs1045985721 Chr.12: 56086671-56086671 on Build GRCh38
rs377384549 Chr.12: 56086497-56086497 on Build GRCh38
rs763914719 Chr.12: 56086613-56086613 on Build GRCh38
rs771120188 Chr.12: 56086680-56086680 on Build GRCh38
rs371016430 Chr.12: 56086651-56086651 on Build GRCh38
rs374619740 Chr.12: 56086546-56086546 on Build GRCh38
rs772576333 Chr.12: 56086667-56086667 on Build GRCh38
rs745388282 Chr.12: 56086523-56086523 on Build GRCh38
rs200978269 Chr.12: 56086618-56086618 on Build GRCh38
rs748544096 Chr.12: 56086555-56086555 on Build GRCh38
rs892530465 Chr.12: 56086542-56086542 on Build GRCh38
rs750714635 Chr.12: 56086491-56086491 on Build GRCh38
rs373842106 Chr.12: 56086676-56086676 on Build GRCh38
rs373295408 Chr.12: 56086506-56086506 on Build GRCh38
rs147412657 Chr.12: 56086565-56086565 on Build GRCh38
rs767692749 Chr.12: 56086490-56086490 on Build GRCh38
rs780804718 Chr.12: 56086512-56086512 on Build GRCh38
rs1018139675 Chr.12: 56086631-56086631 on Build GRCh38

View Full Product Details