Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14056761-14056996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PALM3
Entrez Gene ID
342979
Gene Name
paralemmin 3
Gene Aliases
-
UniGene
Hs.444298
Gene Chromosome Location
Chr.19: 14053367-14062137 on Build GRCh38
Transcripts
NM_001145028.1 XM_005259891.4 XM_011527965.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7272744 c.166G>A p.A56T Chr.19: 14056765-14056765 on Build GRCh38
1480663 c.151C>A p.L51I Chr.19: 14056780-14056780 on Build GRCh38
7187629 c.134C>G p.S45C Chr.19: 14056797-14056797 on Build GRCh38
5456692 c.139C>T p.R47W Chr.19: 14056792-14056792 on Build GRCh38
4886890 c.166G>T p.A56S Chr.19: 14056765-14056765 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370525556 Chr.19: 14056814-14056814 on Build GRCh38
rs950902432 Chr.19: 14056969-14056969 on Build GRCh38
rs924471247 Chr.19: 14056811-14056811 on Build GRCh38
rs999845718 Chr.19: 14056962-14056962 on Build GRCh38
rs995107863 Chr.19: 14056955-14056955 on Build GRCh38
rs899385987 Chr.19: 14056898-14056898 on Build GRCh38
rs34724208 Chr.19: 14056917-14056918 on Build GRCh38
rs1043246145 Chr.19: 14056795-14056795 on Build GRCh38
rs538396990 Chr.19: 14056847-14056847 on Build GRCh38
rs904226587 Chr.19: 14056959-14056959 on Build GRCh38
rs1056660506 Chr.19: 14056871-14056871 on Build GRCh38
rs753204318 Chr.19: 14056822-14056822 on Build GRCh38
rs200135731 Chr.19: 14056918-14056919 on Build GRCh38
rs759941237 Chr.19: 14056828-14056828 on Build GRCh38
rs939529427 Chr.19: 14056868-14056868 on Build GRCh38
rs111983827 Chr.19: 14056917-14056917 on Build GRCh38
rs78497170 Chr.19: 14056972-14056972 on Build GRCh38
rs945623504 Chr.19: 14056893-14056893 on Build GRCh38
rs138552271 Chr.19: 14056915-14056916 on Build GRCh38
rs934742174 Chr.19: 14056813-14056813 on Build GRCh38
rs758913689 Chr.19: 14056904-14056904 on Build GRCh38
rs1028725861 Chr.19: 14056971-14056971 on Build GRCh38
rs776042636 Chr.19: 14056849-14056849 on Build GRCh38
rs926678676 Chr.19: 14056865-14056865 on Build GRCh38
rs892616692 Chr.19: 14056875-14056875 on Build GRCh38
rs544501422 Chr.19: 14056873-14056873 on Build GRCh38

View Full Product Details