Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34108731-34108962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAT10
Entrez Gene ID
55226
Gene Name
N-acetyltransferase 10
Gene Aliases
ALP, NET43
UniGene
Hs.577281
Gene Chromosome Location
Chr.11: 34105564-34146911 on Build GRCh38
Transcripts
NM_001144030.1 NM_024662.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1746218 c.109G>T p.V37L Chr.11: 34108742-34108742 on Build GRCh38
4032509 c.117A>G p.I39M Chr.11: 34108750-34108750 on Build GRCh38
6722017 c.134C>A p.S45Y Chr.11: 34108767-34108767 on Build GRCh38
1297993 c.154C>T p.R52W Chr.11: 34108787-34108787 on Build GRCh38
926599 c.188T>C p.L63P Chr.11: 34108821-34108821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775666850 Chr.11: 34108841-34108841 on Build GRCh38
rs781170067 Chr.11: 34108798-34108798 on Build GRCh38
rs11552143 Chr.11: 34108795-34108795 on Build GRCh38
rs17700360 Chr.11: 34108859-34108859 on Build GRCh38
rs750385600 Chr.11: 34108885-34108885 on Build GRCh38
rs371337207 Chr.11: 34108857-34108857 on Build GRCh38
rs761355482 Chr.11: 34108763-34108763 on Build GRCh38
rs1017673207 Chr.11: 34108926-34108926 on Build GRCh38
rs751404505 Chr.11: 34108789-34108789 on Build GRCh38
rs942977994 Chr.11: 34108801-34108801 on Build GRCh38
rs374561697 Chr.11: 34108850-34108850 on Build GRCh38
rs1012022792 Chr.11: 34108900-34108900 on Build GRCh38
rs61735314 Chr.11: 34108810-34108810 on Build GRCh38
rs764683148 Chr.11: 34108760-34108760 on Build GRCh38
rs199505742 Chr.11: 34108847-34108847 on Build GRCh38
rs758062189 Chr.11: 34108788-34108788 on Build GRCh38
rs746018093 Chr.11: 34108815-34108815 on Build GRCh38
rs534066272 Chr.11: 34108783-34108783 on Build GRCh38
rs755759283 Chr.11: 34108894-34108894 on Build GRCh38
rs893131276 Chr.11: 34108895-34108895 on Build GRCh38
rs772453274 Chr.11: 34108818-34108818 on Build GRCh38
rs762555378 Chr.11: 34108878-34108878 on Build GRCh38
rs564131534 Chr.11: 34108869-34108869 on Build GRCh38
rs367807106 Chr.11: 34108855-34108855 on Build GRCh38
rs776533696 Chr.11: 34108793-34108793 on Build GRCh38
rs150022391 Chr.11: 34108812-34108812 on Build GRCh38
rs552744350 Chr.11: 34108787-34108787 on Build GRCh38
rs754350515 Chr.11: 34108761-34108761 on Build GRCh38

View Full Product Details