Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31085025-31085294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS53
Entrez Gene ID
339105
Gene Name
protease, serine 53
Gene Aliases
POL3S, UNQ308
UniGene
Hs.569575
Gene Chromosome Location
Chr.16: 31083424-31090035 on Build GRCh38
Transcripts
NM_001039503.2 XM_011545816.1 XM_011545817.1 XM_011545818.2 XM_011545819.1 XM_011545820.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3370407 c.982G>T p.G328* Chr.16: 31085162-31085162 on Build GRCh38
5435931 c.983G>C p.G328A Chr.16: 31085161-31085161 on Build GRCh38
7236488 c.893C>G p.S298C Chr.16: 31085251-31085251 on Build GRCh38
5791692 c.954G>A p.L318L Chr.16: 31085190-31085190 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs984219627 Chr.16: 31085058-31085058 on Build GRCh38
rs1003813371 Chr.16: 31085250-31085250 on Build GRCh38
rs773067647 Chr.16: 31085181-31085181 on Build GRCh38
rs745937671 Chr.16: 31085053-31085053 on Build GRCh38
rs865963415 Chr.16: 31085216-31085216 on Build GRCh38
rs762701898 Chr.16: 31085105-31085105 on Build GRCh38
rs908711750 Chr.16: 31085068-31085073 on Build GRCh38
rs779270659 Chr.16: 31085156-31085156 on Build GRCh38
rs765518459 Chr.16: 31085239-31085239 on Build GRCh38
rs746037110 Chr.16: 31085158-31085158 on Build GRCh38
rs779985396 Chr.16: 31085161-31085161 on Build GRCh38
rs902359614 Chr.16: 31085224-31085224 on Build GRCh38
rs762451275 Chr.16: 31085218-31085218 on Build GRCh38
rs370867880 Chr.16: 31085121-31085121 on Build GRCh38
rs766109269 Chr.16: 31085219-31085219 on Build GRCh38
rs757463088 Chr.16: 31085045-31085045 on Build GRCh38
rs375509967 Chr.16: 31085178-31085178 on Build GRCh38
rs368384747 Chr.16: 31085113-31085113 on Build GRCh38
rs375690216 Chr.16: 31085162-31085162 on Build GRCh38
rs369797104 Chr.16: 31085196-31085196 on Build GRCh38
rs768698044 Chr.16: 31085074-31085074 on Build GRCh38
rs764965262 Chr.16: 31085086-31085086 on Build GRCh38
rs751060113 Chr.16: 31085065-31085066 on Build GRCh38
rs751571345 Chr.16: 31085270-31085270 on Build GRCh38
rs72785539 Chr.16: 31085174-31085174 on Build GRCh38
rs758659965 Chr.16: 31085160-31085160 on Build GRCh38
rs776760159 Chr.16: 31085165-31085165 on Build GRCh38
rs772954131 Chr.16: 31085088-31085088 on Build GRCh38
rs761742723 Chr.16: 31085082-31085082 on Build GRCh38
rs371773654 Chr.16: 31085147-31085147 on Build GRCh38
rs199760254 Chr.16: 31085168-31085168 on Build GRCh38
rs764560393 Chr.16: 31085231-31085231 on Build GRCh38
rs758546148 Chr.16: 31085245-31085245 on Build GRCh38
rs374546739 Chr.16: 31085081-31085081 on Build GRCh38
rs1054610897 Chr.16: 31085087-31085087 on Build GRCh38
rs201947413 Chr.16: 31085068-31085068 on Build GRCh38
rs750108535 Chr.16: 31085244-31085244 on Build GRCh38
rs752725867 Chr.16: 31085175-31085180 on Build GRCh38
rs779145638 Chr.16: 31085046-31085046 on Build GRCh38
rs746863050 Chr.16: 31085071-31085071 on Build GRCh38
rs754292430 Chr.16: 31085236-31085236 on Build GRCh38
rs754169438 Chr.16: 31085143-31085143 on Build GRCh38
rs757792377 Chr.16: 31085237-31085237 on Build GRCh38
rs1044646447 Chr.16: 31085124-31085124 on Build GRCh38
rs777607150 Chr.16: 31085270-31085271 on Build GRCh38
rs188342896 Chr.16: 31085140-31085140 on Build GRCh38
rs750858241 Chr.16: 31085243-31085243 on Build GRCh38
rs771997153 Chr.16: 31085067-31085067 on Build GRCh38
rs767525494 Chr.16: 31085130-31085133 on Build GRCh38
rs754561666 Chr.16: 31085067-31085068 on Build GRCh38
rs780366001 Chr.16: 31085257-31085257 on Build GRCh38
rs376206757 Chr.16: 31085139-31085139 on Build GRCh38
rs759123353 Chr.16: 31085226-31085226 on Build GRCh38
rs370538390 Chr.16: 31085222-31085222 on Build GRCh38
rs371720280 Chr.16: 31085163-31085163 on Build GRCh38

View Full Product Details