Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 108176346-108176589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR2E1
Entrez Gene ID
7101
Gene Name
nuclear receptor subfamily 2 group E member 1
Gene Aliases
TLL, TLX, XTLL
UniGene
Hs.157688
Gene Chromosome Location
Chr.6: 108166058-108188809 on Build GRCh38
Transcripts
NM_001286102.1 NM_003269.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
40394 c.298C>T p.R100C Chr.6: 108176541-108176541 on Build GRCh38
3828743 c.260-1G>A p.? Chr.6: 108176502-108176502 on Build GRCh38
1439597 c.310G>A p.A104T Chr.6: 108176553-108176553 on Build GRCh38
6561381 c.334G>C p.E112Q Chr.6: 108176577-108176577 on Build GRCh38
353401 c.324T>C p.R108R Chr.6: 108176567-108176567 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768471900 Chr.6: 108176499-108176499 on Build GRCh38
rs367988764 Chr.6: 108176496-108176496 on Build GRCh38
rs560114573 Chr.6: 108176393-108176393 on Build GRCh38
rs552065706 Chr.6: 108176437-108176437 on Build GRCh38
rs761813335 Chr.6: 108176507-108176507 on Build GRCh38
rs766445753 Chr.6: 108176403-108176403 on Build GRCh38
rs1012549467 Chr.6: 108176406-108176406 on Build GRCh38
rs990324418 Chr.6: 108176402-108176402 on Build GRCh38
rs923314715 Chr.6: 108176494-108176494 on Build GRCh38
rs1031494244 Chr.6: 108176441-108176441 on Build GRCh38
rs773212387 Chr.6: 108176442-108176442 on Build GRCh38
rs776362366 Chr.6: 108176504-108176504 on Build GRCh38
rs759582254 Chr.6: 108176561-108176561 on Build GRCh38
rs756583761 Chr.6: 108176467-108176467 on Build GRCh38
rs907114354 Chr.6: 108176425-108176425 on Build GRCh38
rs751670468 Chr.6: 108176489-108176489 on Build GRCh38
rs1045427019 Chr.6: 108176424-108176424 on Build GRCh38
rs111602818 Chr.6: 108176531-108176531 on Build GRCh38
rs778494221 Chr.6: 108176474-108176474 on Build GRCh38
rs1042157738 Chr.6: 108176426-108176426 on Build GRCh38
rs370543731 Chr.6: 108176465-108176465 on Build GRCh38
rs367602922 Chr.6: 108176455-108176455 on Build GRCh38
rs569609413 Chr.6: 108176386-108176386 on Build GRCh38
rs769686347 Chr.6: 108176522-108176522 on Build GRCh38
rs1017661494 Chr.6: 108176463-108176463 on Build GRCh38
rs374560382 Chr.6: 108176477-108176477 on Build GRCh38
rs574484230 Chr.6: 108176369-108176369 on Build GRCh38
rs998805093 Chr.6: 108176431-108176431 on Build GRCh38
rs773186014 Chr.6: 108176546-108176546 on Build GRCh38
rs771559629 Chr.6: 108176481-108176481 on Build GRCh38
rs757650662 Chr.6: 108176473-108176473 on Build GRCh38
rs778239122 Chr.6: 108176471-108176471 on Build GRCh38
rs755756801 Chr.6: 108176484-108176485 on Build GRCh38
rs532289425 Chr.6: 108176417-108176417 on Build GRCh38
rs540289394 Chr.6: 108176385-108176385 on Build GRCh38
rs902284888 Chr.6: 108176452-108176452 on Build GRCh38
rs993974935 Chr.6: 108176495-108176495 on Build GRCh38

View Full Product Details