Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 27533802-27534061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB18
Entrez Gene ID
22931
Gene Name
RAB18, member RAS oncogene family
Gene Aliases
RAB18LI1, WARBM3
UniGene
Hs.406799
Gene Chromosome Location
Chr.10: 27504174-27542237 on Build GRCh38
Transcripts
NM_001256410.1 NM_001256411.1 NM_001256412.1 NM_001256415.1 NM_021252.4 NR_046172.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1347460 c.368A>G p.K123R Chr.10: 27533843-27533843 on Build GRCh38
1347461 c.393C>T p.V131V Chr.10: 27533942-27533942 on Build GRCh38
4966136 c.332C>T p.T111I Chr.10: 27533807-27533807 on Build GRCh38
917459 c.372C>T p.I124I Chr.10: 27533847-27533847 on Build GRCh38
917460 c.403G>T p.E135* Chr.10: 27533952-27533952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150338094 Chr.10: 27533899-27533899 on Build GRCh38
rs772739893 Chr.10: 27533916-27533916 on Build GRCh38
rs758621104 Chr.10: 27534011-27534011 on Build GRCh38
rs773648790 Chr.10: 27534003-27534003 on Build GRCh38
rs747332052 Chr.10: 27534027-27534027 on Build GRCh38
rs752748726 Chr.10: 27533848-27533848 on Build GRCh38
rs866252168 Chr.10: 27533967-27533967 on Build GRCh38
rs2242356 Chr.10: 27533887-27533887 on Build GRCh38
rs140686479 Chr.10: 27533934-27533934 on Build GRCh38
rs746244679 Chr.10: 27533898-27533898 on Build GRCh38
rs770628607 Chr.10: 27533935-27533935 on Build GRCh38
rs780435909 Chr.10: 27534024-27534024 on Build GRCh38
rs773809598 Chr.10: 27533938-27533938 on Build GRCh38
rs190213580 Chr.10: 27533943-27533943 on Build GRCh38
rs367554299 Chr.10: 27533890-27533890 on Build GRCh38
rs769173543 Chr.10: 27534029-27534029 on Build GRCh38
rs759400796 Chr.10: 27533870-27533871 on Build GRCh38
rs780449726 Chr.10: 27533903-27533903 on Build GRCh38
rs756109187 Chr.10: 27533851-27533851 on Build GRCh38
rs370486903 Chr.10: 27533886-27533886 on Build GRCh38
rs764084774 Chr.10: 27533945-27533945 on Build GRCh38
rs778051423 Chr.10: 27533860-27533860 on Build GRCh38
rs750666780 Chr.10: 27533985-27533985 on Build GRCh38
rs753997206 Chr.10: 27533951-27533951 on Build GRCh38
rs371577624 Chr.10: 27533912-27533912 on Build GRCh38
rs757213835 Chr.10: 27533955-27533955 on Build GRCh38
rs765168595 Chr.10: 27533921-27533922 on Build GRCh38
rs779043957 Chr.10: 27533960-27533960 on Build GRCh38
rs769837798 Chr.10: 27533942-27533942 on Build GRCh38
rs1032076203 Chr.10: 27533999-27533999 on Build GRCh38
rs781620585 Chr.10: 27534032-27534032 on Build GRCh38
rs759222021 Chr.10: 27533940-27533940 on Build GRCh38
rs753942502 Chr.10: 27533880-27533880 on Build GRCh38

View Full Product Details