Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 133073153-133073426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA4
Entrez Gene ID
3308
Gene Name
heat shock protein family A (Hsp70) member 4
Gene Aliases
APG-2, HEL-S-5a, HS24/P52, HSPH2, RY, hsp70, hsp70RY
UniGene
Hs.90093
Gene Chromosome Location
Chr.5: 133051970-133105017 on Build GRCh38
Transcripts
NM_002154.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4550688 c.506G>A p.R169Q Chr.5: 133073306-133073306 on Build GRCh38
4621181 c.449A>G p.D150G Chr.5: 133073249-133073249 on Build GRCh38
7371573 c.513G>T p.M171I Chr.5: 133073313-133073313 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754831914 Chr.5: 133073306-133073306 on Build GRCh38
rs757905051 Chr.5: 133073236-133073236 on Build GRCh38
rs756561855 Chr.5: 133073213-133073214 on Build GRCh38
rs149159356 Chr.5: 133073261-133073261 on Build GRCh38
rs752406094 Chr.5: 133073319-133073319 on Build GRCh38
rs775886603 Chr.5: 133073196-133073196 on Build GRCh38
rs749578528 Chr.5: 133073343-133073343 on Build GRCh38
rs766264363 Chr.5: 133073210-133073210 on Build GRCh38
rs921932873 Chr.5: 133073324-133073324 on Build GRCh38
rs767719973 Chr.5: 133073269-133073269 on Build GRCh38
rs761918529 Chr.5: 133073266-133073266 on Build GRCh38
rs748367872 Chr.5: 133073369-133073369 on Build GRCh38
rs56121115 Chr.5: 133073382-133073382 on Build GRCh38
rs926140221 Chr.5: 133073211-133073214 on Build GRCh38
rs769071727 Chr.5: 133073348-133073348 on Build GRCh38
rs73788295 Chr.5: 133073341-133073341 on Build GRCh38
rs750391463 Chr.5: 133073207-133073207 on Build GRCh38
rs1012164520 Chr.5: 133073386-133073386 on Build GRCh38
rs780411260 Chr.5: 133073371-133073372 on Build GRCh38
rs570264755 Chr.5: 133073359-133073359 on Build GRCh38
rs766468777 Chr.5: 133073277-133073277 on Build GRCh38
rs147934883 Chr.5: 133073374-133073374 on Build GRCh38
rs780635947 Chr.5: 133073342-133073342 on Build GRCh38
rs111333016 Chr.5: 133073226-133073227 on Build GRCh38
rs930751154 Chr.5: 133073337-133073337 on Build GRCh38
rs750714733 Chr.5: 133073350-133073350 on Build GRCh38
rs747570011 Chr.5: 133073226-133073226 on Build GRCh38
rs763085615 Chr.5: 133073198-133073198 on Build GRCh38
rs368418974 Chr.5: 133073281-133073281 on Build GRCh38
rs375412369 Chr.5: 133073225-133073225 on Build GRCh38
rs746357245 Chr.5: 133073252-133073252 on Build GRCh38
rs139240822 Chr.5: 133073331-133073331 on Build GRCh38
rs372766936 Chr.5: 133073213-133073213 on Build GRCh38
rs753706194 Chr.5: 133073212-133073212 on Build GRCh38
rs1053482821 Chr.5: 133073240-133073240 on Build GRCh38
rs762005049 Chr.5: 133073214-133073215 on Build GRCh38
rs753525823 Chr.5: 133073305-133073305 on Build GRCh38
rs762808876 Chr.5: 133073214-133073214 on Build GRCh38
rs944730435 Chr.5: 133073262-133073262 on Build GRCh38
rs145291096 Chr.5: 133073253-133073253 on Build GRCh38
rs761976870 Chr.5: 133073205-133073205 on Build GRCh38
rs764726915 Chr.5: 133073311-133073311 on Build GRCh38
rs767523769 Chr.5: 133073206-133073206 on Build GRCh38
rs61755724 Chr.5: 133073275-133073275 on Build GRCh38
rs756407492 Chr.5: 133073349-133073350 on Build GRCh38
rs770153298 Chr.5: 133073188-133073188 on Build GRCh38
rs149245317 Chr.5: 133073265-133073265 on Build GRCh38
rs773402313 Chr.5: 133073380-133073380 on Build GRCh38
rs774275528 Chr.5: 133073204-133073204 on Build GRCh38
rs767715428 Chr.5: 133073214-133073216 on Build GRCh38
rs185385178 Chr.5: 133073328-133073328 on Build GRCh38
rs150908671 Chr.5: 133073237-133073237 on Build GRCh38
rs764420290 Chr.5: 133073199-133073199 on Build GRCh38
rs7720224 Chr.5: 133073389-133073389 on Build GRCh38
rs755916156 Chr.5: 133073208-133073208 on Build GRCh38

View Full Product Details