Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112978558-112978829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OAS2
Entrez Gene ID
4939
Gene Name
2'-5'-oligoadenylate synthetase 2
Gene Aliases
-
UniGene
Hs.414332
Gene Chromosome Location
Chr.12: 112978395-113011723 on Build GRCh38
Transcripts
NM_001032731.1 NM_002535.2 NM_016817.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5987665 c.85G>A p.E29K Chr.12: 112978693-112978693 on Build GRCh38
5801520 c.152_160delTCCCCCTGG p.F51_V54>L Chr.12: 112978760-112978768 on Build GRCh38
1949002 c.83A>G p.E28G Chr.12: 112978691-112978691 on Build GRCh38
5446999 c.20A>G p.Q7R Chr.12: 112978628-112978628 on Build GRCh38
1628391 c.85G>T p.E29* Chr.12: 112978693-112978693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs45562438 Chr.12: 112978626-112978626 on Build GRCh38
rs751420328 Chr.12: 112978593-112978593 on Build GRCh38
rs754769030 Chr.12: 112978600-112978600 on Build GRCh38
rs752603720 Chr.12: 112978613-112978613 on Build GRCh38
rs771070877 Chr.12: 112978710-112978710 on Build GRCh38
rs56249095 Chr.12: 112978758-112978758 on Build GRCh38
rs771017675 Chr.12: 112978767-112978767 on Build GRCh38
rs550274290 Chr.12: 112978750-112978750 on Build GRCh38
rs182245386 Chr.12: 112978804-112978804 on Build GRCh38
rs561373886 Chr.12: 112978655-112978655 on Build GRCh38
rs146990827 Chr.12: 112978645-112978645 on Build GRCh38
rs763154036 Chr.12: 112978706-112978706 on Build GRCh38
rs528927805 Chr.12: 112978725-112978725 on Build GRCh38
rs774520551 Chr.12: 112978775-112978775 on Build GRCh38
rs759799053 Chr.12: 112978792-112978792 on Build GRCh38
rs368128261 Chr.12: 112978662-112978662 on Build GRCh38
rs778650316 Chr.12: 112978738-112978738 on Build GRCh38
rs766146526 Chr.12: 112978591-112978591 on Build GRCh38
rs780407986 Chr.12: 112978773-112978773 on Build GRCh38
rs752651612 Chr.12: 112978719-112978719 on Build GRCh38
rs777191278 Chr.12: 112978625-112978625 on Build GRCh38
rs1010031248 Chr.12: 112978799-112978799 on Build GRCh38
rs767250727 Chr.12: 112978718-112978718 on Build GRCh38
rs899078633 Chr.12: 112978732-112978732 on Build GRCh38
rs202125427 Chr.12: 112978760-112978760 on Build GRCh38
rs767380192 Chr.12: 112978603-112978603 on Build GRCh38
rs780563212 Chr.12: 112978631-112978631 on Build GRCh38
rs747311190 Chr.12: 112978638-112978638 on Build GRCh38
rs774565552 Chr.12: 112978711-112978711 on Build GRCh38
rs756832935 Chr.12: 112978731-112978731 on Build GRCh38
rs771881227 Chr.12: 112978793-112978793 on Build GRCh38
rs1001446314 Chr.12: 112978733-112978733 on Build GRCh38
rs755361976 Chr.12: 112978743-112978743 on Build GRCh38
rs373240531 Chr.12: 112978720-112978720 on Build GRCh38
rs200545644 Chr.12: 112978745-112978745 on Build GRCh38
rs927394535 Chr.12: 112978610-112978610 on Build GRCh38
rs769892744 Chr.12: 112978687-112978687 on Build GRCh38
rs749303898 Chr.12: 112978763-112978763 on Build GRCh38
rs1051869709 Chr.12: 112978683-112978683 on Build GRCh38
rs545739428 Chr.12: 112978590-112978590 on Build GRCh38
rs1037026002 Chr.12: 112978672-112978679 on Build GRCh38
rs763647879 Chr.12: 112978721-112978721 on Build GRCh38
rs1048080768 Chr.12: 112978727-112978727 on Build GRCh38
rs758741088 Chr.12: 112978608-112978614 on Build GRCh38
rs940836009 Chr.12: 112978649-112978649 on Build GRCh38
rs148059189 Chr.12: 112978716-112978716 on Build GRCh38
rs781694968 Chr.12: 112978658-112978658 on Build GRCh38
rs748641208 Chr.12: 112978679-112978679 on Build GRCh38
rs773220214 Chr.12: 112978693-112978693 on Build GRCh38
rs147801521 Chr.12: 112978623-112978623 on Build GRCh38

View Full Product Details