Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75225363-75225602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGA3
Entrez Gene ID
23163
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 3
Gene Aliases
-
UniGene
Hs.87726
Gene Chromosome Location
Chr.17: 75225464-75262393 on Build GRCh38
Transcripts
XM_011524563.2 NM_001303276.1 NM_024844.4 XM_005257690.2 XM_005257692.3 XM_006722094.1 XM_011525267.1 XM_011525268.1 XM_017025108.1 XM_017025109.1 XM_017025110.1

Gene Symbol
NUP85
Entrez Gene ID
79902
Gene Name
nucleoporin 85
Gene Aliases
FROUNT, Nup75
UniGene
Hs.362817
Gene Chromosome Location
Chr.17: 75205450-75235759 on Build GRCh38
Transcripts
XM_011524563.2 NM_001303276.1 NM_024844.4 XM_005257690.2 XM_005257692.3 XM_006722094.1 XM_011525267.1 XM_011525268.1 XM_017025108.1 XM_017025109.1 XM_017025110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7411012 c.276C>G p.H92Q Chr.17: 75225389-75225389 on Build GRCh38
7316283 c.818C>T p.T273I Chr.17: 75225427-75225427 on Build GRCh38
4406788 c.794G>A p.C265Y Chr.17: 75225403-75225403 on Build GRCh38
7300097 c.754_755delAC p.T252fs*14 Chr.17: 75225363-75225364 on Build GRCh38
4623663 c.300C>T p.Y100Y Chr.17: 75225413-75225413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014121599 Chr.17: 75225561-75225561 on Build GRCh38
rs773708577 Chr.17: 75225419-75225419 on Build GRCh38
rs774652697 Chr.17: 75225431-75225431 on Build GRCh38
rs200495546 Chr.17: 75225395-75225395 on Build GRCh38
rs146236628 Chr.17: 75225461-75225461 on Build GRCh38
rs760285954 Chr.17: 75225460-75225460 on Build GRCh38
rs61760968 Chr.17: 75225432-75225432 on Build GRCh38
rs1034447731 Chr.17: 75225547-75225547 on Build GRCh38
rs775254781 Chr.17: 75225459-75225459 on Build GRCh38
rs138086614 Chr.17: 75225569-75225569 on Build GRCh38
rs1005611266 Chr.17: 75225534-75225534 on Build GRCh38
rs866622891 Chr.17: 75225393-75225393 on Build GRCh38
rs781103310 Chr.17: 75225515-75225515 on Build GRCh38
rs562632765 Chr.17: 75225557-75225557 on Build GRCh38
rs374408998 Chr.17: 75225582-75225582 on Build GRCh38
rs1047027011 Chr.17: 75225505-75225505 on Build GRCh38
rs925684897 Chr.17: 75225388-75225388 on Build GRCh38
rs571750459 Chr.17: 75225579-75225579 on Build GRCh38
rs753583486 Chr.17: 75225526-75225526 on Build GRCh38
rs1041267140 Chr.17: 75225444-75225444 on Build GRCh38
rs1026682451 Chr.17: 75225567-75225568 on Build GRCh38
rs1050134772 Chr.17: 75225396-75225396 on Build GRCh38
rs901473821 Chr.17: 75225481-75225481 on Build GRCh38
rs756983192 Chr.17: 75225486-75225486 on Build GRCh38
rs138444280 Chr.17: 75225425-75225425 on Build GRCh38
rs747560348 Chr.17: 75225491-75225491 on Build GRCh38
rs531797747 Chr.17: 75225562-75225562 on Build GRCh38
rs967354498 Chr.17: 75225571-75225571 on Build GRCh38
rs559887603 Chr.17: 75225504-75225504 on Build GRCh38
rs766886650 Chr.17: 75225433-75225433 on Build GRCh38
rs889989024 Chr.17: 75225446-75225446 on Build GRCh38
rs770324180 Chr.17: 75225411-75225411 on Build GRCh38
rs367852869 Chr.17: 75225512-75225512 on Build GRCh38
rs371760418 Chr.17: 75225482-75225482 on Build GRCh38
rs201624012 Chr.17: 75225492-75225492 on Build GRCh38
rs563377292 Chr.17: 75225409-75225409 on Build GRCh38
rs763940285 Chr.17: 75225473-75225473 on Build GRCh38
rs368833946 Chr.17: 75225428-75225428 on Build GRCh38

View Full Product Details