Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81940888-81941145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYADML2
Entrez Gene ID
255275
Gene Name
myeloid associated differentiation marker like 2
Gene Aliases
-
UniGene
Hs.729645
Gene Chromosome Location
Chr.17: 81939645-81947233 on Build GRCh38
Transcripts
NM_001145113.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5833471 c.795_796insC p.N266fs*29 Chr.17: 81940946-81940947 on Build GRCh38
985990 c.853T>C p.Y285H Chr.17: 81940889-81940889 on Build GRCh38
5495553 c.835G>A p.V279M Chr.17: 81940907-81940907 on Build GRCh38
5585841 c.793C>T p.P265S Chr.17: 81940949-81940949 on Build GRCh38
3388310 c.631G>A p.V211M Chr.17: 81941111-81941111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021530171 Chr.17: 81940943-81940943 on Build GRCh38
rs748912551 Chr.17: 81941015-81941015 on Build GRCh38
rs868737913 Chr.17: 81940920-81940921 on Build GRCh38
rs760736785 Chr.17: 81941050-81941050 on Build GRCh38
rs748414196 Chr.17: 81940968-81940968 on Build GRCh38
rs773013862 Chr.17: 81941049-81941049 on Build GRCh38
rs145619726 Chr.17: 81940954-81940954 on Build GRCh38
rs758676686 Chr.17: 81941112-81941112 on Build GRCh38
rs1009735862 Chr.17: 81940929-81940929 on Build GRCh38
rs774177998 Chr.17: 81940926-81940926 on Build GRCh38
rs563167581 Chr.17: 81941029-81941029 on Build GRCh38
rs189201910 Chr.17: 81941103-81941103 on Build GRCh38
rs767360602 Chr.17: 81941047-81941047 on Build GRCh38
rs916981709 Chr.17: 81940986-81940986 on Build GRCh38
rs770248037 Chr.17: 81940985-81940986 on Build GRCh38
rs557230569 Chr.17: 81940907-81940907 on Build GRCh38
rs140886468 Chr.17: 81940936-81940936 on Build GRCh38
rs373462435 Chr.17: 81941088-81941088 on Build GRCh38
rs761451771 Chr.17: 81941044-81941044 on Build GRCh38
rs572847285 Chr.17: 81940955-81940955 on Build GRCh38
rs775017420 Chr.17: 81941010-81941010 on Build GRCh38
rs372698458 Chr.17: 81940924-81940924 on Build GRCh38
rs755292173 Chr.17: 81940947-81940947 on Build GRCh38
rs751976251 Chr.17: 81941126-81941126 on Build GRCh38
rs766537154 Chr.17: 81941056-81941056 on Build GRCh38
rs773431171 Chr.17: 81941098-81941104 on Build GRCh38
rs377112990 Chr.17: 81940944-81940944 on Build GRCh38
rs555491751 Chr.17: 81941046-81941046 on Build GRCh38
rs532700062 Chr.17: 81941003-81941003 on Build GRCh38
rs774020399 Chr.17: 81941038-81941038 on Build GRCh38
rs748338882 Chr.17: 81940961-81940961 on Build GRCh38
rs201601082 Chr.17: 81940913-81940913 on Build GRCh38
rs187029546 Chr.17: 81940979-81940979 on Build GRCh38
rs150154051 Chr.17: 81940945-81940945 on Build GRCh38
rs767320390 Chr.17: 81940937-81940937 on Build GRCh38
rs1053495041 Chr.17: 81940928-81940928 on Build GRCh38
rs375074526 Chr.17: 81940980-81940980 on Build GRCh38
rs778267212 Chr.17: 81941114-81941114 on Build GRCh38
rs564528690 Chr.17: 81940998-81940998 on Build GRCh38
rs544481628 Chr.17: 81941009-81941009 on Build GRCh38
rs752976354 Chr.17: 81941111-81941111 on Build GRCh38
rs768245360 Chr.17: 81941024-81941024 on Build GRCh38
rs758792872 Chr.17: 81940975-81940975 on Build GRCh38
rs984103495 Chr.17: 81940963-81940963 on Build GRCh38
rs62077244 Chr.17: 81941057-81941057 on Build GRCh38

View Full Product Details