Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 25428689-25428962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RARB
Entrez Gene ID
5915
Gene Name
retinoic acid receptor beta
Gene Aliases
HAP, MCOPS12, NR1B2, RARbeta1, RRB2
UniGene
Hs.543218
Gene Chromosome Location
Chr.3: 24829323-25597932 on Build GRCh38
Transcripts
NM_000965.4 NM_001290216.2 NM_001290266.1 NM_001290276.1 NM_001290277.1 NM_001290300.1 NM_016152.3 NR_110892.1 NR_110893.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
446309 c.129C>T p.T43T Chr.3: 25428860-25428860 on Build GRCh38
3365147 c.113G>A p.S38N Chr.3: 25428844-25428844 on Build GRCh38
6762961 c.69G>A p.P23P Chr.3: 25428800-25428800 on Build GRCh38
1422181 c.62C>T p.A21V Chr.3: 25428793-25428793 on Build GRCh38
6256648 c.90G>T p.E30D Chr.3: 25428821-25428821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751811703 Chr.3: 25428811-25428811 on Build GRCh38
rs201424655 Chr.3: 25428777-25428777 on Build GRCh38
rs1002995251 Chr.3: 25428753-25428753 on Build GRCh38
rs778514781 Chr.3: 25428742-25428742 on Build GRCh38
rs147798097 Chr.3: 25428826-25428826 on Build GRCh38
rs115942154 Chr.3: 25428771-25428771 on Build GRCh38
rs773868160 Chr.3: 25428931-25428931 on Build GRCh38
rs77740700 Chr.3: 25428920-25428920 on Build GRCh38
rs943782896 Chr.3: 25428912-25428912 on Build GRCh38
rs748551833 Chr.3: 25428924-25428924 on Build GRCh38
rs762241586 Chr.3: 25428793-25428793 on Build GRCh38
rs768354497 Chr.3: 25428780-25428780 on Build GRCh38
rs749077401 Chr.3: 25428723-25428723 on Build GRCh38
rs765882393 Chr.3: 25428794-25428794 on Build GRCh38
rs577970077 Chr.3: 25428836-25428836 on Build GRCh38
rs977039906 Chr.3: 25428842-25428842 on Build GRCh38
rs753673931 Chr.3: 25428849-25428849 on Build GRCh38
rs923393778 Chr.3: 25428918-25428918 on Build GRCh38
rs771435459 Chr.3: 25428758-25428758 on Build GRCh38
rs768238601 Chr.3: 25428915-25428915 on Build GRCh38
rs116259656 Chr.3: 25428916-25428916 on Build GRCh38
rs778791519 Chr.3: 25428744-25428744 on Build GRCh38
rs761745119 Chr.3: 25428929-25428929 on Build GRCh38
rs747714002 Chr.3: 25428921-25428921 on Build GRCh38
rs781662331 Chr.3: 25428907-25428909 on Build GRCh38
rs777364835 Chr.3: 25428718-25428718 on Build GRCh38
rs745750943 Chr.3: 25428866-25428866 on Build GRCh38
rs373305056 Chr.3: 25428789-25428789 on Build GRCh38
rs746518650 Chr.3: 25428911-25428911 on Build GRCh38
rs764613965 Chr.3: 25428803-25428803 on Build GRCh38
rs766309157 Chr.3: 25428807-25428807 on Build GRCh38
rs962337521 Chr.3: 25428846-25428846 on Build GRCh38
rs370403223 Chr.3: 25428788-25428788 on Build GRCh38
rs990515725 Chr.3: 25428835-25428835 on Build GRCh38
rs745416080 Chr.3: 25428749-25428749 on Build GRCh38
rs1021557007 Chr.3: 25428815-25428815 on Build GRCh38
rs201745230 Chr.3: 25428860-25428860 on Build GRCh38
rs768498439 Chr.3: 25428887-25428887 on Build GRCh38
rs909529483 Chr.3: 25428839-25428839 on Build GRCh38
rs779502214 Chr.3: 25428907-25428907 on Build GRCh38
rs755442951 Chr.3: 25428898-25428898 on Build GRCh38
rs771732428 Chr.3: 25428800-25428800 on Build GRCh38
rs538896356 Chr.3: 25428920-25428921 on Build GRCh38
rs77824017 Chr.3: 25428921-25428921 on Build GRCh38

View Full Product Details