Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111543850-111544111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAYN
Entrez Gene ID
143903
Gene Name
layilin
Gene Aliases
-
UniGene
Hs.503831
Gene Chromosome Location
Chr.11: 111540280-111563464 on Build GRCh38
Transcripts
NM_001258390.1 NM_001258391.1 NM_001318799.1 NM_178834.4 XM_006718769.3 XM_006718770.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6007785 c.203G>A p.G68D Chr.11: 111544040-111544040 on Build GRCh38
4017771 c.277T>C p.F93L Chr.11: 111544090-111544090 on Build GRCh38
1297640 c.235C>A p.Q79K Chr.11: 111544072-111544072 on Build GRCh38
3868406 c.104G>A p.G35E Chr.11: 111543941-111543941 on Build GRCh38
5360474 c.163T>G p.F55V Chr.11: 111543976-111543976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766557678 Chr.11: 111544006-111544006 on Build GRCh38
rs759541337 Chr.11: 111544028-111544028 on Build GRCh38
rs200578625 Chr.11: 111544001-111544001 on Build GRCh38
rs574688254 Chr.11: 111543944-111543944 on Build GRCh38
rs754903225 Chr.11: 111544032-111544032 on Build GRCh38
rs140344376 Chr.11: 111544044-111544044 on Build GRCh38
rs778151629 Chr.11: 111544066-111544066 on Build GRCh38
rs781274219 Chr.11: 111543986-111543986 on Build GRCh38
rs201747451 Chr.11: 111543902-111543902 on Build GRCh38
rs769505552 Chr.11: 111543996-111543996 on Build GRCh38
rs773532897 Chr.11: 111543998-111543998 on Build GRCh38
rs778489334 Chr.11: 111543953-111543953 on Build GRCh38
rs992891369 Chr.11: 111544054-111544054 on Build GRCh38
rs755569988 Chr.11: 111543943-111543943 on Build GRCh38
rs779588947 Chr.11: 111543971-111543971 on Build GRCh38
rs772732084 Chr.11: 111544034-111544034 on Build GRCh38
rs753723537 Chr.11: 111544016-111544016 on Build GRCh38
rs749415260 Chr.11: 111544012-111544013 on Build GRCh38
rs750693804 Chr.11: 111543892-111543892 on Build GRCh38
rs141049653 Chr.11: 111544069-111544069 on Build GRCh38
rs777974609 Chr.11: 111543949-111543950 on Build GRCh38
rs866895364 Chr.11: 111543989-111543989 on Build GRCh38
rs919463140 Chr.11: 111544068-111544068 on Build GRCh38
rs766602564 Chr.11: 111543910-111543910 on Build GRCh38
rs765440796 Chr.11: 111543875-111543875 on Build GRCh38
rs971768137 Chr.11: 111544047-111544047 on Build GRCh38
rs1000154255 Chr.11: 111543899-111543899 on Build GRCh38
rs756451753 Chr.11: 111543949-111543949 on Build GRCh38
rs563598463 Chr.11: 111544040-111544040 on Build GRCh38
rs200355216 Chr.11: 111543937-111543937 on Build GRCh38
rs368995215 Chr.11: 111543991-111543991 on Build GRCh38
rs377682012 Chr.11: 111543874-111543874 on Build GRCh38
rs745552777 Chr.11: 111543954-111543954 on Build GRCh38
rs753041812 Chr.11: 111543946-111543946 on Build GRCh38
rs11827718 Chr.11: 111544009-111544009 on Build GRCh38
rs149755521 Chr.11: 111543938-111543938 on Build GRCh38
rs746915929 Chr.11: 111543984-111543984 on Build GRCh38
rs756463419 Chr.11: 111544057-111544057 on Build GRCh38

View Full Product Details