Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154478713-154478962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAGE3
Entrez Gene ID
8270
Gene Name
L antigen family member 3
Gene Aliases
CVG5, DXS9879E, DXS9951E, ESO3, ITBA2, Pcc1
UniGene
Hs.444619
Gene Chromosome Location
Chr.X: 154477769-154479257 on Build GRCh38
Transcripts
NM_006014.4

SNP Detail

SNP ID Chromosome Location
rs782601504 Chr.X: 154478804-154478804 on Build GRCh38
rs781965466 Chr.X: 154478754-154478754 on Build GRCh38
rs984350197 Chr.X: 154478910-154478910 on Build GRCh38
rs782016401 Chr.X: 154478815-154478815 on Build GRCh38
rs2276463 Chr.X: 154478825-154478825 on Build GRCh38
rs782088780 Chr.X: 154478839-154478839 on Build GRCh38
rs942873497 Chr.X: 154478928-154478928 on Build GRCh38
rs782019621 Chr.X: 154478887-154478887 on Build GRCh38
rs782053419 Chr.X: 154478779-154478779 on Build GRCh38
rs370078755 Chr.X: 154478792-154478792 on Build GRCh38
rs1012023429 Chr.X: 154478883-154478883 on Build GRCh38
rs782309060 Chr.X: 154478821-154478821 on Build GRCh38
rs74922846 Chr.X: 154478870-154478870 on Build GRCh38
rs1001156590 Chr.X: 154478865-154478865 on Build GRCh38
rs374765412 Chr.X: 154478794-154478794 on Build GRCh38
rs368054910 Chr.X: 154478796-154478796 on Build GRCh38
rs782717564 Chr.X: 154478780-154478780 on Build GRCh38
rs1062915 Chr.X: 154478760-154478760 on Build GRCh38
rs1041213718 Chr.X: 154478929-154478929 on Build GRCh38
rs1038771485 Chr.X: 154478826-154478826 on Build GRCh38
rs917931041 Chr.X: 154478739-154478739 on Build GRCh38
rs782809328 Chr.X: 154478774-154478774 on Build GRCh38
rs781818854 Chr.X: 154478889-154478889 on Build GRCh38
rs1025038377 Chr.X: 154478884-154478884 on Build GRCh38
rs782494587 Chr.X: 154478790-154478790 on Build GRCh38
rs781861801 Chr.X: 154478856-154478856 on Build GRCh38
rs781904042 Chr.X: 154478777-154478777 on Build GRCh38
rs782788929 Chr.X: 154478866-154478866 on Build GRCh38
rs782163014 Chr.X: 154478819-154478819 on Build GRCh38
rs782172701 Chr.X: 154478801-154478801 on Build GRCh38
rs997212905 Chr.X: 154478855-154478855 on Build GRCh38
rs1042444602 Chr.X: 154478860-154478860 on Build GRCh38
rs782323206 Chr.X: 154478802-154478802 on Build GRCh38
rs782752201 Chr.X: 154478845-154478845 on Build GRCh38
rs782065337 Chr.X: 154478747-154478747 on Build GRCh38
rs782060612 Chr.X: 154478908-154478908 on Build GRCh38
rs782021588 Chr.X: 154478946-154478946 on Build GRCh38
rs782634958 Chr.X: 154478925-154478925 on Build GRCh38
rs911405244 Chr.X: 154478927-154478927 on Build GRCh38
rs11538409 Chr.X: 154478737-154478737 on Build GRCh38
rs889919994 Chr.X: 154478817-154478817 on Build GRCh38
rs11538408 Chr.X: 154478846-154478846 on Build GRCh38
rs782228821 Chr.X: 154478807-154478807 on Build GRCh38
rs781952793 Chr.X: 154478943-154478943 on Build GRCh38
rs782685188 Chr.X: 154478797-154478797 on Build GRCh38
rs781999457 Chr.X: 154478782-154478782 on Build GRCh38
rs782729135 Chr.X: 154478751-154478751 on Build GRCh38

View Full Product Details