Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176883720-176883991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HK3
Entrez Gene ID
3101
Gene Name
hexokinase 3
Gene Aliases
HKIII, HXK3
UniGene
Hs.411695
Gene Chromosome Location
Chr.5: 176880869-176899373 on Build GRCh38
Transcripts
NM_002115.2 XM_017009411.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6680608 c.2025C>T p.D675D Chr.5: 176883798-176883798 on Build GRCh38
3853914 c.1992G>A p.T664T Chr.5: 176883831-176883831 on Build GRCh38
3726475 c.2030G>A p.R677H Chr.5: 176883793-176883793 on Build GRCh38
3729327 c.1961A>G p.E654G Chr.5: 176883862-176883862 on Build GRCh38
4392141 c.2034C>T p.C678C Chr.5: 176883789-176883789 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764452797 Chr.5: 176883771-176883771 on Build GRCh38
rs199684264 Chr.5: 176883797-176883797 on Build GRCh38
rs749924458 Chr.5: 176883776-176883776 on Build GRCh38
rs200553299 Chr.5: 176883760-176883760 on Build GRCh38
rs760956658 Chr.5: 176883770-176883770 on Build GRCh38
rs770077768 Chr.5: 176883882-176883882 on Build GRCh38
rs140261586 Chr.5: 176883789-176883789 on Build GRCh38
rs779271345 Chr.5: 176883796-176883796 on Build GRCh38
rs1024564206 Chr.5: 176883759-176883759 on Build GRCh38
rs776048582 Chr.5: 176883761-176883761 on Build GRCh38
rs375101205 Chr.5: 176883742-176883742 on Build GRCh38
rs372237392 Chr.5: 176883907-176883907 on Build GRCh38
rs201414393 Chr.5: 176883892-176883892 on Build GRCh38
rs757795706 Chr.5: 176883794-176883794 on Build GRCh38
rs373320988 Chr.5: 176883912-176883912 on Build GRCh38
rs755973686 Chr.5: 176883911-176883911 on Build GRCh38
rs750004073 Chr.5: 176883780-176883780 on Build GRCh38
rs377712743 Chr.5: 176883913-176883913 on Build GRCh38
rs761477115 Chr.5: 176883777-176883777 on Build GRCh38
rs771160414 Chr.5: 176883745-176883745 on Build GRCh38
rs891588657 Chr.5: 176883812-176883812 on Build GRCh38
rs200692931 Chr.5: 176883908-176883908 on Build GRCh38
rs754765379 Chr.5: 176883792-176883792 on Build GRCh38
rs751478472 Chr.5: 176883829-176883829 on Build GRCh38
rs748527973 Chr.5: 176883872-176883872 on Build GRCh38
rs553544325 Chr.5: 176883949-176883949 on Build GRCh38
rs771143869 Chr.5: 176883807-176883807 on Build GRCh38
rs775865208 Chr.5: 176883799-176883799 on Build GRCh38
rs149452459 Chr.5: 176883909-176883909 on Build GRCh38
rs774624506 Chr.5: 176883750-176883750 on Build GRCh38
rs772794959 Chr.5: 176883816-176883816 on Build GRCh38
rs759654848 Chr.5: 176883753-176883753 on Build GRCh38
rs769243803 Chr.5: 176883868-176883868 on Build GRCh38
rs761278572 Chr.5: 176883740-176883741 on Build GRCh38
rs148264728 Chr.5: 176883963-176883963 on Build GRCh38
rs757892659 Chr.5: 176883788-176883788 on Build GRCh38
rs190254695 Chr.5: 176883901-176883901 on Build GRCh38
rs145354682 Chr.5: 176883838-176883838 on Build GRCh38
rs780995517 Chr.5: 176883793-176883793 on Build GRCh38
rs966823819 Chr.5: 176883940-176883940 on Build GRCh38
rs550062516 Chr.5: 176883904-176883904 on Build GRCh38
rs764928358 Chr.5: 176883810-176883810 on Build GRCh38
rs1015745893 Chr.5: 176883786-176883786 on Build GRCh38
rs772217261 Chr.5: 176883798-176883798 on Build GRCh38
rs776992861 Chr.5: 176883869-176883869 on Build GRCh38
rs559889465 Chr.5: 176883804-176883804 on Build GRCh38
rs754994587 Chr.5: 176883832-176883832 on Build GRCh38
rs753685728 Chr.5: 176883790-176883790 on Build GRCh38
rs200657973 Chr.5: 176883805-176883805 on Build GRCh38
rs755957085 Chr.5: 176883844-176883844 on Build GRCh38
rs143896985 Chr.5: 176883960-176883960 on Build GRCh38
rs114213052 Chr.5: 176883801-176883801 on Build GRCh38
rs138002696 Chr.5: 176883848-176883848 on Build GRCh38
rs772882998 Chr.5: 176883895-176883895 on Build GRCh38
rs554140413 Chr.5: 176883831-176883831 on Build GRCh38
rs905561306 Chr.5: 176883841-176883841 on Build GRCh38
rs758109758 Chr.5: 176883813-176883813 on Build GRCh38
rs144556523 Chr.5: 176883794-176883794 on Build GRCh38
rs773955084 Chr.5: 176883906-176883906 on Build GRCh38
rs762455016 Chr.5: 176883820-176883820 on Build GRCh38
rs753464254 Chr.5: 176883773-176883773 on Build GRCh38
rs747036529 Chr.5: 176883861-176883861 on Build GRCh38
rs746016190 Chr.5: 176883850-176883850 on Build GRCh38
rs1001144198 Chr.5: 176883854-176883854 on Build GRCh38
rs995593737 Chr.5: 176883956-176883956 on Build GRCh38
rs762162785 Chr.5: 176883806-176883806 on Build GRCh38
rs766926957 Chr.5: 176883752-176883752 on Build GRCh38
rs1035440256 Chr.5: 176883875-176883875 on Build GRCh38
rs765839316 Chr.5: 176883827-176883827 on Build GRCh38
rs780132854 Chr.5: 176883852-176883852 on Build GRCh38
rs752602164 Chr.5: 176883843-176883843 on Build GRCh38

View Full Product Details