Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 51432087-51432356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFATC2
Entrez Gene ID
4773
Gene Name
nuclear factor of activated T-cells 2
Gene Aliases
NFAT1, NFATP
UniGene
Hs.744148
Gene Chromosome Location
Chr.20: 51386957-51562857 on Build GRCh38
Transcripts
NM_001136021.2 NM_001258292.1 NM_001258294.1 NM_001258295.1 NM_001258296.1 NM_001258297.1 NM_012340.4 NM_173091.3 XM_011528824.2 XM_011528825.2 XM_011528826.2 XM_017027850.1 XM_017027851.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6725270 c.2606C>A p.P869H Chr.20: 51432123-51432123 on Build GRCh38
6725302 c.2564T>G p.L855R Chr.20: 51432225-51432225 on Build GRCh38
6160581 c.2672G>T p.G891V Chr.20: 51432117-51432117 on Build GRCh38
5657829 c.2630A>G p.N877S Chr.20: 51432159-51432159 on Build GRCh38
4878953 c.2677A>G p.T893A Chr.20: 51432112-51432112 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139027658 Chr.20: 51432293-51432293 on Build GRCh38
rs774423532 Chr.20: 51432240-51432240 on Build GRCh38
rs199948885 Chr.20: 51432278-51432278 on Build GRCh38
rs376673351 Chr.20: 51432252-51432252 on Build GRCh38
rs753841433 Chr.20: 51432153-51432153 on Build GRCh38
rs56050792 Chr.20: 51432180-51432180 on Build GRCh38
rs774316511 Chr.20: 51432143-51432143 on Build GRCh38
rs772115227 Chr.20: 51432301-51432301 on Build GRCh38
rs145231526 Chr.20: 51432246-51432246 on Build GRCh38
rs75923936 Chr.20: 51432158-51432158 on Build GRCh38
rs775570233 Chr.20: 51432147-51432148 on Build GRCh38
rs776529996 Chr.20: 51432247-51432247 on Build GRCh38
rs765940316 Chr.20: 51432273-51432273 on Build GRCh38
rs374799660 Chr.20: 51432179-51432179 on Build GRCh38
rs751031284 Chr.20: 51432169-51432169 on Build GRCh38
rs546062557 Chr.20: 51432245-51432245 on Build GRCh38
rs746140198 Chr.20: 51432295-51432295 on Build GRCh38
rs770338559 Chr.20: 51432120-51432120 on Build GRCh38
rs762655231 Chr.20: 51432316-51432316 on Build GRCh38
rs749627820 Chr.20: 51432292-51432292 on Build GRCh38
rs373570203 Chr.20: 51432327-51432327 on Build GRCh38
rs6067777 Chr.20: 51432123-51432123 on Build GRCh38
rs773262587 Chr.20: 51432313-51432313 on Build GRCh38
rs368935350 Chr.20: 51432207-51432207 on Build GRCh38
rs754580402 Chr.20: 51432171-51432171 on Build GRCh38
rs773544975 Chr.20: 51432124-51432124 on Build GRCh38
rs200642234 Chr.20: 51432328-51432328 on Build GRCh38
rs372957875 Chr.20: 51432218-51432218 on Build GRCh38
rs749477538 Chr.20: 51432213-51432213 on Build GRCh38
rs149166265 Chr.20: 51432277-51432277 on Build GRCh38
rs376022750 Chr.20: 51432289-51432289 on Build GRCh38
rs201968807 Chr.20: 51432253-51432253 on Build GRCh38
rs763099970 Chr.20: 51432152-51432153 on Build GRCh38
rs1032159772 Chr.20: 51432268-51432268 on Build GRCh38
rs144500154 Chr.20: 51432331-51432331 on Build GRCh38
rs146886244 Chr.20: 51432149-51432149 on Build GRCh38
rs765068247 Chr.20: 51432154-51432154 on Build GRCh38
rs778996693 Chr.20: 51432294-51432294 on Build GRCh38
rs55980737 Chr.20: 51432150-51432150 on Build GRCh38
rs764980438 Chr.20: 51432251-51432251 on Build GRCh38
rs143217939 Chr.20: 51432287-51432287 on Build GRCh38
rs150039512 Chr.20: 51432219-51432219 on Build GRCh38
rs771002433 Chr.20: 51432142-51432142 on Build GRCh38
rs763124205 Chr.20: 51432136-51432136 on Build GRCh38
rs553983994 Chr.20: 51432166-51432166 on Build GRCh38
rs369906770 Chr.20: 51432119-51432119 on Build GRCh38
rs770250812 Chr.20: 51432182-51432182 on Build GRCh38
rs920197166 Chr.20: 51432127-51432127 on Build GRCh38
rs997266756 Chr.20: 51432203-51432203 on Build GRCh38
rs143896328 Chr.20: 51432144-51432144 on Build GRCh38
rs6067778 Chr.20: 51432212-51432212 on Build GRCh38
rs761782021 Chr.20: 51432248-51432248 on Build GRCh38
rs141596667 Chr.20: 51432311-51432311 on Build GRCh38
rs768952512 Chr.20: 51432154-51432155 on Build GRCh38
rs775565853 Chr.20: 51432308-51432308 on Build GRCh38
rs528842461 Chr.20: 51432322-51432322 on Build GRCh38
rs1042885456 Chr.20: 51432206-51432206 on Build GRCh38
rs778132625 Chr.20: 51432291-51432291 on Build GRCh38
rs773854579 Chr.20: 51432323-51432323 on Build GRCh38
rs373320619 Chr.20: 51432167-51432167 on Build GRCh38
rs748033725 Chr.20: 51432309-51432309 on Build GRCh38

View Full Product Details